Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153982574-153983165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB13
Entrez Gene ID
5872
Gene Name
RAB13, member RAS oncogene family
Gene Aliases
GIG4
UniGene
Hs.151536
Gene Chromosome Location
Chr.1: 153981617-153990644 on Build GRCh38
Transcripts
NM_001272038.1 NM_002870.3 NR_073553.1 XM_017001959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896483 c.418G>A p.A140T Chr.1: 153982597-153982597 on Build GRCh38
4988706 c.355C>A p.L119M Chr.1: 153982778-153982778 on Build GRCh38
6761260 c.378G>T p.E126D Chr.1: 153982755-153982755 on Build GRCh38
213612 c.380C>T p.A127V Chr.1: 153982753-153982753 on Build GRCh38
4576299 c.423A>G p.R141R Chr.1: 153982592-153982592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767421720 Chr.1: 153982690-153982690 on Build GRCh38
rs1049605926 Chr.1: 153982705-153982705 on Build GRCh38
rs201720331 Chr.1: 153982813-153982813 on Build GRCh38
rs748391066 Chr.1: 153982781-153982781 on Build GRCh38
rs549643334 Chr.1: 153982868-153982868 on Build GRCh38
rs1015204331 Chr.1: 153983028-153983028 on Build GRCh38
rs562321650 Chr.1: 153982655-153982655 on Build GRCh38
rs766351999 Chr.1: 153982812-153982812 on Build GRCh38
rs867456942 Chr.1: 153982898-153982898 on Build GRCh38
rs200896887 Chr.1: 153982723-153982723 on Build GRCh38
rs539299083 Chr.1: 153983041-153983041 on Build GRCh38
rs779539207 Chr.1: 153983026-153983026 on Build GRCh38
rs757844951 Chr.1: 153982724-153982724 on Build GRCh38
rs1015091372 Chr.1: 153983087-153983087 on Build GRCh38
rs1055619825 Chr.1: 153982989-153982989 on Build GRCh38
rs756924739 Chr.1: 153982714-153982714 on Build GRCh38
rs562089951 Chr.1: 153983078-153983078 on Build GRCh38
rs753632958 Chr.1: 153982851-153982851 on Build GRCh38
rs911161000 Chr.1: 153983126-153983127 on Build GRCh38
rs572015035 Chr.1: 153983039-153983039 on Build GRCh38
rs370214600 Chr.1: 153982617-153982617 on Build GRCh38
rs756977938 Chr.1: 153982854-153982854 on Build GRCh38
rs745309724 Chr.1: 153982737-153982737 on Build GRCh38
rs543780422 Chr.1: 153983062-153983062 on Build GRCh38
rs780668783 Chr.1: 153982725-153982725 on Build GRCh38
rs545276571 Chr.1: 153982640-153982640 on Build GRCh38
rs879571650 Chr.1: 153983125-153983126 on Build GRCh38
rs752569127 Chr.1: 153982698-153982698 on Build GRCh38
rs747451550 Chr.1: 153982755-153982755 on Build GRCh38
rs763159582 Chr.1: 153982801-153982801 on Build GRCh38
rs997658090 Chr.1: 153983044-153983044 on Build GRCh38
rs757413947 Chr.1: 153982793-153982793 on Build GRCh38
rs748930016 Chr.1: 153982982-153982982 on Build GRCh38
rs776941556 Chr.1: 153982778-153982778 on Build GRCh38
rs779771794 Chr.1: 153982981-153982981 on Build GRCh38
rs985398877 Chr.1: 153983126-153983126 on Build GRCh38
rs375247680 Chr.1: 153983137-153983138 on Build GRCh38
rs931260449 Chr.1: 153982688-153982688 on Build GRCh38
rs919608758 Chr.1: 153983108-153983108 on Build GRCh38
rs760656575 Chr.1: 153982817-153982817 on Build GRCh38
rs553922996 Chr.1: 153983035-153983035 on Build GRCh38
rs776890093 Chr.1: 153982624-153982624 on Build GRCh38
rs931014477 Chr.1: 153982998-153982998 on Build GRCh38
rs1048536473 Chr.1: 153982999-153982999 on Build GRCh38
rs750465602 Chr.1: 153982650-153982650 on Build GRCh38
rs533626473 Chr.1: 153982759-153982759 on Build GRCh38
rs894218542 Chr.1: 153982627-153982627 on Build GRCh38
rs370008084 Chr.1: 153983137-153983137 on Build GRCh38
rs568125087 Chr.1: 153982955-153982955 on Build GRCh38
rs183837316 Chr.1: 153983049-153983049 on Build GRCh38
rs930958433 Chr.1: 153983111-153983111 on Build GRCh38
rs377035618 Chr.1: 153982710-153982710 on Build GRCh38
rs191665541 Chr.1: 153982844-153982844 on Build GRCh38
rs1004277400 Chr.1: 153983025-153983025 on Build GRCh38
rs778582488 Chr.1: 153982716-153982716 on Build GRCh38
rs893782460 Chr.1: 153982926-153982926 on Build GRCh38
rs367560794 Chr.1: 153983016-153983016 on Build GRCh38
rs369273010 Chr.1: 153982855-153982855 on Build GRCh38
rs1022126848 Chr.1: 153983100-153983100 on Build GRCh38
rs768466241 Chr.1: 153982987-153982987 on Build GRCh38
rs188353397 Chr.1: 153982684-153982684 on Build GRCh38
rs776280609 Chr.1: 153983106-153983106 on Build GRCh38
rs772575766 Chr.1: 153982605-153982605 on Build GRCh38
rs367690717 Chr.1: 153982594-153982594 on Build GRCh38
rs565253301 Chr.1: 153982864-153982864 on Build GRCh38
rs541278504 Chr.1: 153983101-153983101 on Build GRCh38
rs894239833 Chr.1: 153982992-153982992 on Build GRCh38
rs769424595 Chr.1: 153982887-153982887 on Build GRCh38
rs886815647 Chr.1: 153983000-153983000 on Build GRCh38
rs547968945 Chr.1: 153982711-153982711 on Build GRCh38
rs200451111 Chr.1: 153982787-153982787 on Build GRCh38
rs372725515 Chr.1: 153982800-153982800 on Build GRCh38
rs574072676 Chr.1: 153983086-153983086 on Build GRCh38
rs953472273 Chr.1: 153983074-153983074 on Build GRCh38
rs945958075 Chr.1: 153982952-153982952 on Build GRCh38
rs773169319 Chr.1: 153982788-153982788 on Build GRCh38
rs575992062 Chr.1: 153983057-153983057 on Build GRCh38
rs529492772 Chr.1: 153982704-153982704 on Build GRCh38
rs372299956 Chr.1: 153982621-153982621 on Build GRCh38
rs976207581 Chr.1: 153982757-153982757 on Build GRCh38
rs564976582 Chr.1: 153982985-153982985 on Build GRCh38
rs902085428 Chr.1: 153983040-153983040 on Build GRCh38

View Full Product Details