Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 82916835-82917338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THAP9
Entrez Gene ID
79725
Gene Name
THAP domain containing 9
Gene Aliases
hTh9
UniGene
Hs.582050
Gene Chromosome Location
Chr.4: 82900650-82920283 on Build GRCh38
Transcripts
NM_001317776.1 NM_024672.5 XM_005263220.2 XM_011532246.2 XM_017008603.1 XM_017008604.1 XM_017008605.1 XM_017008606.1 XM_017008607.1 XM_017008608.1 XM_017008609.1 XM_017008610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5024450 c.800T>C p.V267A Chr.4: 82917012-82917012 on Build GRCh38
1267746 c.956C>T p.S319L Chr.4: 82917168-82917168 on Build GRCh38
5425643 c.897G>T p.L299F Chr.4: 82917109-82917109 on Build GRCh38
1058213 c.782C>A p.S261Y Chr.4: 82916994-82916994 on Build GRCh38
39154 c.846_860del15 p.S283_K287del Chr.4: 82917058-82917072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768660702 Chr.4: 82917270-82917280 on Build GRCh38
rs754062845 Chr.4: 82917157-82917157 on Build GRCh38
rs368120049 Chr.4: 82917046-82917046 on Build GRCh38
rs199536519 Chr.4: 82917070-82917070 on Build GRCh38
rs1047931024 Chr.4: 82917096-82917096 on Build GRCh38
rs749058717 Chr.4: 82917004-82917004 on Build GRCh38
rs747095049 Chr.4: 82917253-82917253 on Build GRCh38
rs764046504 Chr.4: 82917138-82917138 on Build GRCh38
rs764480070 Chr.4: 82916950-82916950 on Build GRCh38
rs778595696 Chr.4: 82917015-82917015 on Build GRCh38
rs1025694220 Chr.4: 82916864-82916864 on Build GRCh38
rs567428950 Chr.4: 82916908-82916908 on Build GRCh38
rs767923664 Chr.4: 82917111-82917111 on Build GRCh38
rs764871857 Chr.4: 82917043-82917043 on Build GRCh38
rs770618574 Chr.4: 82916988-82916988 on Build GRCh38
rs760906128 Chr.4: 82917131-82917131 on Build GRCh38
rs775815298 Chr.4: 82916893-82916893 on Build GRCh38
rs376013905 Chr.4: 82917206-82917206 on Build GRCh38
rs188609851 Chr.4: 82917047-82917047 on Build GRCh38
rs780499253 Chr.4: 82917228-82917229 on Build GRCh38
rs772630906 Chr.4: 82917296-82917296 on Build GRCh38
rs758965702 Chr.4: 82917229-82917229 on Build GRCh38
rs147886071 Chr.4: 82917132-82917132 on Build GRCh38
rs759019875 Chr.4: 82916942-82916942 on Build GRCh38
rs770480270 Chr.4: 82917208-82917208 on Build GRCh38
rs757622109 Chr.4: 82917055-82917055 on Build GRCh38
rs897945 Chr.4: 82917109-82917109 on Build GRCh38
rs750519999 Chr.4: 82917119-82917119 on Build GRCh38
rs1051655356 Chr.4: 82917243-82917243 on Build GRCh38
rs551600505 Chr.4: 82917016-82917016 on Build GRCh38
rs369859993 Chr.4: 82917197-82917198 on Build GRCh38
rs774613739 Chr.4: 82917095-82917095 on Build GRCh38
rs373501397 Chr.4: 82916881-82916881 on Build GRCh38
rs141560028 Chr.4: 82917186-82917186 on Build GRCh38
rs999999748 Chr.4: 82916910-82916910 on Build GRCh38
rs958272242 Chr.4: 82916998-82916998 on Build GRCh38
rs948599518 Chr.4: 82917086-82917086 on Build GRCh38
rs780706065 Chr.4: 82917213-82917213 on Build GRCh38
rs564924897 Chr.4: 82917029-82917029 on Build GRCh38
rs751537124 Chr.4: 82917308-82917308 on Build GRCh38
rs200386399 Chr.4: 82917006-82917006 on Build GRCh38
rs147028810 Chr.4: 82917201-82917201 on Build GRCh38
rs986964457 Chr.4: 82916891-82916891 on Build GRCh38
rs766684750 Chr.4: 82917156-82917156 on Build GRCh38
rs746491309 Chr.4: 82916940-82916940 on Build GRCh38
rs995994609 Chr.4: 82916863-82916867 on Build GRCh38
rs759301770 Chr.4: 82917037-82917037 on Build GRCh38
rs774721320 Chr.4: 82917275-82917275 on Build GRCh38
rs771905292 Chr.4: 82917018-82917018 on Build GRCh38
rs376098095 Chr.4: 82916969-82916969 on Build GRCh38
rs377556286 Chr.4: 82916916-82916916 on Build GRCh38
rs74789298 Chr.4: 82917203-82917203 on Build GRCh38
rs1026168041 Chr.4: 82917202-82917202 on Build GRCh38
rs779333217 Chr.4: 82917070-82917073 on Build GRCh38
rs780483417 Chr.4: 82916989-82916989 on Build GRCh38
rs866865367 Chr.4: 82917260-82917260 on Build GRCh38
rs151087610 Chr.4: 82916958-82916958 on Build GRCh38
rs778698755 Chr.4: 82917183-82917183 on Build GRCh38
rs941653909 Chr.4: 82917144-82917144 on Build GRCh38
rs772808524 Chr.4: 82916926-82916926 on Build GRCh38
rs762144579 Chr.4: 82917276-82917276 on Build GRCh38
rs757730324 Chr.4: 82916962-82916962 on Build GRCh38
rs745486010 Chr.4: 82917237-82917237 on Build GRCh38
rs201472958 Chr.4: 82917193-82917193 on Build GRCh38
rs746409313 Chr.4: 82917175-82917176 on Build GRCh38
rs776892304 Chr.4: 82917282-82917283 on Build GRCh38
rs369223139 Chr.4: 82917284-82917284 on Build GRCh38
rs1031639 Chr.4: 82917064-82917064 on Build GRCh38
rs773902967 Chr.4: 82917314-82917314 on Build GRCh38
rs369047725 Chr.4: 82916981-82916981 on Build GRCh38
rs180770321 Chr.4: 82917267-82917267 on Build GRCh38
rs755185508 Chr.4: 82917180-82917180 on Build GRCh38

View Full Product Details