Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174567702-174568230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WIPF1
Entrez Gene ID
7456
Gene Name
WAS/WASL interacting protein family member 1
Gene Aliases
PRPL-2, WAS2, WASPIP, WIP
UniGene
Hs.128067
Gene Chromosome Location
Chr.2: 174559574-174682899 on Build GRCh38
Transcripts
NM_001077269.1 NM_003387.4 XM_011511780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523849 c.1237G>A p.G413R Chr.2: 174567966-174567966 on Build GRCh38
5394976 c.1318G>A p.G440S Chr.2: 174567885-174567885 on Build GRCh38
5653233 c.1178G>A p.R393Q Chr.2: 174568025-174568025 on Build GRCh38
4523847 c.1237G>A p.G413R Chr.2: 174567966-174567966 on Build GRCh38
3407071 c.1284T>G p.P428P Chr.2: 174567919-174567919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866667559 Chr.2: 174567750-174567750 on Build GRCh38
rs765734549 Chr.2: 174567849-174567849 on Build GRCh38
rs71417493 Chr.2: 174568053-174568053 on Build GRCh38
rs564527991 Chr.2: 174568126-174568126 on Build GRCh38
rs981678700 Chr.2: 174567761-174567761 on Build GRCh38
rs201327694 Chr.2: 174568139-174568139 on Build GRCh38
rs4441436 Chr.2: 174568088-174568088 on Build GRCh38
rs76731102 Chr.2: 174567907-174567907 on Build GRCh38
rs773426790 Chr.2: 174568109-174568109 on Build GRCh38
rs970254579 Chr.2: 174567760-174567760 on Build GRCh38
rs938241940 Chr.2: 174568086-174568086 on Build GRCh38
rs371289527 Chr.2: 174567831-174567831 on Build GRCh38
rs777176983 Chr.2: 174567939-174567939 on Build GRCh38
rs571077444 Chr.2: 174567806-174567807 on Build GRCh38
rs747009480 Chr.2: 174567923-174567923 on Build GRCh38
rs908677157 Chr.2: 174567787-174567788 on Build GRCh38
rs367686097 Chr.2: 174567970-174567970 on Build GRCh38
rs750915536 Chr.2: 174567869-174567869 on Build GRCh38
rs77346261 Chr.2: 174567841-174567841 on Build GRCh38
rs761413428 Chr.2: 174568096-174568096 on Build GRCh38
rs761842397 Chr.2: 174567963-174567963 on Build GRCh38
rs760919514 Chr.2: 174567829-174567829 on Build GRCh38
rs954688051 Chr.2: 174568113-174568113 on Build GRCh38
rs986855403 Chr.2: 174567965-174567965 on Build GRCh38
rs921892999 Chr.2: 174568110-174568110 on Build GRCh38
rs749199595 Chr.2: 174567921-174567921 on Build GRCh38
rs755626909 Chr.2: 174568025-174568025 on Build GRCh38
rs148300861 Chr.2: 174567908-174567908 on Build GRCh38
rs1028287821 Chr.2: 174567937-174567937 on Build GRCh38
rs577852991 Chr.2: 174567976-174567976 on Build GRCh38
rs370232631 Chr.2: 174567825-174567825 on Build GRCh38
rs1053596413 Chr.2: 174568174-174568174 on Build GRCh38
rs778809828 Chr.2: 174568071-174568071 on Build GRCh38
rs147238026 Chr.2: 174568158-174568158 on Build GRCh38
rs1013296046 Chr.2: 174567872-174567872 on Build GRCh38
rs765045748 Chr.2: 174568038-174568038 on Build GRCh38
rs748781288 Chr.2: 174567985-174567985 on Build GRCh38
rs755377964 Chr.2: 174568014-174568014 on Build GRCh38
rs12052997 Chr.2: 174567890-174567890 on Build GRCh38
rs747335347 Chr.2: 174567887-174567887 on Build GRCh38
rs1044133629 Chr.2: 174568172-174568172 on Build GRCh38
rs776908211 Chr.2: 174567961-174567961 on Build GRCh38
rs371993338 Chr.2: 174567951-174567951 on Build GRCh38
rs147511624 Chr.2: 174567778-174567778 on Build GRCh38
rs974710774 Chr.2: 174567835-174567835 on Build GRCh38
rs145467227 Chr.2: 174567914-174567914 on Build GRCh38
rs769870479 Chr.2: 174568103-174568103 on Build GRCh38
rs879255352 Chr.2: 174567957-174567957 on Build GRCh38
rs748180254 Chr.2: 174568167-174568167 on Build GRCh38
rs142991283 Chr.2: 174567974-174567974 on Build GRCh38
rs771963360 Chr.2: 174567811-174567811 on Build GRCh38
rs377212270 Chr.2: 174568083-174568083 on Build GRCh38
rs143156793 Chr.2: 174568026-174568026 on Build GRCh38
rs771873177 Chr.2: 174568077-174568077 on Build GRCh38
rs753877257 Chr.2: 174567838-174567838 on Build GRCh38
rs763066609 Chr.2: 174568116-174568116 on Build GRCh38
rs949070351 Chr.2: 174568150-174568150 on Build GRCh38
rs756045105 Chr.2: 174567967-174567982 on Build GRCh38
rs878897138 Chr.2: 174568141-174568141 on Build GRCh38
rs763806469 Chr.2: 174568080-174568081 on Build GRCh38
rs752363182 Chr.2: 174568018-174568018 on Build GRCh38
rs767047241 Chr.2: 174568122-174568122 on Build GRCh38
rs751981976 Chr.2: 174568009-174568009 on Build GRCh38
rs41270199 Chr.2: 174568039-174568039 on Build GRCh38
rs34236584 Chr.2: 174567919-174567920 on Build GRCh38
rs569519657 Chr.2: 174567779-174567779 on Build GRCh38
rs939984536 Chr.2: 174568137-174568137 on Build GRCh38
rs769341820 Chr.2: 174567912-174567912 on Build GRCh38
rs745730470 Chr.2: 174568076-174568076 on Build GRCh38
rs1042079427 Chr.2: 174568156-174568156 on Build GRCh38
rs768818973 Chr.2: 174567953-174567953 on Build GRCh38
rs1051018084 Chr.2: 174567886-174567886 on Build GRCh38
rs561800755 Chr.2: 174568136-174568136 on Build GRCh38
rs914622631 Chr.2: 174567827-174567827 on Build GRCh38
rs543519687 Chr.2: 174568128-174568128 on Build GRCh38
rs200542640 Chr.2: 174567820-174567820 on Build GRCh38
rs932515850 Chr.2: 174567878-174567878 on Build GRCh38
rs780591761 Chr.2: 174567870-174567870 on Build GRCh38
rs921279221 Chr.2: 174567753-174567753 on Build GRCh38
rs896117953 Chr.2: 174568146-174568146 on Build GRCh38
rs71417494 Chr.2: 174568056-174568056 on Build GRCh38
rs762802902 Chr.2: 174567814-174567814 on Build GRCh38
rs754680076 Chr.2: 174567896-174567896 on Build GRCh38
rs1022616082 Chr.2: 174567755-174567755 on Build GRCh38
rs766924474 Chr.2: 174567999-174567999 on Build GRCh38
rs537157130 Chr.2: 174567782-174567782 on Build GRCh38
rs770870596 Chr.2: 174568070-174568070 on Build GRCh38
rs905539978 Chr.2: 174567986-174567986 on Build GRCh38
rs371524394 Chr.2: 174568079-174568079 on Build GRCh38
rs940131375 Chr.2: 174568004-174568004 on Build GRCh38
rs2288622 Chr.2: 174568127-174568127 on Build GRCh38

View Full Product Details