Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102717371-102717928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD9
Entrez Gene ID
84804
Gene Name
major facilitator superfamily domain containing 9
Gene Aliases
-
UniGene
Hs.590898
Gene Chromosome Location
Chr.2: 102713867-102736888 on Build GRCh38
Transcripts
NM_001322080.1 NM_001322081.1 NM_032718.4 NR_136173.1 NR_136174.1 XM_011512005.2 XM_011512006.2 XM_011512007.2

SNP Detail

SNP ID Chromosome Location
rs557706287 Chr.2: 102717607-102717607 on Build GRCh38
rs756262268 Chr.2: 102717465-102717465 on Build GRCh38
rs1013228340 Chr.2: 102717765-102717765 on Build GRCh38
rs780353989 Chr.2: 102717581-102717581 on Build GRCh38
rs925765567 Chr.2: 102717587-102717587 on Build GRCh38
rs1043607039 Chr.2: 102717646-102717646 on Build GRCh38
rs535267486 Chr.2: 102717436-102717436 on Build GRCh38
rs13405814 Chr.2: 102717520-102717520 on Build GRCh38
rs976315236 Chr.2: 102717567-102717567 on Build GRCh38
rs774449514 Chr.2: 102717606-102717606 on Build GRCh38
rs1033354980 Chr.2: 102717433-102717433 on Build GRCh38
rs750516891 Chr.2: 102717396-102717396 on Build GRCh38
rs933360578 Chr.2: 102717602-102717602 on Build GRCh38
rs903730157 Chr.2: 102717699-102717699 on Build GRCh38
rs866303650 Chr.2: 102717498-102717498 on Build GRCh38
rs945185315 Chr.2: 102717628-102717628 on Build GRCh38
rs555533742 Chr.2: 102717855-102717855 on Build GRCh38
rs937952597 Chr.2: 102717719-102717719 on Build GRCh38
rs374088161 Chr.2: 102717710-102717711 on Build GRCh38
rs576013129 Chr.2: 102717805-102717805 on Build GRCh38
rs1030722392 Chr.2: 102717869-102717869 on Build GRCh38
rs975032920 Chr.2: 102717861-102717861 on Build GRCh38
rs997608000 Chr.2: 102717813-102717813 on Build GRCh38
rs573200756 Chr.2: 102717664-102717664 on Build GRCh38
rs1026006128 Chr.2: 102717766-102717766 on Build GRCh38
rs752385172 Chr.2: 102717780-102717780 on Build GRCh38
rs374089170 Chr.2: 102717832-102717832 on Build GRCh38
rs965028816 Chr.2: 102717547-102717547 on Build GRCh38
rs1029236118 Chr.2: 102717826-102717826 on Build GRCh38
rs1045680901 Chr.2: 102717622-102717622 on Build GRCh38
rs1027746927 Chr.2: 102717437-102717437 on Build GRCh38
rs905881055 Chr.2: 102717416-102717416 on Build GRCh38
rs984951522 Chr.2: 102717517-102717517 on Build GRCh38
rs1055170620 Chr.2: 102717761-102717761 on Build GRCh38
rs540633932 Chr.2: 102717674-102717674 on Build GRCh38
rs374039281 Chr.2: 102717584-102717584 on Build GRCh38
rs977266533 Chr.2: 102717715-102717715 on Build GRCh38
rs949841440 Chr.2: 102717612-102717612 on Build GRCh38
rs72524311 Chr.2: 102717688-102717688 on Build GRCh38
rs1006341681 Chr.2: 102717804-102717804 on Build GRCh38
rs950866649 Chr.2: 102717503-102717503 on Build GRCh38
rs1013303074 Chr.2: 102717480-102717480 on Build GRCh38
rs116071755 Chr.2: 102717478-102717478 on Build GRCh38
rs761971846 Chr.2: 102717763-102717763 on Build GRCh38
rs988878625 Chr.2: 102717605-102717605 on Build GRCh38
rs570377584 Chr.2: 102717580-102717580 on Build GRCh38
rs945900745 Chr.2: 102717705-102717705 on Build GRCh38
rs543045176 Chr.2: 102717886-102717886 on Build GRCh38
rs905741065 Chr.2: 102717623-102717623 on Build GRCh38
rs866791411 Chr.2: 102717723-102717723 on Build GRCh38

View Full Product Details