Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50795086-50795603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS4
Entrez Gene ID
55653
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
CNOL
UniGene
Hs.381178
Gene Chromosome Location
Chr.20: 50794894-50877736 on Build GRCh38
Transcripts
NM_001010974.2 NM_017843.4 NM_198799.3 XM_011528886.2 XM_011528887.2 XM_017027932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381209 c.130A>G p.S44G Chr.20: 50795123-50795123 on Build GRCh38
5631508 c.147C>T p.L49L Chr.20: 50795140-50795140 on Build GRCh38
3720966 c.168G>T p.E56D Chr.20: 50795161-50795161 on Build GRCh38
3720965 c.168G>T p.E56D Chr.20: 50795161-50795161 on Build GRCh38
5631510 c.147C>T p.L49L Chr.20: 50795140-50795140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990021608 Chr.20: 50795506-50795506 on Build GRCh38
rs530011560 Chr.20: 50795201-50795201 on Build GRCh38
rs960689189 Chr.20: 50795495-50795495 on Build GRCh38
rs999515018 Chr.20: 50795440-50795440 on Build GRCh38
rs1022348898 Chr.20: 50795417-50795417 on Build GRCh38
rs1040416683 Chr.20: 50795390-50795390 on Build GRCh38
rs1050652632 Chr.20: 50795197-50795197 on Build GRCh38
rs758469699 Chr.20: 50795120-50795120 on Build GRCh38
rs945003448 Chr.20: 50795578-50795578 on Build GRCh38
rs919692424 Chr.20: 50795567-50795567 on Build GRCh38
rs973798633 Chr.20: 50795341-50795341 on Build GRCh38
rs980545936 Chr.20: 50795293-50795293 on Build GRCh38
rs892451081 Chr.20: 50795401-50795401 on Build GRCh38
rs908194150 Chr.20: 50795330-50795330 on Build GRCh38
rs958317815 Chr.20: 50795481-50795481 on Build GRCh38
rs767425001 Chr.20: 50795252-50795252 on Build GRCh38
rs978293923 Chr.20: 50795264-50795264 on Build GRCh38
rs568068151 Chr.20: 50795416-50795416 on Build GRCh38
rs1019332545 Chr.20: 50795215-50795215 on Build GRCh38
rs927728696 Chr.20: 50795552-50795552 on Build GRCh38
rs903925215 Chr.20: 50795426-50795426 on Build GRCh38
rs1033895297 Chr.20: 50795467-50795467 on Build GRCh38
rs924223342 Chr.20: 50795318-50795318 on Build GRCh38
rs1008915954 Chr.20: 50795205-50795205 on Build GRCh38
rs1022109863 Chr.20: 50795269-50795269 on Build GRCh38
rs770828267 Chr.20: 50795162-50795162 on Build GRCh38
rs756148826 Chr.20: 50795347-50795347 on Build GRCh38
rs986351778 Chr.20: 50795572-50795572 on Build GRCh38
rs967843557 Chr.20: 50795280-50795280 on Build GRCh38
rs1019583594 Chr.20: 50795212-50795212 on Build GRCh38
rs185600142 Chr.20: 50795582-50795582 on Build GRCh38
rs773750758 Chr.20: 50795554-50795555 on Build GRCh38
rs867562161 Chr.20: 50795535-50795535 on Build GRCh38
rs116207530 Chr.20: 50795568-50795568 on Build GRCh38
rs372701999 Chr.20: 50795363-50795363 on Build GRCh38
rs527771676 Chr.20: 50795326-50795326 on Build GRCh38
rs551994531 Chr.20: 50795344-50795344 on Build GRCh38
rs924128505 Chr.20: 50795105-50795105 on Build GRCh38
rs1033619991 Chr.20: 50795305-50795305 on Build GRCh38
rs944947604 Chr.20: 50795389-50795389 on Build GRCh38
rs995038358 Chr.20: 50795376-50795376 on Build GRCh38
rs1014742939 Chr.20: 50795170-50795170 on Build GRCh38
rs965574691 Chr.20: 50795229-50795229 on Build GRCh38
rs954235019 Chr.20: 50795157-50795168 on Build GRCh38
rs553712948 Chr.20: 50795504-50795504 on Build GRCh38
rs866274827 Chr.20: 50795556-50795556 on Build GRCh38
rs973702442 Chr.20: 50795566-50795566 on Build GRCh38
rs754141391 Chr.20: 50795488-50795488 on Build GRCh38
rs764248920 Chr.20: 50795428-50795428 on Build GRCh38
rs757394402 Chr.20: 50795520-50795520 on Build GRCh38
rs570311641 Chr.20: 50795355-50795355 on Build GRCh38
rs548688125 Chr.20: 50795288-50795288 on Build GRCh38
rs780045321 Chr.20: 50795126-50795126 on Build GRCh38
rs150892114 Chr.20: 50795552-50795553 on Build GRCh38
rs756921768 Chr.20: 50795176-50795176 on Build GRCh38
rs954157113 Chr.20: 50795198-50795198 on Build GRCh38
rs2272962 Chr.20: 50795161-50795161 on Build GRCh38
rs76593448 Chr.20: 50795325-50795325 on Build GRCh38
rs375879128 Chr.20: 50795558-50795558 on Build GRCh38
rs117388674 Chr.20: 50795152-50795152 on Build GRCh38
rs747248326 Chr.20: 50795143-50795143 on Build GRCh38
rs898981583 Chr.20: 50795182-50795182 on Build GRCh38
rs777462572 Chr.20: 50795158-50795158 on Build GRCh38
rs920474297 Chr.20: 50795564-50795564 on Build GRCh38
rs752837883 Chr.20: 50795337-50795337 on Build GRCh38
rs192529506 Chr.20: 50795441-50795441 on Build GRCh38
rs956848860 Chr.20: 50795226-50795226 on Build GRCh38
rs910709399 Chr.20: 50795577-50795577 on Build GRCh38
rs994713212 Chr.20: 50795183-50795183 on Build GRCh38
rs936862957 Chr.20: 50795110-50795110 on Build GRCh38
rs555999847 Chr.20: 50795379-50795379 on Build GRCh38
rs985619400 Chr.20: 50795507-50795522 on Build GRCh38

View Full Product Details