Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 109425908-109426484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf87
Entrez Gene ID
399947
Gene Name
chromosome 11 open reading frame 87
Gene Aliases
LOH11CR1A, NEURIM1
UniGene
Hs.172982
Gene Chromosome Location
Chr.11: 109422120-109433003 on Build GRCh38
Transcripts
NM_207645.3 XM_011542817.2 XM_011542818.2

SNP Detail

SNP ID Chromosome Location
rs939777898 Chr.11: 109426388-109426388 on Build GRCh38
rs941025894 Chr.11: 109426075-109426075 on Build GRCh38
rs191545707 Chr.11: 109425967-109425967 on Build GRCh38
rs1038098602 Chr.11: 109426403-109426403 on Build GRCh38
rs956766087 Chr.11: 109425987-109425987 on Build GRCh38
rs374009971 Chr.11: 109425989-109425989 on Build GRCh38
rs151033061 Chr.11: 109426384-109426384 on Build GRCh38
rs1018512899 Chr.11: 109425933-109425933 on Build GRCh38
rs1020338459 Chr.11: 109426277-109426277 on Build GRCh38
rs934128186 Chr.11: 109426082-109426082 on Build GRCh38
rs909553656 Chr.11: 109426057-109426057 on Build GRCh38
rs551795932 Chr.11: 109426404-109426404 on Build GRCh38
rs867011274 Chr.11: 109426256-109426256 on Build GRCh38
rs566912003 Chr.11: 109426418-109426418 on Build GRCh38
rs964253512 Chr.11: 109426376-109426376 on Build GRCh38
rs971225661 Chr.11: 109426224-109426224 on Build GRCh38
rs1003899685 Chr.11: 109426114-109426114 on Build GRCh38
rs183732200 Chr.11: 109426104-109426104 on Build GRCh38
rs934242637 Chr.11: 109426377-109426377 on Build GRCh38
rs539405100 Chr.11: 109426398-109426398 on Build GRCh38
rs988216424 Chr.11: 109426259-109426259 on Build GRCh38
rs1018518101 Chr.11: 109426372-109426372 on Build GRCh38
rs140187758 Chr.11: 109426231-109426231 on Build GRCh38
rs939200818 Chr.11: 109426198-109426198 on Build GRCh38
rs560730508 Chr.11: 109426278-109426278 on Build GRCh38
rs530603291 Chr.11: 109426038-109426038 on Build GRCh38
rs904548982 Chr.11: 109426281-109426282 on Build GRCh38
rs868245813 Chr.11: 109426095-109426095 on Build GRCh38
rs1013267945 Chr.11: 109426185-109426185 on Build GRCh38
rs562622604 Chr.11: 109426143-109426143 on Build GRCh38
rs921552685 Chr.11: 109426349-109426349 on Build GRCh38
rs971194809 Chr.11: 109425950-109425950 on Build GRCh38
rs559764380 Chr.11: 109426238-109426238 on Build GRCh38
rs921437341 Chr.11: 109426046-109426046 on Build GRCh38
rs976079388 Chr.11: 109426333-109426333 on Build GRCh38
rs539837296 Chr.11: 109425944-109425944 on Build GRCh38
rs531555173 Chr.11: 109426289-109426289 on Build GRCh38
rs1038473369 Chr.11: 109426153-109426153 on Build GRCh38
rs534479814 Chr.11: 109426458-109426458 on Build GRCh38
rs1022738050 Chr.11: 109426302-109426302 on Build GRCh38
rs144935313 Chr.11: 109425981-109425981 on Build GRCh38
rs544612498 Chr.11: 109426232-109426232 on Build GRCh38
rs898132354 Chr.11: 109426158-109426158 on Build GRCh38
rs879124792 Chr.11: 109426347-109426347 on Build GRCh38
rs1010754901 Chr.11: 109426250-109426250 on Build GRCh38
rs1000180961 Chr.11: 109426363-109426363 on Build GRCh38
rs982968669 Chr.11: 109426421-109426421 on Build GRCh38
rs147524530 Chr.11: 109425992-109425992 on Build GRCh38
rs142836310 Chr.11: 109426378-109426378 on Build GRCh38
rs1039935157 Chr.11: 109426021-109426021 on Build GRCh38
rs1022840102 Chr.11: 109426199-109426199 on Build GRCh38
rs982194198 Chr.11: 109425956-109425956 on Build GRCh38
rs80246075 Chr.11: 109425988-109425988 on Build GRCh38
rs527574103 Chr.11: 109426240-109426240 on Build GRCh38
rs968038453 Chr.11: 109426312-109426312 on Build GRCh38
rs11213073 Chr.11: 109426249-109426249 on Build GRCh38
rs982531421 Chr.11: 109426040-109426040 on Build GRCh38
rs1002739539 Chr.11: 109426228-109426228 on Build GRCh38
rs779623022 Chr.11: 109426244-109426244 on Build GRCh38
rs375510942 Chr.11: 109426113-109426113 on Build GRCh38
rs751995406 Chr.11: 109426059-109426060 on Build GRCh38
rs868353532 Chr.11: 109425978-109425978 on Build GRCh38

View Full Product Details