Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57633501-57634001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4562490 c.835G>A p.E279K Chr.15: 57633736-57633736 on Build GRCh38
963301 c.923G>A p.R308H Chr.15: 57633731-57633731 on Build GRCh38
3887246 c.808G>A p.E270K Chr.15: 57633616-57633616 on Build GRCh38
7208861 c.860C>G p.A287G Chr.15: 57633668-57633668 on Build GRCh38
5555209 c.898G>A p.G300R Chr.15: 57633706-57633706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929189993 Chr.15: 57633915-57633915 on Build GRCh38
rs546495468 Chr.15: 57633833-57633833 on Build GRCh38
rs766046051 Chr.15: 57633655-57633655 on Build GRCh38
rs771663802 Chr.15: 57633705-57633705 on Build GRCh38
rs151226893 Chr.15: 57633754-57633754 on Build GRCh38
rs779272418 Chr.15: 57633734-57633734 on Build GRCh38
rs370661037 Chr.15: 57633723-57633723 on Build GRCh38
rs144667495 Chr.15: 57633731-57633731 on Build GRCh38
rs745534187 Chr.15: 57633703-57633703 on Build GRCh38
rs752135885 Chr.15: 57633823-57633823 on Build GRCh38
rs539136507 Chr.15: 57633631-57633631 on Build GRCh38
rs771575962 Chr.15: 57633637-57633637 on Build GRCh38
rs138772571 Chr.15: 57633661-57633661 on Build GRCh38
rs148672365 Chr.15: 57633675-57633675 on Build GRCh38
rs775013553 Chr.15: 57633706-57633706 on Build GRCh38
rs775400978 Chr.15: 57633556-57633556 on Build GRCh38
rs765127132 Chr.15: 57633717-57633717 on Build GRCh38
rs79326877 Chr.15: 57633920-57633920 on Build GRCh38
rs924244490 Chr.15: 57633815-57633815 on Build GRCh38
rs748180170 Chr.15: 57633585-57633585 on Build GRCh38
rs780463115 Chr.15: 57633582-57633582 on Build GRCh38
rs373898639 Chr.15: 57633620-57633620 on Build GRCh38
rs546156676 Chr.15: 57633726-57633726 on Build GRCh38
rs746193755 Chr.15: 57633737-57633737 on Build GRCh38
rs755165474 Chr.15: 57633604-57633604 on Build GRCh38
rs774005642 Chr.15: 57633627-57633627 on Build GRCh38
rs529210276 Chr.15: 57633872-57633872 on Build GRCh38
rs199506323 Chr.15: 57633571-57633571 on Build GRCh38
rs975627512 Chr.15: 57633592-57633592 on Build GRCh38
rs201828277 Chr.15: 57633626-57633626 on Build GRCh38
rs150236562 Chr.15: 57633855-57633855 on Build GRCh38
rs752669069 Chr.15: 57633650-57633650 on Build GRCh38
rs186008498 Chr.15: 57633641-57633641 on Build GRCh38
rs140373684 Chr.15: 57633783-57633783 on Build GRCh38
rs570667256 Chr.15: 57633538-57633538 on Build GRCh38
rs760392279 Chr.15: 57633639-57633639 on Build GRCh38
rs191608007 Chr.15: 57633860-57633860 on Build GRCh38
rs866088824 Chr.15: 57633658-57633658 on Build GRCh38
rs776140348 Chr.15: 57633766-57633766 on Build GRCh38
rs564106023 Chr.15: 57633809-57633809 on Build GRCh38
rs763965444 Chr.15: 57633570-57633570 on Build GRCh38
rs969359759 Chr.15: 57633808-57633808 on Build GRCh38
rs541779682 Chr.15: 57633774-57633774 on Build GRCh38
rs948560675 Chr.15: 57633645-57633645 on Build GRCh38
rs1041101381 Chr.15: 57633948-57633948 on Build GRCh38
rs142159417 Chr.15: 57633684-57633684 on Build GRCh38
rs757704684 Chr.15: 57633733-57633733 on Build GRCh38
rs756208577 Chr.15: 57633730-57633730 on Build GRCh38
rs200890662 Chr.15: 57633779-57633779 on Build GRCh38
rs758679982 Chr.15: 57633580-57633580 on Build GRCh38
rs768117859 Chr.15: 57633764-57633764 on Build GRCh38
rs770577032 Chr.15: 57633587-57633587 on Build GRCh38
rs577399655 Chr.15: 57633712-57633712 on Build GRCh38
rs1008233495 Chr.15: 57633933-57633933 on Build GRCh38
rs767003871 Chr.15: 57633565-57633565 on Build GRCh38
rs761805041 Chr.15: 57633714-57633714 on Build GRCh38
rs951531514 Chr.15: 57633545-57633545 on Build GRCh38
rs61997919 Chr.15: 57633853-57633853 on Build GRCh38
rs760464970 Chr.15: 57633569-57633569 on Build GRCh38
rs749658073 Chr.15: 57633617-57633617 on Build GRCh38
rs920461497 Chr.15: 57633905-57633905 on Build GRCh38
rs79247688 Chr.15: 57633855-57633855 on Build GRCh38
rs146453491 Chr.15: 57633609-57633609 on Build GRCh38
rs373998282 Chr.15: 57633600-57633600 on Build GRCh38
rs778218288 Chr.15: 57633612-57633612 on Build GRCh38
rs16977629 Chr.15: 57633638-57633638 on Build GRCh38
rs748691063 Chr.15: 57633608-57633608 on Build GRCh38
rs759187975 Chr.15: 57633632-57633632 on Build GRCh38
rs746542048 Chr.15: 57633711-57633711 on Build GRCh38
rs764443252 Chr.15: 57633952-57633952 on Build GRCh38
rs1019769534 Chr.15: 57633557-57633557 on Build GRCh38
rs767713926 Chr.15: 57633648-57633648 on Build GRCh38
rs550455447 Chr.15: 57633527-57633527 on Build GRCh38
rs751896236 Chr.15: 57633643-57633643 on Build GRCh38
rs1047651898 Chr.15: 57633918-57633918 on Build GRCh38
rs779186470 Chr.15: 57633695-57633695 on Build GRCh38
rs752701091 Chr.15: 57633567-57633567 on Build GRCh38
rs35565683 Chr.15: 57633785-57633786 on Build GRCh38
rs11637944 Chr.15: 57633792-57633792 on Build GRCh38
rs548999050 Chr.15: 57633876-57633876 on Build GRCh38
rs758959774 Chr.15: 57633895-57633895 on Build GRCh38
rs200396643 Chr.15: 57633646-57633646 on Build GRCh38
rs568909825 Chr.15: 57633881-57633881 on Build GRCh38
rs200587063 Chr.15: 57633672-57633672 on Build GRCh38
rs781415700 Chr.15: 57633605-57633605 on Build GRCh38
rs773080564 Chr.15: 57633719-57633719 on Build GRCh38
rs372234545 Chr.15: 57633753-57633753 on Build GRCh38
rs533122419 Chr.15: 57633830-57633830 on Build GRCh38
rs374320645 Chr.15: 57633749-57633749 on Build GRCh38
rs761560230 Chr.15: 57633640-57633640 on Build GRCh38
rs1037483603 Chr.15: 57633904-57633904 on Build GRCh38
rs367589212 Chr.15: 57633786-57633786 on Build GRCh38
rs533062360 Chr.15: 57633539-57633539 on Build GRCh38

View Full Product Details