Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48518711-48519211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFKFB4
Entrez Gene ID
5210
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4
Gene Aliases
-
UniGene
Hs.476217
Gene Chromosome Location
Chr.3: 48517684-48562383 on Build GRCh38
Transcripts
NM_001317134.1 NM_001317135.1 NM_001317136.1 NM_001317137.1 NM_001317138.1 NM_004567.3 XM_011533829.2 XM_017006614.1 XM_017006615.1 XM_017006616.1 XM_017006617.1

SNP Detail

SNP ID Chromosome Location
rs886334387 Chr.3: 48518914-48518914 on Build GRCh38
rs1003891973 Chr.3: 48518916-48518916 on Build GRCh38
rs1000996740 Chr.3: 48519029-48519029 on Build GRCh38
rs766107948 Chr.3: 48519159-48519159 on Build GRCh38
rs1002215952 Chr.3: 48518736-48518736 on Build GRCh38
rs754440565 Chr.3: 48519189-48519189 on Build GRCh38
rs1010087456 Chr.3: 48518771-48518771 on Build GRCh38
rs539637030 Chr.3: 48519112-48519112 on Build GRCh38
rs914908226 Chr.3: 48518819-48518819 on Build GRCh38
rs1021022478 Chr.3: 48518776-48518776 on Build GRCh38
rs1035054428 Chr.3: 48518741-48518741 on Build GRCh38
rs941862959 Chr.3: 48518868-48518868 on Build GRCh38
rs1017761354 Chr.3: 48519054-48519054 on Build GRCh38
rs189734105 Chr.3: 48518978-48518978 on Build GRCh38
rs772263559 Chr.3: 48518818-48518818 on Build GRCh38
rs959141081 Chr.3: 48518799-48518799 on Build GRCh38
rs899247953 Chr.3: 48518891-48518891 on Build GRCh38
rs1034853142 Chr.3: 48518957-48518957 on Build GRCh38
rs1039173497 Chr.3: 48518879-48518879 on Build GRCh38
rs765075957 Chr.3: 48519060-48519060 on Build GRCh38
rs968632010 Chr.3: 48518777-48518777 on Build GRCh38
rs977706076 Chr.3: 48519135-48519135 on Build GRCh38
rs943457783 Chr.3: 48518860-48518860 on Build GRCh38
rs553738467 Chr.3: 48518840-48518840 on Build GRCh38
rs531213070 Chr.3: 48518747-48518747 on Build GRCh38
rs2267846 Chr.3: 48518906-48518906 on Build GRCh38
rs780587671 Chr.3: 48518850-48518850 on Build GRCh38
rs13094578 Chr.3: 48518798-48518798 on Build GRCh38
rs557728093 Chr.3: 48519113-48519113 on Build GRCh38
rs994805346 Chr.3: 48518905-48518905 on Build GRCh38
rs968119889 Chr.3: 48518994-48518994 on Build GRCh38
rs964845698 Chr.3: 48519080-48519080 on Build GRCh38
rs557969678 Chr.3: 48519163-48519163 on Build GRCh38
rs534280420 Chr.3: 48518857-48518857 on Build GRCh38
rs1044220327 Chr.3: 48518885-48518885 on Build GRCh38
rs934926708 Chr.3: 48518849-48518849 on Build GRCh38
rs924838505 Chr.3: 48518841-48518841 on Build GRCh38
rs905688108 Chr.3: 48518898-48518898 on Build GRCh38
rs766953484 Chr.3: 48518746-48518746 on Build GRCh38
rs946349044 Chr.3: 48518821-48518821 on Build GRCh38
rs556718960 Chr.3: 48519003-48519003 on Build GRCh38
rs577075863 Chr.3: 48519093-48519093 on Build GRCh38
rs923627246 Chr.3: 48519160-48519160 on Build GRCh38
rs148233066 Chr.3: 48519081-48519081 on Build GRCh38
rs552566010 Chr.3: 48518773-48518773 on Build GRCh38
rs182711256 Chr.3: 48518984-48518984 on Build GRCh38
rs1057052020 Chr.3: 48518910-48518910 on Build GRCh38

View Full Product Details