Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71157806-71158317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN8
Entrez Gene ID
7103
Gene Name
tetraspanin 8
Gene Aliases
CO-029, TM4SF3
UniGene
Hs.170563
Gene Chromosome Location
Chr.12: 71125093-71157999 on Build GRCh38
Transcripts
NM_004616.2 XM_006719583.3 XM_017019913.1

SNP Detail

SNP ID Chromosome Location
rs1047785102 Chr.12: 71157850-71157850 on Build GRCh38
rs929991563 Chr.12: 71158146-71158146 on Build GRCh38
rs11834208 Chr.12: 71157866-71157866 on Build GRCh38
rs1008652087 Chr.12: 71158059-71158059 on Build GRCh38
rs917246891 Chr.12: 71158048-71158048 on Build GRCh38
rs982679655 Chr.12: 71158167-71158167 on Build GRCh38
rs964608717 Chr.12: 71158179-71158179 on Build GRCh38
rs540321122 Chr.12: 71158229-71158229 on Build GRCh38
rs535287160 Chr.12: 71158197-71158197 on Build GRCh38
rs906034421 Chr.12: 71157940-71157940 on Build GRCh38
rs572966511 Chr.12: 71158007-71158007 on Build GRCh38
rs776939225 Chr.12: 71158009-71158009 on Build GRCh38
rs1005824336 Chr.12: 71157872-71157872 on Build GRCh38
rs527364257 Chr.12: 71157894-71157894 on Build GRCh38
rs1039560200 Chr.12: 71157899-71157899 on Build GRCh38
rs899544786 Chr.12: 71157900-71157900 on Build GRCh38
rs140008352 Chr.12: 71157829-71157829 on Build GRCh38
rs979310346 Chr.12: 71158006-71158006 on Build GRCh38
rs544601984 Chr.12: 71158289-71158289 on Build GRCh38
rs191910363 Chr.12: 71157897-71157897 on Build GRCh38
rs747455385 Chr.12: 71157992-71157992 on Build GRCh38
rs780426781 Chr.12: 71157844-71157844 on Build GRCh38
rs769199953 Chr.12: 71158237-71158237 on Build GRCh38
rs1044509833 Chr.12: 71157893-71157893 on Build GRCh38
rs956405693 Chr.12: 71157973-71157973 on Build GRCh38
rs556070688 Chr.12: 71158024-71158024 on Build GRCh38
rs553650516 Chr.12: 71158201-71158201 on Build GRCh38
rs997891976 Chr.12: 71157931-71157931 on Build GRCh38
rs570308846 Chr.12: 71157911-71157911 on Build GRCh38
rs183235624 Chr.12: 71158099-71158099 on Build GRCh38
rs1029280397 Chr.12: 71157943-71157943 on Build GRCh38
rs950099350 Chr.12: 71158263-71158263 on Build GRCh38
rs950151251 Chr.12: 71157867-71157867 on Build GRCh38
rs189140545 Chr.12: 71158270-71158270 on Build GRCh38
rs899701181 Chr.12: 71158252-71158252 on Build GRCh38
rs954408627 Chr.12: 71158038-71158038 on Build GRCh38
rs983388666 Chr.12: 71158228-71158228 on Build GRCh38
rs114719636 Chr.12: 71158226-71158226 on Build GRCh38
rs1017850512 Chr.12: 71158148-71158148 on Build GRCh38
rs1010430426 Chr.12: 71157979-71157979 on Build GRCh38
rs114123347 Chr.12: 71157996-71157996 on Build GRCh38
rs893451263 Chr.12: 71157980-71157980 on Build GRCh38
rs147198760 Chr.12: 71158284-71158284 on Build GRCh38
rs941302531 Chr.12: 71158207-71158207 on Build GRCh38

View Full Product Details