Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103337731-103338262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5
Entrez Gene ID
1983
Gene Name
eukaryotic translation initiation factor 5
Gene Aliases
EIF-5, EIF-5A
UniGene
Hs.433702
Gene Chromosome Location
Chr.14: 103334002-103345025 on Build GRCh38
Transcripts
NM_001969.4 NM_183004.4 NR_002964.1

Gene Symbol
SNORA28
Entrez Gene ID
677811
Gene Name
small nucleolar RNA, H/ACA box 28
Gene Aliases
ACA28
UniGene
Hs.658237
Gene Chromosome Location
Chr.14: 103337849-103337974 on Build GRCh38
Transcripts
NM_001969.4 NM_183004.4 NR_002964.1

SNP Detail

SNP ID Chromosome Location
rs550449250 Chr.14: 103337817-103337817 on Build GRCh38
rs200543633 Chr.14: 103337949-103337949 on Build GRCh38
rs375202609 Chr.14: 103337824-103337824 on Build GRCh38
rs933660286 Chr.14: 103338166-103338166 on Build GRCh38
rs899555053 Chr.14: 103338058-103338058 on Build GRCh38
rs201174936 Chr.14: 103337954-103337954 on Build GRCh38
rs768231628 Chr.14: 103337944-103337944 on Build GRCh38
rs182987635 Chr.14: 103337828-103337828 on Build GRCh38
rs764702644 Chr.14: 103338081-103338082 on Build GRCh38
rs945852588 Chr.14: 103337774-103337774 on Build GRCh38
rs996790371 Chr.14: 103337882-103337882 on Build GRCh38
rs560125919 Chr.14: 103337778-103337778 on Build GRCh38
rs762186210 Chr.14: 103337966-103337966 on Build GRCh38
rs191428158 Chr.14: 103337939-103337939 on Build GRCh38
rs774027834 Chr.14: 103337884-103337884 on Build GRCh38
rs749113819 Chr.14: 103338012-103338013 on Build GRCh38
rs574069166 Chr.14: 103338086-103338086 on Build GRCh38
rs975069703 Chr.14: 103338006-103338006 on Build GRCh38
rs746867235 Chr.14: 103337962-103337963 on Build GRCh38
rs967620504 Chr.14: 103337974-103337974 on Build GRCh38
rs760457499 Chr.14: 103337953-103337953 on Build GRCh38
rs369452409 Chr.14: 103337920-103337920 on Build GRCh38
rs199848523 Chr.14: 103337971-103337971 on Build GRCh38
rs780068905 Chr.14: 103337885-103337886 on Build GRCh38
rs940185170 Chr.14: 103338179-103338179 on Build GRCh38
rs757719588 Chr.14: 103337831-103337831 on Build GRCh38
rs759352953 Chr.14: 103337955-103337955 on Build GRCh38
rs781074324 Chr.14: 103338118-103338119 on Build GRCh38
rs941286719 Chr.14: 103338185-103338185 on Build GRCh38
rs920944514 Chr.14: 103338232-103338232 on Build GRCh38
rs770369359 Chr.14: 103337837-103337837 on Build GRCh38
rs955097211 Chr.14: 103337892-103337892 on Build GRCh38
rs761401525 Chr.14: 103337887-103337887 on Build GRCh38
rs992684732 Chr.14: 103337765-103337765 on Build GRCh38
rs777640006 Chr.14: 103337999-103338003 on Build GRCh38
rs769738411 Chr.14: 103337972-103337973 on Build GRCh38
rs760617003 Chr.14: 103337903-103337903 on Build GRCh38
rs756475920 Chr.14: 103337989-103337989 on Build GRCh38
rs754397117 Chr.14: 103337994-103337994 on Build GRCh38
rs752762030 Chr.14: 103337803-103337803 on Build GRCh38
rs185024481 Chr.14: 103337792-103337792 on Build GRCh38
rs774936961 Chr.14: 103337823-103337823 on Build GRCh38
rs1002202941 Chr.14: 103338109-103338109 on Build GRCh38
rs386780954 Chr.14: 103337792-103337794 on Build GRCh38
rs180723198 Chr.14: 103337770-103337770 on Build GRCh38
rs977173283 Chr.14: 103337975-103337975 on Build GRCh38
rs2274191 Chr.14: 103338080-103338080 on Build GRCh38
rs377171654 Chr.14: 103337857-103337858 on Build GRCh38
rs758637320 Chr.14: 103337878-103337881 on Build GRCh38
rs34047817 Chr.14: 103337858-103337859 on Build GRCh38
rs79484170 Chr.14: 103337804-103337804 on Build GRCh38
rs975041551 Chr.14: 103338153-103338153 on Build GRCh38
rs1010873279 Chr.14: 103338139-103338139 on Build GRCh38
rs1014142950 Chr.14: 103338138-103338138 on Build GRCh38
rs757491975 Chr.14: 103337936-103337936 on Build GRCh38
rs573534091 Chr.14: 103338125-103338125 on Build GRCh38
rs920893983 Chr.14: 103338162-103338162 on Build GRCh38
rs941064567 Chr.14: 103338032-103338033 on Build GRCh38
rs114716984 Chr.14: 103338151-103338151 on Build GRCh38
rs768230423 Chr.14: 103337868-103337868 on Build GRCh38
rs765305283 Chr.14: 103337802-103337802 on Build GRCh38
rs773047451 Chr.14: 103337951-103337951 on Build GRCh38
rs745803571 Chr.14: 103338186-103338186 on Build GRCh38
rs1033716096 Chr.14: 103338116-103338116 on Build GRCh38
rs780576519 Chr.14: 103337941-103337941 on Build GRCh38
rs898606008 Chr.14: 103338104-103338104 on Build GRCh38
rs756755204 Chr.14: 103337940-103337940 on Build GRCh38
rs184067682 Chr.14: 103338132-103338132 on Build GRCh38
rs1024658520 Chr.14: 103338140-103338140 on Build GRCh38
rs764242369 Chr.14: 103337958-103337958 on Build GRCh38
rs934278693 Chr.14: 103338062-103338062 on Build GRCh38
rs1041867324 Chr.14: 103337788-103337788 on Build GRCh38
rs749562172 Chr.14: 103337795-103337795 on Build GRCh38
rs767851034 Chr.14: 103337969-103337969 on Build GRCh38
rs113982504 Chr.14: 103338000-103338000 on Build GRCh38
rs529675265 Chr.14: 103337858-103337858 on Build GRCh38
rs536617634 Chr.14: 103338101-103338101 on Build GRCh38
rs1032741832 Chr.14: 103338150-103338150 on Build GRCh38
rs568999693 Chr.14: 103337982-103337982 on Build GRCh38
rs546345987 Chr.14: 103338106-103338106 on Build GRCh38
rs748823990 Chr.14: 103337864-103337864 on Build GRCh38
rs908817659 Chr.14: 103338178-103338178 on Build GRCh38
rs772924687 Chr.14: 103337890-103337890 on Build GRCh38
rs779223057 Chr.14: 103337829-103337831 on Build GRCh38
rs747027417 Chr.14: 103337970-103337971 on Build GRCh38
rs746395063 Chr.14: 103337924-103337924 on Build GRCh38
rs376331676 Chr.14: 103337761-103337761 on Build GRCh38
rs909539229 Chr.14: 103338015-103338015 on Build GRCh38
rs925705979 Chr.14: 103337998-103337998 on Build GRCh38
rs377042799 Chr.14: 103337928-103337928 on Build GRCh38
rs887242619 Chr.14: 103338090-103338090 on Build GRCh38
rs967207341 Chr.14: 103338142-103338142 on Build GRCh38
rs188829448 Chr.14: 103337927-103337927 on Build GRCh38
rs1039427092 Chr.14: 103338048-103338048 on Build GRCh38
rs987042725 Chr.14: 103338175-103338175 on Build GRCh38
rs1039458321 Chr.14: 103337877-103337877 on Build GRCh38
rs767330819 Chr.14: 103337889-103337889 on Build GRCh38
rs977685036 Chr.14: 103338144-103338144 on Build GRCh38
rs552309514 Chr.14: 103337959-103337959 on Build GRCh38
rs940197036 Chr.14: 103338097-103338097 on Build GRCh38
rs752761248 Chr.14: 103337926-103337926 on Build GRCh38
rs1031290010 Chr.14: 103338124-103338124 on Build GRCh38
rs534024507 Chr.14: 103338138-103338138 on Build GRCh38
rs146310156 Chr.14: 103338112-103338112 on Build GRCh38
rs975017285 Chr.14: 103338190-103338190 on Build GRCh38
rs565345079 Chr.14: 103337797-103337797 on Build GRCh38
rs780229484 Chr.14: 103337990-103337990 on Build GRCh38
rs751840283 Chr.14: 103337929-103337929 on Build GRCh38
rs757643898 Chr.14: 103337822-103337823 on Build GRCh38
rs781658399 Chr.14: 103337832-103337832 on Build GRCh38
rs190216704 Chr.14: 103337794-103337794 on Build GRCh38
rs746386837 Chr.14: 103337834-103337834 on Build GRCh38
rs969848556 Chr.14: 103338146-103338146 on Build GRCh38
rs775137611 Chr.14: 103338040-103338040 on Build GRCh38
rs748796284 Chr.14: 103337943-103337943 on Build GRCh38
rs1035249236 Chr.14: 103338159-103338159 on Build GRCh38

View Full Product Details