Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 94869147-94869654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC442028
Entrez Gene ID
442028
Gene Name
uncharacterized LOC442028
Gene Aliases
-
UniGene
Hs.611545
Gene Chromosome Location
Chr.2: 94868685-94947342 on Build GRCh38
Transcripts
NR_037597.1 XM_005263876.2 XM_011510670.1

Gene Symbol
TEKT4
Entrez Gene ID
150483
Gene Name
tektin 4
Gene Aliases
-
UniGene
Hs.631517
Gene Chromosome Location
Chr.2: 94867767-94876829 on Build GRCh38
Transcripts
NR_037597.1 XM_005263876.2 XM_011510670.1

SNP Detail

SNP ID Chromosome Location
rs143556866 Chr.2: 94869307-94869307 on Build GRCh38
rs763813316 Chr.2: 94869416-94869416 on Build GRCh38
rs79002427 Chr.2: 94869630-94869630 on Build GRCh38
rs140626490 Chr.2: 94869373-94869373 on Build GRCh38
rs112926053 Chr.2: 94869228-94869228 on Build GRCh38
rs5014839 Chr.2: 94869402-94869402 on Build GRCh38
rs111848380 Chr.2: 94869264-94869264 on Build GRCh38
rs115152992 Chr.2: 94869184-94869184 on Build GRCh38
rs111399154 Chr.2: 94869552-94869552 on Build GRCh38
rs77140257 Chr.2: 94869214-94869214 on Build GRCh38
rs5014842 Chr.2: 94869377-94869377 on Build GRCh38
rs373803595 Chr.2: 94869287-94869287 on Build GRCh38
rs75982729 Chr.2: 94869180-94869180 on Build GRCh38
rs782110779 Chr.2: 94869620-94869620 on Build GRCh38
rs111246321 Chr.2: 94869197-94869197 on Build GRCh38
rs774301035 Chr.2: 94869439-94869439 on Build GRCh38
rs535770604 Chr.2: 94869622-94869622 on Build GRCh38
rs781940032 Chr.2: 94869226-94869226 on Build GRCh38
rs5014843 Chr.2: 94869345-94869345 on Build GRCh38
rs551534504 Chr.2: 94869329-94869329 on Build GRCh38
rs113672190 Chr.2: 94869564-94869564 on Build GRCh38
rs113808654 Chr.2: 94869168-94869168 on Build GRCh38
rs113915994 Chr.2: 94869321-94869321 on Build GRCh38
rs111581694 Chr.2: 94869203-94869203 on Build GRCh38
rs5014841 Chr.2: 94869391-94869391 on Build GRCh38
rs5014840 Chr.2: 94869392-94869392 on Build GRCh38
rs181364812 Chr.2: 94869351-94869351 on Build GRCh38
rs5014838 Chr.2: 94869437-94869437 on Build GRCh38
rs144290615 Chr.2: 94869475-94869475 on Build GRCh38
rs568192953 Chr.2: 94869334-94869334 on Build GRCh38
rs113020535 Chr.2: 94869569-94869569 on Build GRCh38
rs112481853 Chr.2: 94869516-94869516 on Build GRCh38
rs548798401 Chr.2: 94869263-94869263 on Build GRCh38
rs113823321 Chr.2: 94869285-94869285 on Build GRCh38
rs139693059 Chr.2: 94869253-94869253 on Build GRCh38
rs78442760 Chr.2: 94869619-94869619 on Build GRCh38
rs762711610 Chr.2: 94869347-94869347 on Build GRCh38
rs547664861 Chr.2: 94869207-94869207 on Build GRCh38
rs190659347 Chr.2: 94869477-94869477 on Build GRCh38

View Full Product Details