Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 85676909-85677420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG5
Entrez Gene ID
387066
Gene Name
small nucleolar RNA host gene 5
Gene Aliases
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
UniGene
Hs.292457
Gene Chromosome Location
Chr.6: 85677007-85678733 on Build GRCh38
Transcripts
NR_003038.2 NR_002743.2

Gene Symbol
SNORD50A
Entrez Gene ID
26799
Gene Name
small nucleolar RNA, C/D box 50A
Gene Aliases
RNU50, U50
UniGene
Gene Chromosome Location
Chr.6: 85677294-85677368 on Build GRCh38
Transcripts
NR_003038.2 NR_002743.2

SNP Detail

SNP ID Chromosome Location
rs1023232430 Chr.6: 85677180-85677180 on Build GRCh38
rs903087010 Chr.6: 85677382-85677382 on Build GRCh38
rs779720993 Chr.6: 85677395-85677396 on Build GRCh38
rs770810849 Chr.6: 85677358-85677358 on Build GRCh38
rs777059782 Chr.6: 85677378-85677378 on Build GRCh38
rs923142948 Chr.6: 85677041-85677041 on Build GRCh38
rs11545947 Chr.6: 85677138-85677138 on Build GRCh38
rs1011419707 Chr.6: 85676999-85676999 on Build GRCh38
rs758640144 Chr.6: 85677348-85677348 on Build GRCh38
rs986089861 Chr.6: 85677063-85677064 on Build GRCh38
rs746019269 Chr.6: 85677377-85677377 on Build GRCh38
rs765587748 Chr.6: 85677334-85677334 on Build GRCh38
rs143503129 Chr.6: 85677314-85677315 on Build GRCh38
rs780370069 Chr.6: 85677350-85677350 on Build GRCh38
rs1013480160 Chr.6: 85677171-85677171 on Build GRCh38
rs908790484 Chr.6: 85677251-85677251 on Build GRCh38
rs757458579 Chr.6: 85677372-85677372 on Build GRCh38
rs758833702 Chr.6: 85677392-85677392 on Build GRCh38
rs555669127 Chr.6: 85677352-85677352 on Build GRCh38
rs112084658 Chr.6: 85676993-85676993 on Build GRCh38
rs967361032 Chr.6: 85677059-85677059 on Build GRCh38
rs1059374 Chr.6: 85677064-85677064 on Build GRCh38
rs149080830 Chr.6: 85677244-85677244 on Build GRCh38
rs79011497 Chr.6: 85677125-85677125 on Build GRCh38
rs755770974 Chr.6: 85677329-85677330 on Build GRCh38
rs774088392 Chr.6: 85677303-85677303 on Build GRCh38
rs919898790 Chr.6: 85677216-85677216 on Build GRCh38
rs971786192 Chr.6: 85677012-85677012 on Build GRCh38
rs772033283 Chr.6: 85677253-85677261 on Build GRCh38
rs188165652 Chr.6: 85677256-85677256 on Build GRCh38
rs933142508 Chr.6: 85677048-85677048 on Build GRCh38
rs771116678 Chr.6: 85677265-85677265 on Build GRCh38
rs763442375 Chr.6: 85677319-85677319 on Build GRCh38
rs964459501 Chr.6: 85677130-85677130 on Build GRCh38
rs771863432 Chr.6: 85677206-85677207 on Build GRCh38
rs757688274 Chr.6: 85677250-85677250 on Build GRCh38
rs527270213 Chr.6: 85677387-85677387 on Build GRCh38
rs1001319671 Chr.6: 85676987-85676987 on Build GRCh38
rs773239342 Chr.6: 85677273-85677273 on Build GRCh38
rs1020160769 Chr.6: 85677006-85677006 on Build GRCh38
rs1005889338 Chr.6: 85677239-85677239 on Build GRCh38
rs773127533 Chr.6: 85677256-85677256 on Build GRCh38
rs1059376 Chr.6: 85677060-85677060 on Build GRCh38
rs753283093 Chr.6: 85677318-85677319 on Build GRCh38
rs774842558 Chr.6: 85677312-85677312 on Build GRCh38
rs947422481 Chr.6: 85677079-85677079 on Build GRCh38
rs775282327 Chr.6: 85677375-85677375 on Build GRCh38
rs1057227914 Chr.6: 85677147-85677147 on Build GRCh38
rs760990566 Chr.6: 85677264-85677265 on Build GRCh38
rs745709329 Chr.6: 85677366-85677366 on Build GRCh38
rs866054139 Chr.6: 85677046-85677046 on Build GRCh38
rs969101605 Chr.6: 85676958-85676958 on Build GRCh38
rs143092599 Chr.6: 85677391-85677391 on Build GRCh38
rs754431576 Chr.6: 85677329-85677329 on Build GRCh38
rs1055481096 Chr.6: 85677284-85677284 on Build GRCh38
rs377740413 Chr.6: 85677332-85677332 on Build GRCh38
rs780942790 Chr.6: 85677331-85677331 on Build GRCh38
rs750600656 Chr.6: 85677336-85677336 on Build GRCh38
rs748067653 Chr.6: 85677270-85677270 on Build GRCh38
rs954563406 Chr.6: 85677308-85677308 on Build GRCh38
rs1059373 Chr.6: 85677067-85677067 on Build GRCh38
rs80236938 Chr.6: 85676970-85676970 on Build GRCh38
rs754123809 Chr.6: 85677248-85677248 on Build GRCh38
rs542657781 Chr.6: 85677072-85677072 on Build GRCh38
rs931313296 Chr.6: 85677241-85677241 on Build GRCh38
rs771594685 Chr.6: 85677370-85677370 on Build GRCh38
rs896012867 Chr.6: 85677152-85677152 on Build GRCh38
rs779098225 Chr.6: 85677254-85677254 on Build GRCh38
rs751491874 Chr.6: 85677197-85677197 on Build GRCh38
rs759608310 Chr.6: 85677275-85677276 on Build GRCh38
rs770907006 Chr.6: 85677285-85677285 on Build GRCh38
rs751868483 Chr.6: 85677319-85677320 on Build GRCh38
rs934769303 Chr.6: 85677134-85677134 on Build GRCh38
rs759294747 Chr.6: 85677304-85677304 on Build GRCh38
rs754803693 Chr.6: 85677289-85677289 on Build GRCh38
rs749884745 Chr.6: 85676936-85676936 on Build GRCh38
rs572387330 Chr.6: 85677376-85677376 on Build GRCh38
rs749251084 Chr.6: 85677355-85677355 on Build GRCh38
rs1059375 Chr.6: 85677061-85677061 on Build GRCh38
rs769593949 Chr.6: 85677272-85677272 on Build GRCh38
rs767004773 Chr.6: 85677307-85677307 on Build GRCh38
rs908971980 Chr.6: 85676980-85676980 on Build GRCh38
rs762834781 Chr.6: 85677276-85677276 on Build GRCh38
rs777558541 Chr.6: 85677354-85677354 on Build GRCh38
rs1033957668 Chr.6: 85676992-85676992 on Build GRCh38
rs766503797 Chr.6: 85677395-85677395 on Build GRCh38
rs533177250 Chr.6: 85677265-85677265 on Build GRCh38
rs776603952 Chr.6: 85677274-85677274 on Build GRCh38
rs765532685 Chr.6: 85677388-85677388 on Build GRCh38
rs562674040 Chr.6: 85677260-85677260 on Build GRCh38
rs192599183 Chr.6: 85677315-85677315 on Build GRCh38

View Full Product Details