Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111901297-111901846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM1A
Entrez Gene ID
8759
Gene Name
ADAM metallopeptidase domain 1A (pseudogene)
Gene Aliases
ADAM1, ADAM1P, FTNAP, Ftna, PH-30a
UniGene
Hs.661708
Gene Chromosome Location
Chr.12: 111899063-111901902 on Build GRCh38
Transcripts
NR_036636.1 XM_017020127.1 XM_017020128.1 XM_017020132.1 XM_017020133.1 XM_017020134.1 XM_017020135.1 XM_017020136.1 XM_017020138.1 XM_011538948.2 XM_017020187.1

Gene Symbol
MAPKAPK5
Entrez Gene ID
8550
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Gene Aliases
MAPKAP-K5, MK-5, MK5, PRAK
UniGene
Hs.333120
Gene Chromosome Location
Chr.12: 111842228-111902225 on Build GRCh38
Transcripts
NR_036636.1 XM_017020127.1 XM_017020128.1 XM_017020132.1 XM_017020133.1 XM_017020134.1 XM_017020135.1 XM_017020136.1 XM_017020138.1 XM_011538948.2 XM_017020187.1

Gene Symbol
TMEM116
Entrez Gene ID
89894
Gene Name
transmembrane protein 116
Gene Aliases
-
UniGene
Hs.506815
Gene Chromosome Location
Chr.12: 111894936-112013219 on Build GRCh38
Transcripts
NR_036636.1 XM_017020127.1 XM_017020128.1 XM_017020132.1 XM_017020133.1 XM_017020134.1 XM_017020135.1 XM_017020136.1 XM_017020138.1 XM_011538948.2 XM_017020187.1

SNP Detail

SNP ID Chromosome Location
rs184565202 Chr.12: 111901691-111901691 on Build GRCh38
rs902293716 Chr.12: 111901751-111901751 on Build GRCh38
rs1044250313 Chr.12: 111901585-111901585 on Build GRCh38
rs977576525 Chr.12: 111901497-111901498 on Build GRCh38
rs985647320 Chr.12: 111901445-111901445 on Build GRCh38
rs745534312 Chr.12: 111901742-111901742 on Build GRCh38
rs968827221 Chr.12: 111901494-111901496 on Build GRCh38
rs561559847 Chr.12: 111901558-111901558 on Build GRCh38
rs1038156756 Chr.12: 111901557-111901557 on Build GRCh38
rs900130498 Chr.12: 111901659-111901661 on Build GRCh38
rs3741999 Chr.12: 111901804-111901804 on Build GRCh38
rs770893337 Chr.12: 111901327-111901327 on Build GRCh38
rs1022006607 Chr.12: 111901470-111901470 on Build GRCh38
rs192124639 Chr.12: 111901469-111901469 on Build GRCh38
rs564398631 Chr.12: 111901711-111901712 on Build GRCh38
rs968941862 Chr.12: 111901328-111901328 on Build GRCh38
rs915936315 Chr.12: 111901535-111901535 on Build GRCh38
rs924846731 Chr.12: 111901498-111901500 on Build GRCh38
rs1021258789 Chr.12: 111901616-111901616 on Build GRCh38
rs748287091 Chr.12: 111901466-111901466 on Build GRCh38
rs968926974 Chr.12: 111901444-111901444 on Build GRCh38
rs1037538178 Chr.12: 111901377-111901377 on Build GRCh38
rs745708367 Chr.12: 111901384-111901385 on Build GRCh38
rs930028134 Chr.12: 111901719-111901719 on Build GRCh38
rs770492623 Chr.12: 111901636-111901637 on Build GRCh38
rs1006527806 Chr.12: 111901565-111901565 on Build GRCh38
rs968979217 Chr.12: 111901621-111901621 on Build GRCh38
rs776572351 Chr.12: 111901337-111901337 on Build GRCh38
rs955716205 Chr.12: 111901408-111901408 on Build GRCh38
rs938659908 Chr.12: 111901333-111901334 on Build GRCh38
rs1057447194 Chr.12: 111901333-111901336 on Build GRCh38
rs977607684 Chr.12: 111901332-111901332 on Build GRCh38
rs905721503 Chr.12: 111901587-111901587 on Build GRCh38
rs1002692723 Chr.12: 111901589-111901589 on Build GRCh38
rs865969593 Chr.12: 111901419-111901419 on Build GRCh38
rs948849854 Chr.12: 111901376-111901376 on Build GRCh38
rs924876415 Chr.12: 111901332-111901333 on Build GRCh38
rs547776959 Chr.12: 111901430-111901430 on Build GRCh38
rs890366721 Chr.12: 111901638-111901640 on Build GRCh38
rs12317129 Chr.12: 111901647-111901647 on Build GRCh38
rs1035480305 Chr.12: 111901590-111901590 on Build GRCh38
rs111353084 Chr.12: 111901656-111901658 on Build GRCh38
rs948751874 Chr.12: 111901550-111901550 on Build GRCh38
rs911353614 Chr.12: 111901448-111901448 on Build GRCh38
rs189399231 Chr.12: 111901738-111901738 on Build GRCh38
rs915983481 Chr.12: 111901333-111901333 on Build GRCh38
rs779850992 Chr.12: 111901422-111901422 on Build GRCh38
rs774894414 Chr.12: 111901784-111901784 on Build GRCh38
rs746788717 Chr.12: 111901438-111901438 on Build GRCh38
rs1017718078 Chr.12: 111901464-111901466 on Build GRCh38
rs778886503 Chr.12: 111901334-111901334 on Build GRCh38
rs909857969 Chr.12: 111901791-111901791 on Build GRCh38
rs1031833808 Chr.12: 111901775-111901775 on Build GRCh38
rs1048373511 Chr.12: 111901542-111901542 on Build GRCh38
rs1009916471 Chr.12: 111901609-111901609 on Build GRCh38
rs546090506 Chr.12: 111901664-111901666 on Build GRCh38
rs1047620442 Chr.12: 111901605-111901605 on Build GRCh38
rs898959260 Chr.12: 111901382-111901382 on Build GRCh38
rs965954393 Chr.12: 111901666-111901666 on Build GRCh38
rs545420990 Chr.12: 111901453-111901453 on Build GRCh38
rs918706689 Chr.12: 111901704-111901704 on Build GRCh38
rs116121301 Chr.12: 111901499-111901499 on Build GRCh38
rs1989866 Chr.12: 111901465-111901465 on Build GRCh38
rs116888411 Chr.12: 111901769-111901769 on Build GRCh38
rs955769100 Chr.12: 111901596-111901596 on Build GRCh38
rs558881427 Chr.12: 111901339-111901339 on Build GRCh38
rs1044729072 Chr.12: 111901809-111901809 on Build GRCh38
rs986088892 Chr.12: 111901641-111901641 on Build GRCh38
rs887703559 Chr.12: 111901560-111901560 on Build GRCh38
rs1035432806 Chr.12: 111901407-111901407 on Build GRCh38
rs767203103 Chr.12: 111901498-111901503 on Build GRCh38
rs976945551 Chr.12: 111901703-111901703 on Build GRCh38
rs76932235 Chr.12: 111901497-111901497 on Build GRCh38
rs187206454 Chr.12: 111901426-111901426 on Build GRCh38
rs558612614 Chr.12: 111901529-111901529 on Build GRCh38
rs754951941 Chr.12: 111901443-111901443 on Build GRCh38
rs758025063 Chr.12: 111901335-111901335 on Build GRCh38
rs768158686 Chr.12: 111901513-111901513 on Build GRCh38
rs1021974560 Chr.12: 111901670-111901681 on Build GRCh38

View Full Product Details