Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100193870-100194419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM30C
Entrez Gene ID
644444
Gene Name
transmembrane protein 30C
Gene Aliases
CDC50C
UniGene
Hs.652353
Gene Chromosome Location
Chr.3: 100185824-100194186 on Build GRCh38
Transcripts
NR_028357.1

SNP Detail

SNP ID Chromosome Location
rs906349901 Chr.3: 100194282-100194282 on Build GRCh38
rs192972193 Chr.3: 100194128-100194128 on Build GRCh38
rs1003352907 Chr.3: 100194294-100194294 on Build GRCh38
rs911359636 Chr.3: 100194108-100194108 on Build GRCh38
rs768893836 Chr.3: 100194166-100194166 on Build GRCh38
rs981460581 Chr.3: 100194028-100194028 on Build GRCh38
rs767724588 Chr.3: 100193950-100193950 on Build GRCh38
rs756864127 Chr.3: 100194107-100194107 on Build GRCh38
rs769967983 Chr.3: 100194254-100194254 on Build GRCh38
rs560251979 Chr.3: 100193990-100193990 on Build GRCh38
rs763572388 Chr.3: 100194038-100194038 on Build GRCh38
rs760206903 Chr.3: 100194005-100194005 on Build GRCh38
rs1053496044 Chr.3: 100194180-100194180 on Build GRCh38
rs962983069 Chr.3: 100194099-100194099 on Build GRCh38
rs1040824147 Chr.3: 100193915-100193915 on Build GRCh38
rs973144259 Chr.3: 100194337-100194337 on Build GRCh38
rs900351406 Chr.3: 100194311-100194311 on Build GRCh38
rs964073181 Chr.3: 100193967-100193967 on Build GRCh38
rs961656342 Chr.3: 100194051-100194051 on Build GRCh38
rs182573725 Chr.3: 100194220-100194220 on Build GRCh38
rs911495570 Chr.3: 100194363-100194363 on Build GRCh38
rs540550571 Chr.3: 100193931-100193931 on Build GRCh38
rs776786137 Chr.3: 100194200-100194200 on Build GRCh38
rs891227466 Chr.3: 100194355-100194359 on Build GRCh38
rs972790002 Chr.3: 100194066-100194066 on Build GRCh38
rs985681493 Chr.3: 100194358-100194358 on Build GRCh38
rs996944300 Chr.3: 100193975-100193975 on Build GRCh38
rs772356408 Chr.3: 100193897-100193897 on Build GRCh38
rs775110888 Chr.3: 100194231-100194231 on Build GRCh38
rs902284495 Chr.3: 100194149-100194149 on Build GRCh38
rs779972977 Chr.3: 100193979-100193979 on Build GRCh38
rs758323889 Chr.3: 100194022-100194022 on Build GRCh38
rs931594732 Chr.3: 100194095-100194095 on Build GRCh38
rs78455505 Chr.3: 100193955-100193955 on Build GRCh38
rs113536040 Chr.3: 100193896-100193896 on Build GRCh38
rs767154735 Chr.3: 100194292-100194292 on Build GRCh38
rs146242419 Chr.3: 100194164-100194164 on Build GRCh38
rs763302269 Chr.3: 100194288-100194288 on Build GRCh38
rs750475783 Chr.3: 100193952-100193952 on Build GRCh38
rs952701576 Chr.3: 100194355-100194355 on Build GRCh38
rs76181857 Chr.3: 100194326-100194326 on Build GRCh38
rs757756155 Chr.3: 100194349-100194363 on Build GRCh38
rs570604424 Chr.3: 100194381-100194381 on Build GRCh38
rs920165843 Chr.3: 100194072-100194072 on Build GRCh38
rs775562665 Chr.3: 100194279-100194279 on Build GRCh38
rs187113430 Chr.3: 100193964-100193964 on Build GRCh38
rs1030042123 Chr.3: 100193991-100193991 on Build GRCh38
rs955391438 Chr.3: 100194029-100194029 on Build GRCh38
rs1795130 Chr.3: 100194273-100194273 on Build GRCh38
rs1032659594 Chr.3: 100193962-100193962 on Build GRCh38
rs1042299762 Chr.3: 100193907-100193907 on Build GRCh38
rs527777405 Chr.3: 100194100-100194100 on Build GRCh38
rs1012031758 Chr.3: 100193977-100193977 on Build GRCh38
rs1026949733 Chr.3: 100194342-100194342 on Build GRCh38
rs752668514 Chr.3: 100194019-100194019 on Build GRCh38
rs868200674 Chr.3: 100194111-100194111 on Build GRCh38
rs1040793071 Chr.3: 100194129-100194129 on Build GRCh38
rs970540585 Chr.3: 100194021-100194021 on Build GRCh38
rs866499477 Chr.3: 100194297-100194297 on Build GRCh38
rs546526079 Chr.3: 100194103-100194103 on Build GRCh38
rs796708517 Chr.3: 100194310-100194310 on Build GRCh38
rs961519541 Chr.3: 100194318-100194318 on Build GRCh38
rs1015617477 Chr.3: 100194364-100194364 on Build GRCh38
rs762188086 Chr.3: 100193946-100193946 on Build GRCh38
rs188038486 Chr.3: 100194240-100194240 on Build GRCh38
rs1009865218 Chr.3: 100193906-100193906 on Build GRCh38
rs962933038 Chr.3: 100194375-100194375 on Build GRCh38
rs765248623 Chr.3: 100194250-100194250 on Build GRCh38
rs893465739 Chr.3: 100194218-100194218 on Build GRCh38
rs1036169693 Chr.3: 100194317-100194317 on Build GRCh38

View Full Product Details