Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 168812202-168812769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFEM1P
Entrez Gene ID
93556
Gene Name
EGF like and EMI domain containing 1, pseudogene
Gene Aliases
C3orf50, NCRNA00259
UniGene
Hs.478158
Gene Chromosome Location
Chr.3: 168249522-168830599 on Build GRCh38
Transcripts
NR_021485.2

SNP Detail

SNP ID Chromosome Location
rs867251702 Chr.3: 168812473-168812473 on Build GRCh38
rs1054783143 Chr.3: 168812593-168812593 on Build GRCh38
rs1054348981 Chr.3: 168812286-168812286 on Build GRCh38
rs778448320 Chr.3: 168812300-168812300 on Build GRCh38
rs563833320 Chr.3: 168812497-168812497 on Build GRCh38
rs1028836743 Chr.3: 168812455-168812455 on Build GRCh38
rs753850422 Chr.3: 168812278-168812278 on Build GRCh38
rs879071463 Chr.3: 168812259-168812259 on Build GRCh38
rs1044123404 Chr.3: 168812566-168812566 on Build GRCh38
rs57394291 Chr.3: 168812259-168812260 on Build GRCh38
rs927309363 Chr.3: 168812545-168812545 on Build GRCh38
rs866326283 Chr.3: 168812346-168812346 on Build GRCh38
rs201354995 Chr.3: 168812506-168812506 on Build GRCh38
rs892977229 Chr.3: 168812287-168812287 on Build GRCh38
rs917387264 Chr.3: 168812535-168812535 on Build GRCh38
rs1008773745 Chr.3: 168812657-168812657 on Build GRCh38
rs937245203 Chr.3: 168812271-168812271 on Build GRCh38
rs898747243 Chr.3: 168812686-168812686 on Build GRCh38
rs956028579 Chr.3: 168812719-168812719 on Build GRCh38
rs763570027 Chr.3: 168812433-168812433 on Build GRCh38
rs746596856 Chr.3: 168812415-168812415 on Build GRCh38
rs1044542509 Chr.3: 168812323-168812323 on Build GRCh38
rs1017641290 Chr.3: 168812679-168812679 on Build GRCh38
rs550552360 Chr.3: 168812393-168812393 on Build GRCh38
rs769116902 Chr.3: 168812426-168812426 on Build GRCh38
rs901585250 Chr.3: 168812368-168812368 on Build GRCh38
rs948908235 Chr.3: 168812551-168812551 on Build GRCh38
rs374007019 Chr.3: 168812258-168812258 on Build GRCh38
rs768776627 Chr.3: 168812256-168812256 on Build GRCh38
rs35629118 Chr.3: 168812519-168812520 on Build GRCh38
rs572738106 Chr.3: 168812516-168812516 on Build GRCh38
rs972920909 Chr.3: 168812484-168812487 on Build GRCh38
rs527943801 Chr.3: 168812691-168812691 on Build GRCh38
rs375768872 Chr.3: 168812543-168812543 on Build GRCh38
rs558442005 Chr.3: 168812526-168812526 on Build GRCh38
rs1024991886 Chr.3: 168812741-168812741 on Build GRCh38
rs796086729 Chr.3: 168812276-168812277 on Build GRCh38
rs192073171 Chr.3: 168812476-168812476 on Build GRCh38
rs1041866182 Chr.3: 168812335-168812335 on Build GRCh38
rs1047408598 Chr.3: 168812371-168812371 on Build GRCh38
rs1031675973 Chr.3: 168812716-168812716 on Build GRCh38
rs548064633 Chr.3: 168812260-168812261 on Build GRCh38
rs1000317787 Chr.3: 168812697-168812697 on Build GRCh38
rs749946541 Chr.3: 168812301-168812301 on Build GRCh38
rs764054475 Chr.3: 168812435-168812435 on Build GRCh38
rs370608849 Chr.3: 168812257-168812258 on Build GRCh38
rs886345405 Chr.3: 168812391-168812391 on Build GRCh38
rs918708745 Chr.3: 168812496-168812496 on Build GRCh38
rs534008954 Chr.3: 168812522-168812522 on Build GRCh38
rs1022154513 Chr.3: 168812685-168812685 on Build GRCh38
rs997209009 Chr.3: 168812375-168812375 on Build GRCh38
rs372165656 Chr.3: 168812236-168812236 on Build GRCh38
rs577272622 Chr.3: 168812625-168812625 on Build GRCh38
rs189536970 Chr.3: 168812667-168812667 on Build GRCh38
rs901622907 Chr.3: 168812651-168812651 on Build GRCh38
rs761315718 Chr.3: 168812258-168812259 on Build GRCh38
rs1008801936 Chr.3: 168812392-168812392 on Build GRCh38
rs781382203 Chr.3: 168812439-168812439 on Build GRCh38
rs994766031 Chr.3: 168812715-168812715 on Build GRCh38
rs993062567 Chr.3: 168812732-168812732 on Build GRCh38
rs375021725 Chr.3: 168812329-168812329 on Build GRCh38
rs924637185 Chr.3: 168812491-168812491 on Build GRCh38
rs595594 Chr.3: 168812671-168812671 on Build GRCh38
rs536179125 Chr.3: 168812463-168812463 on Build GRCh38
rs184340380 Chr.3: 168812648-168812648 on Build GRCh38
rs80213882 Chr.3: 168812464-168812464 on Build GRCh38
rs3732462 Chr.3: 168812652-168812652 on Build GRCh38
rs568471282 Chr.3: 168812429-168812429 on Build GRCh38
rs969056093 Chr.3: 168812255-168812255 on Build GRCh38
rs757841476 Chr.3: 168812312-168812312 on Build GRCh38
rs930423894 Chr.3: 168812627-168812627 on Build GRCh38
rs748758665 Chr.3: 168812459-168812459 on Build GRCh38
rs115336079 Chr.3: 168812440-168812440 on Build GRCh38
rs544341124 Chr.3: 168812629-168812629 on Build GRCh38
rs772994138 Chr.3: 168812277-168812278 on Build GRCh38
rs757135801 Chr.3: 168812295-168812295 on Build GRCh38
rs571334663 Chr.3: 168812277-168812277 on Build GRCh38
rs945904175 Chr.3: 168812634-168812634 on Build GRCh38
rs1040700214 Chr.3: 168812664-168812664 on Build GRCh38
rs748181472 Chr.3: 168812509-168812509 on Build GRCh38
rs538326965 Chr.3: 168812328-168812328 on Build GRCh38
rs920325488 Chr.3: 168812583-168812583 on Build GRCh38
rs866786963 Chr.3: 168812401-168812401 on Build GRCh38

View Full Product Details