Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127559656-127559887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO7B
Entrez Gene ID
4648
Gene Name
myosin VIIB
Gene Aliases
-
UniGene
Hs.154578
Gene Chromosome Location
Chr.2: 127535689-127637728 on Build GRCh38
Transcripts
NM_001080527.1 XM_006712539.3 XM_011511218.2 XM_017004169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
262269 c.5C>T p.S2L Chr.2: 127559727-127559727 on Build GRCh38
4449183 c.18+1G>T p.? Chr.2: 127559741-127559741 on Build GRCh38
4449182 c.18+1G>T p.? Chr.2: 127559741-127559741 on Build GRCh38
5502516 c.18+1G>A p.? Chr.2: 127559741-127559741 on Build GRCh38
3836757 c.12C>T p.F4F Chr.2: 127559734-127559734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377192509 Chr.2: 127559768-127559768 on Build GRCh38
rs779578963 Chr.2: 127559710-127559710 on Build GRCh38
rs780969803 Chr.2: 127559718-127559718 on Build GRCh38
rs1053942086 Chr.2: 127559760-127559760 on Build GRCh38
rs747796244 Chr.2: 127559726-127559726 on Build GRCh38
rs750810519 Chr.2: 127559712-127559712 on Build GRCh38
rs769236077 Chr.2: 127559678-127559678 on Build GRCh38
rs368709931 Chr.2: 127559777-127559777 on Build GRCh38
rs779261025 Chr.2: 127559769-127559769 on Build GRCh38
rs376887824 Chr.2: 127559804-127559804 on Build GRCh38
rs759316863 Chr.2: 127559751-127559751 on Build GRCh38
rs1056586989 Chr.2: 127559737-127559737 on Build GRCh38
rs774071718 Chr.2: 127559695-127559695 on Build GRCh38
rs1001310375 Chr.2: 127559843-127559843 on Build GRCh38
rs571733176 Chr.2: 127559812-127559812 on Build GRCh38
rs148264172 Chr.2: 127559841-127559841 on Build GRCh38
rs371672192 Chr.2: 127559803-127559803 on Build GRCh38
rs758889393 Chr.2: 127559714-127559714 on Build GRCh38
rs764120354 Chr.2: 127559704-127559704 on Build GRCh38
rs867610551 Chr.2: 127559692-127559692 on Build GRCh38
rs28618206 Chr.2: 127559853-127559853 on Build GRCh38
rs765842841 Chr.2: 127559675-127559675 on Build GRCh38
rs776160939 Chr.2: 127559727-127559727 on Build GRCh38
rs113869867 Chr.2: 127559817-127559817 on Build GRCh38
rs758950038 Chr.2: 127559810-127559810 on Build GRCh38
rs199510820 Chr.2: 127559700-127559700 on Build GRCh38
rs757658558 Chr.2: 127559708-127559708 on Build GRCh38
rs1017521358 Chr.2: 127559859-127559859 on Build GRCh38
rs189648068 Chr.2: 127559728-127559728 on Build GRCh38
rs773165611 Chr.2: 127559679-127559679 on Build GRCh38
rs965013403 Chr.2: 127559859-127559860 on Build GRCh38
rs753983663 Chr.2: 127559706-127559706 on Build GRCh38
rs943640752 Chr.2: 127559754-127559754 on Build GRCh38
rs759333392 Chr.2: 127559698-127559698 on Build GRCh38
rs771907122 Chr.2: 127559765-127559765 on Build GRCh38
rs748840608 Chr.2: 127559730-127559730 on Build GRCh38
rs535187718 Chr.2: 127559732-127559732 on Build GRCh38
rs369000497 Chr.2: 127559715-127559715 on Build GRCh38

View Full Product Details