Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180495382-180495639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD6
Entrez Gene ID
84320
Gene Name
acyl-CoA binding domain containing 6
Gene Aliases
-
UniGene
Hs.200051
Gene Chromosome Location
Chr.1: 180269663-180502967 on Build GRCh38
Transcripts
NM_032360.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668247 c.265G>T p.D89Y Chr.1: 180495483-180495483 on Build GRCh38
6591889 c.285delA p.K95fs*23 Chr.1: 180495463-180495463 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757400788 Chr.1: 180495430-180495430 on Build GRCh38
rs751301067 Chr.1: 180495527-180495527 on Build GRCh38
rs373691265 Chr.1: 180495417-180495417 on Build GRCh38
rs946728090 Chr.1: 180495535-180495535 on Build GRCh38
rs369393620 Chr.1: 180495546-180495546 on Build GRCh38
rs773950939 Chr.1: 180495559-180495559 on Build GRCh38
rs767304492 Chr.1: 180495532-180495532 on Build GRCh38
rs779123354 Chr.1: 180495467-180495467 on Build GRCh38
rs770484958 Chr.1: 180495479-180495479 on Build GRCh38
rs751414597 Chr.1: 180495421-180495421 on Build GRCh38
rs755424723 Chr.1: 180495573-180495573 on Build GRCh38
rs762765644 Chr.1: 180495521-180495521 on Build GRCh38
rs775168344 Chr.1: 180495512-180495512 on Build GRCh38
rs151129855 Chr.1: 180495531-180495531 on Build GRCh38
rs994940290 Chr.1: 180495597-180495597 on Build GRCh38
rs749748842 Chr.1: 180495447-180495447 on Build GRCh38
rs367797962 Chr.1: 180495445-180495445 on Build GRCh38
rs759272432 Chr.1: 180495493-180495493 on Build GRCh38
rs372225794 Chr.1: 180495501-180495501 on Build GRCh38
rs753966657 Chr.1: 180495568-180495568 on Build GRCh38
rs898807995 Chr.1: 180495589-180495589 on Build GRCh38
rs34463124 Chr.1: 180495519-180495520 on Build GRCh38
rs75982541 Chr.1: 180495609-180495609 on Build GRCh38
rs780122121 Chr.1: 180495446-180495446 on Build GRCh38
rs764144492 Chr.1: 180495525-180495525 on Build GRCh38
rs868267521 Chr.1: 180495556-180495556 on Build GRCh38
rs375543733 Chr.1: 180495506-180495506 on Build GRCh38
rs756352300 Chr.1: 180495548-180495548 on Build GRCh38
rs780069635 Chr.1: 180495558-180495558 on Build GRCh38
rs748601454 Chr.1: 180495478-180495478 on Build GRCh38
rs752653705 Chr.1: 180495540-180495541 on Build GRCh38
rs745947205 Chr.1: 180495432-180495432 on Build GRCh38
rs781538883 Chr.1: 180495431-180495431 on Build GRCh38
rs141033870 Chr.1: 180495602-180495602 on Build GRCh38
rs756121124 Chr.1: 180495442-180495442 on Build GRCh38
rs769237721 Chr.1: 180495448-180495448 on Build GRCh38
rs776355636 Chr.1: 180495487-180495487 on Build GRCh38
rs201717287 Chr.1: 180495413-180495413 on Build GRCh38

View Full Product Details