Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45038224-45038497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2
Entrez Gene ID
83605
Gene Name
CCM2 scaffolding protein
Gene Aliases
C7orf22, OSM, PP10187
UniGene
Hs.148272
Gene Chromosome Location
Chr.7: 44999746-45076470 on Build GRCh38
Transcripts
NM_001029835.2 NM_001167934.1 NM_001167935.1 NM_031443.3 NR_030770.1 XM_006715785.3 XM_006715786.3 XM_011515561.2 XM_011515562.1 XM_011515563.2 XM_011515564.2 XM_017012671.1 XM_017012672.1 XM_017012673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4970300 c.267+8G>A p.? Chr.7: 45038434-45038434 on Build GRCh38
7171561 c.126C>T p.F42F Chr.7: 45038285-45038285 on Build GRCh38
4996843 c.146G>C p.R49T Chr.7: 45038305-45038305 on Build GRCh38
3263224 c.136G>A p.E46K Chr.7: 45038295-45038295 on Build GRCh38
1090027 c.217C>T p.R73C Chr.7: 45038376-45038376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761427273 Chr.7: 45038456-45038456 on Build GRCh38
rs765327096 Chr.7: 45038438-45038438 on Build GRCh38
rs771577643 Chr.7: 45038385-45038385 on Build GRCh38
rs773845177 Chr.7: 45038381-45038381 on Build GRCh38
rs755800734 Chr.7: 45038277-45038277 on Build GRCh38
rs2107732 Chr.7: 45038379-45038379 on Build GRCh38
rs780210490 Chr.7: 45038454-45038454 on Build GRCh38
rs142320735 Chr.7: 45038270-45038270 on Build GRCh38
rs763778029 Chr.7: 45038402-45038402 on Build GRCh38
rs759155563 Chr.7: 45038254-45038254 on Build GRCh38
rs763981736 Chr.7: 45038279-45038279 on Build GRCh38
rs760398669 Chr.7: 45038401-45038401 on Build GRCh38
rs770271552 Chr.7: 45038474-45038474 on Build GRCh38
rs116596883 Chr.7: 45038473-45038473 on Build GRCh38
rs753670430 Chr.7: 45038317-45038317 on Build GRCh38
rs376855404 Chr.7: 45038434-45038434 on Build GRCh38
rs751679565 Chr.7: 45038457-45038457 on Build GRCh38
rs746014166 Chr.7: 45038352-45038352 on Build GRCh38
rs781380226 Chr.7: 45038472-45038472 on Build GRCh38
rs751071658 Chr.7: 45038436-45038437 on Build GRCh38
rs747256904 Chr.7: 45038360-45038360 on Build GRCh38
rs199865749 Chr.7: 45038450-45038450 on Build GRCh38
rs748579964 Chr.7: 45038378-45038378 on Build GRCh38
rs781634033 Chr.7: 45038376-45038376 on Build GRCh38
rs774965400 Chr.7: 45038400-45038400 on Build GRCh38
rs372598381 Chr.7: 45038433-45038433 on Build GRCh38
rs922721845 Chr.7: 45038437-45038437 on Build GRCh38
rs146579519 Chr.7: 45038348-45038348 on Build GRCh38
rs780362886 Chr.7: 45038354-45038354 on Build GRCh38
rs910903710 Chr.7: 45038396-45038396 on Build GRCh38
rs942924855 Chr.7: 45038403-45038403 on Build GRCh38
rs759104121 Chr.7: 45038382-45038382 on Build GRCh38
rs747384033 Chr.7: 45038266-45038266 on Build GRCh38
rs755950481 Chr.7: 45038340-45038340 on Build GRCh38
rs139113713 Chr.7: 45038361-45038361 on Build GRCh38
rs547769770 Chr.7: 45038455-45038455 on Build GRCh38
rs370580229 Chr.7: 45038451-45038451 on Build GRCh38
rs368976298 Chr.7: 45038463-45038463 on Build GRCh38

View Full Product Details