Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120548725-120548998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNTB1
Entrez Gene ID
6641
Gene Name
syntrophin beta 1
Gene Aliases
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43
UniGene
Hs.46701
Gene Chromosome Location
Chr.8: 120535745-120812069 on Build GRCh38
Transcripts
NM_021021.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1700591 c.1163G>A p.G388E Chr.8: 120548932-120548932 on Build GRCh38
6856628 c.1137-4_1137-3insTC p.? Chr.8: 120548961-120548962 on Build GRCh38
1724310 c.1137-5_1137-4delTC p.? Chr.8: 120548962-120548963 on Build GRCh38
5976612 c.1143C>T p.V381V Chr.8: 120548952-120548952 on Build GRCh38
4522313 c.1156G>A p.G386R Chr.8: 120548939-120548939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749027044 Chr.8: 120548961-120548962 on Build GRCh38
rs369675242 Chr.8: 120548814-120548814 on Build GRCh38
rs201759508 Chr.8: 120548913-120548913 on Build GRCh38
rs762326336 Chr.8: 120548846-120548846 on Build GRCh38
rs762447055 Chr.8: 120548891-120548891 on Build GRCh38
rs752407948 Chr.8: 120548887-120548887 on Build GRCh38
rs746999114 Chr.8: 120548797-120548797 on Build GRCh38
rs779936049 Chr.8: 120548905-120548905 on Build GRCh38
rs894013871 Chr.8: 120548971-120548971 on Build GRCh38
rs546161138 Chr.8: 120548859-120548859 on Build GRCh38
rs759040938 Chr.8: 120548872-120548872 on Build GRCh38
rs147938962 Chr.8: 120548922-120548922 on Build GRCh38
rs367672113 Chr.8: 120548968-120548968 on Build GRCh38
rs931729446 Chr.8: 120548786-120548786 on Build GRCh38
rs768714966 Chr.8: 120548918-120548918 on Build GRCh38
rs753293640 Chr.8: 120548757-120548757 on Build GRCh38
rs539908856 Chr.8: 120548966-120548966 on Build GRCh38
rs765901308 Chr.8: 120548754-120548754 on Build GRCh38
rs373006836 Chr.8: 120548900-120548900 on Build GRCh38
rs150921542 Chr.8: 120548761-120548761 on Build GRCh38
rs769073888 Chr.8: 120548841-120548841 on Build GRCh38
rs775770688 Chr.8: 120548838-120548838 on Build GRCh38
rs756279078 Chr.8: 120548898-120548898 on Build GRCh38
rs751464905 Chr.8: 120548952-120548952 on Build GRCh38
rs61762674 Chr.8: 120548788-120548788 on Build GRCh38
rs752499760 Chr.8: 120548760-120548760 on Build GRCh38
rs774814006 Chr.8: 120548949-120548949 on Build GRCh38
rs754671121 Chr.8: 120548758-120548758 on Build GRCh38
rs779386567 Chr.8: 120548921-120548921 on Build GRCh38
rs776156441 Chr.8: 120548863-120548863 on Build GRCh38
rs764361263 Chr.8: 120548857-120548857 on Build GRCh38
rs141740285 Chr.8: 120548938-120548938 on Build GRCh38
rs373208447 Chr.8: 120548833-120548833 on Build GRCh38
rs138424014 Chr.8: 120548823-120548823 on Build GRCh38
rs781472720 Chr.8: 120548810-120548810 on Build GRCh38
rs138861807 Chr.8: 120548962-120548963 on Build GRCh38
rs763971821 Chr.8: 120548893-120548893 on Build GRCh38
rs1054360843 Chr.8: 120548933-120548933 on Build GRCh38
rs773697841 Chr.8: 120548932-120548932 on Build GRCh38
rs982797889 Chr.8: 120548796-120548796 on Build GRCh38
rs911380205 Chr.8: 120548832-120548832 on Build GRCh38
rs772635723 Chr.8: 120548927-120548927 on Build GRCh38
rs199549031 Chr.8: 120548896-120548896 on Build GRCh38
rs377444423 Chr.8: 120548895-120548895 on Build GRCh38
rs370983314 Chr.8: 120548942-120548942 on Build GRCh38
rs768463931 Chr.8: 120548962-120548965 on Build GRCh38
rs774535850 Chr.8: 120548845-120548845 on Build GRCh38
rs758198750 Chr.8: 120548768-120548768 on Build GRCh38
rs764836354 Chr.8: 120548759-120548759 on Build GRCh38
rs61752911 Chr.8: 120548894-120548894 on Build GRCh38
rs920477778 Chr.8: 120548779-120548779 on Build GRCh38
rs374593019 Chr.8: 120548951-120548951 on Build GRCh38
rs764855445 Chr.8: 120548880-120548880 on Build GRCh38
rs757212699 Chr.8: 120548803-120548803 on Build GRCh38
rs376545139 Chr.8: 120548825-120548825 on Build GRCh38

View Full Product Details