Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44055038-44055311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF223
Entrez Gene ID
7766
Gene Name
zinc finger protein 223
Gene Aliases
-
UniGene
Hs.720813
Gene Chromosome Location
Chr.19: 44051372-44069902 on Build GRCh38
Transcripts
NM_013361.5 XM_017027258.1 XM_017027259.1

SNP Detail

SNP ID Chromosome Location
rs771861914 Chr.19: 44055106-44055106 on Build GRCh38
rs976143929 Chr.19: 44055247-44055248 on Build GRCh38
rs777997822 Chr.19: 44055095-44055095 on Build GRCh38
rs547664867 Chr.19: 44055057-44055057 on Build GRCh38
rs781514349 Chr.19: 44055115-44055115 on Build GRCh38
rs35667802 Chr.19: 44055116-44055117 on Build GRCh38
rs10164320 Chr.19: 44055069-44055069 on Build GRCh38
rs762665627 Chr.19: 44055239-44055239 on Build GRCh38
rs750105018 Chr.19: 44055124-44055124 on Build GRCh38
rs769663667 Chr.19: 44055073-44055074 on Build GRCh38
rs368242899 Chr.19: 44055114-44055114 on Build GRCh38
rs745624731 Chr.19: 44055162-44055162 on Build GRCh38
rs749368745 Chr.19: 44055238-44055238 on Build GRCh38
rs753098785 Chr.19: 44055194-44055194 on Build GRCh38
rs201089953 Chr.19: 44055143-44055143 on Build GRCh38
rs899531291 Chr.19: 44055076-44055076 on Build GRCh38
rs188597251 Chr.19: 44055098-44055098 on Build GRCh38
rs1002941923 Chr.19: 44055145-44055145 on Build GRCh38
rs761418672 Chr.19: 44055118-44055118 on Build GRCh38
rs371084484 Chr.19: 44055119-44055119 on Build GRCh38
rs775445514 Chr.19: 44055228-44055229 on Build GRCh38
rs756203071 Chr.19: 44055204-44055204 on Build GRCh38
rs766116068 Chr.19: 44055133-44055133 on Build GRCh38
rs755563617 Chr.19: 44055059-44055059 on Build GRCh38
rs376253926 Chr.19: 44055274-44055274 on Build GRCh38
rs962164796 Chr.19: 44055134-44055134 on Build GRCh38
rs567565506 Chr.19: 44055243-44055243 on Build GRCh38
rs755263076 Chr.19: 44055151-44055151 on Build GRCh38
rs991355274 Chr.19: 44055214-44055214 on Build GRCh38
rs923255257 Chr.19: 44055259-44055259 on Build GRCh38
rs781736121 Chr.19: 44055179-44055179 on Build GRCh38
rs781527289 Chr.19: 44055060-44055060 on Build GRCh38
rs191983492 Chr.19: 44055109-44055109 on Build GRCh38
rs777766432 Chr.19: 44055211-44055211 on Build GRCh38
rs776402173 Chr.19: 44055116-44055116 on Build GRCh38
rs762759818 Chr.19: 44055125-44055125 on Build GRCh38
rs769821972 Chr.19: 44055075-44055075 on Build GRCh38

View Full Product Details