Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52833898-52834161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT79
Entrez Gene ID
338785
Gene Name
keratin 79
Gene Aliases
K6L, KRT6L
UniGene
Hs.711471
Gene Chromosome Location
Chr.12: 52821447-52834293 on Build GRCh38
Transcripts
NM_175834.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311002 c.104C>A p.S35* Chr.12: 52834157-52834157 on Build GRCh38
1299559 c.196G>A p.G66R Chr.12: 52834065-52834065 on Build GRCh38
7204904 c.252T>A p.A84A Chr.12: 52834009-52834009 on Build GRCh38
261725 c.179G>A p.R60Q Chr.12: 52834082-52834082 on Build GRCh38
6254999 c.118C>A p.R40R Chr.12: 52834143-52834143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139252457 Chr.12: 52834004-52834004 on Build GRCh38
rs369790667 Chr.12: 52834118-52834119 on Build GRCh38
rs775985976 Chr.12: 52834135-52834135 on Build GRCh38
rs757618925 Chr.12: 52834088-52834088 on Build GRCh38
rs779227257 Chr.12: 52834116-52834116 on Build GRCh38
rs755319583 Chr.12: 52834060-52834061 on Build GRCh38
rs760206986 Chr.12: 52834079-52834079 on Build GRCh38
rs774268069 Chr.12: 52834002-52834002 on Build GRCh38
rs745966432 Chr.12: 52833965-52833965 on Build GRCh38
rs749904634 Chr.12: 52834008-52834008 on Build GRCh38
rs752930073 Chr.12: 52833955-52833955 on Build GRCh38
rs2638497 Chr.12: 52834019-52834019 on Build GRCh38
rs764726714 Chr.12: 52834139-52834139 on Build GRCh38
rs750599632 Chr.12: 52833934-52833935 on Build GRCh38
rs767131022 Chr.12: 52833919-52833919 on Build GRCh38
rs140098626 Chr.12: 52834105-52834105 on Build GRCh38
rs143471796 Chr.12: 52833950-52833950 on Build GRCh38
rs928955491 Chr.12: 52834055-52834055 on Build GRCh38
rs759113149 Chr.12: 52834136-52834136 on Build GRCh38
rs771177506 Chr.12: 52834031-52834031 on Build GRCh38
rs1000255684 Chr.12: 52833934-52833934 on Build GRCh38
rs767950892 Chr.12: 52834091-52834091 on Build GRCh38
rs754719555 Chr.12: 52834017-52834017 on Build GRCh38
rs762748757 Chr.12: 52833979-52833979 on Build GRCh38
rs932749377 Chr.12: 52834113-52834113 on Build GRCh38
rs779875886 Chr.12: 52833941-52833941 on Build GRCh38
rs1014832358 Chr.12: 52834125-52834125 on Build GRCh38
rs758807543 Chr.12: 52833987-52833988 on Build GRCh38
rs954629041 Chr.12: 52833921-52833922 on Build GRCh38
rs778287919 Chr.12: 52833951-52833951 on Build GRCh38
rs142983721 Chr.12: 52834033-52834033 on Build GRCh38
rs761692317 Chr.12: 52834063-52834063 on Build GRCh38
rs757708838 Chr.12: 52834021-52834021 on Build GRCh38
rs771539896 Chr.12: 52833959-52833959 on Build GRCh38
rs745879478 Chr.12: 52834032-52834032 on Build GRCh38
rs762650014 Chr.12: 52834143-52834143 on Build GRCh38
rs780164981 Chr.12: 52834035-52834035 on Build GRCh38
rs773993524 Chr.12: 52834060-52834060 on Build GRCh38
rs146288989 Chr.12: 52833928-52833928 on Build GRCh38
rs775692198 Chr.12: 52833974-52833974 on Build GRCh38
rs1014386231 Chr.12: 52833918-52833918 on Build GRCh38
rs749992742 Chr.12: 52833922-52833922 on Build GRCh38
rs780648343 Chr.12: 52834001-52834001 on Build GRCh38
rs141221786 Chr.12: 52833949-52833949 on Build GRCh38
rs373848280 Chr.12: 52834129-52834130 on Build GRCh38
rs144706200 Chr.12: 52834124-52834124 on Build GRCh38
rs749465670 Chr.12: 52834048-52834048 on Build GRCh38
rs773201448 Chr.12: 52834123-52834123 on Build GRCh38
rs376794232 Chr.12: 52834013-52834013 on Build GRCh38
rs191073791 Chr.12: 52834084-52834084 on Build GRCh38
rs112108733 Chr.12: 52834115-52834115 on Build GRCh38
rs747500796 Chr.12: 52834038-52834039 on Build GRCh38
rs147982249 Chr.12: 52833958-52833958 on Build GRCh38
rs759084678 Chr.12: 52834083-52834083 on Build GRCh38
rs573136966 Chr.12: 52834118-52834118 on Build GRCh38
rs768322135 Chr.12: 52833996-52833996 on Build GRCh38
rs146117735 Chr.12: 52834069-52834069 on Build GRCh38
rs980934138 Chr.12: 52834030-52834030 on Build GRCh38
rs756679057 Chr.12: 52834103-52834103 on Build GRCh38
rs750913221 Chr.12: 52834097-52834097 on Build GRCh38
rs765976515 Chr.12: 52834082-52834082 on Build GRCh38
rs769960334 Chr.12: 52833973-52833973 on Build GRCh38
rs767613356 Chr.12: 52834006-52834006 on Build GRCh38
rs150909244 Chr.12: 52834014-52834014 on Build GRCh38
rs373385213 Chr.12: 52834142-52834142 on Build GRCh38
rs747461471 Chr.12: 52833954-52833954 on Build GRCh38
rs201617351 Chr.12: 52834104-52834104 on Build GRCh38
rs768963365 Chr.12: 52834053-52834053 on Build GRCh38
rs772094718 Chr.12: 52834065-52834065 on Build GRCh38
rs781221492 Chr.12: 52833964-52833964 on Build GRCh38
rs371411949 Chr.12: 52834132-52834132 on Build GRCh38
rs772008601 Chr.12: 52834122-52834122 on Build GRCh38

View Full Product Details