Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58586473-58586732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSMAF
Entrez Gene ID
8439
Gene Name
neutral sphingomyelinase activation associated factor
Gene Aliases
FAN, GRAMD5
UniGene
Hs.372000
Gene Chromosome Location
Chr.8: 58583504-58659845 on Build GRCh38
Transcripts
NM_001144772.1 NM_003580.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3650024 c.2404C>T p.P802S Chr.8: 58586500-58586500 on Build GRCh38
5965374 c.2389-2A>G p.? Chr.8: 58586610-58586610 on Build GRCh38
1624104 c.2388C>G p.T796T Chr.8: 58586516-58586516 on Build GRCh38
7314330 c.2397C>A p.H799Q Chr.8: 58586507-58586507 on Build GRCh38
6193705 c.2430C>G p.D810E Chr.8: 58586474-58586474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760234860 Chr.8: 58586609-58586609 on Build GRCh38
rs745637347 Chr.8: 58586521-58586521 on Build GRCh38
rs375475310 Chr.8: 58586517-58586517 on Build GRCh38
rs990872384 Chr.8: 58586692-58586692 on Build GRCh38
rs753529419 Chr.8: 58586633-58586633 on Build GRCh38
rs756892524 Chr.8: 58586636-58586636 on Build GRCh38
rs776192883 Chr.8: 58586550-58586550 on Build GRCh38
rs775970470 Chr.8: 58586599-58586599 on Build GRCh38
rs368448102 Chr.8: 58586626-58586626 on Build GRCh38
rs144059643 Chr.8: 58586564-58586564 on Build GRCh38
rs533202658 Chr.8: 58586547-58586547 on Build GRCh38
rs371173698 Chr.8: 58586641-58586641 on Build GRCh38
rs547244877 Chr.8: 58586639-58586639 on Build GRCh38
rs771141347 Chr.8: 58586579-58586579 on Build GRCh38
rs758147904 Chr.8: 58586640-58586640 on Build GRCh38
rs780495086 Chr.8: 58586618-58586618 on Build GRCh38
rs138530641 Chr.8: 58586523-58586523 on Build GRCh38
rs769994066 Chr.8: 58586562-58586562 on Build GRCh38
rs767913242 Chr.8: 58586598-58586598 on Build GRCh38
rs773631020 Chr.8: 58586563-58586563 on Build GRCh38
rs984172283 Chr.8: 58586613-58586613 on Build GRCh38
rs367876230 Chr.8: 58586647-58586647 on Build GRCh38
rs976225018 Chr.8: 58586584-58586584 on Build GRCh38
rs3816846 Chr.8: 58586678-58586678 on Build GRCh38
rs542801540 Chr.8: 58586666-58586666 on Build GRCh38
rs949027566 Chr.8: 58586663-58586663 on Build GRCh38
rs759885433 Chr.8: 58586597-58586597 on Build GRCh38
rs753476022 Chr.8: 58586509-58586509 on Build GRCh38
rs147332065 Chr.8: 58586638-58586638 on Build GRCh38
rs965697159 Chr.8: 58586520-58586520 on Build GRCh38
rs917971772 Chr.8: 58586693-58586693 on Build GRCh38
rs756315849 Chr.8: 58586658-58586658 on Build GRCh38
rs748245286 Chr.8: 58586557-58586557 on Build GRCh38
rs774597540 Chr.8: 58586586-58586586 on Build GRCh38
rs757125055 Chr.8: 58586513-58586513 on Build GRCh38

View Full Product Details