Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120575023-120575280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNTB1
Entrez Gene ID
6641
Gene Name
syntrophin beta 1
Gene Aliases
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43
UniGene
Hs.46701
Gene Chromosome Location
Chr.8: 120535745-120812069 on Build GRCh38
Transcripts
NM_021021.3 XM_011517239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520921 c.1083G>A p.R361R Chr.8: 120575139-120575139 on Build GRCh38
3896483 c.1020G>A p.Q340Q Chr.8: 120575202-120575202 on Build GRCh38
5408752 c.1003G>A p.G335R Chr.8: 120575219-120575219 on Build GRCh38
1700592 c.1023G>A p.W341* Chr.8: 120575199-120575199 on Build GRCh38
1288117 c.1057C>G p.L353V Chr.8: 120575165-120575165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781535683 Chr.8: 120575123-120575123 on Build GRCh38
rs1003793300 Chr.8: 120575098-120575098 on Build GRCh38
rs372322110 Chr.8: 120575130-120575130 on Build GRCh38
rs996987796 Chr.8: 120575058-120575058 on Build GRCh38
rs764294820 Chr.8: 120575231-120575231 on Build GRCh38
rs370175109 Chr.8: 120575061-120575061 on Build GRCh38
rs760719104 Chr.8: 120575055-120575055 on Build GRCh38
rs755001518 Chr.8: 120575078-120575078 on Build GRCh38
rs746326215 Chr.8: 120575125-120575125 on Build GRCh38
rs561464293 Chr.8: 120575088-120575088 on Build GRCh38
rs752979349 Chr.8: 120575220-120575220 on Build GRCh38
rs1033914737 Chr.8: 120575203-120575203 on Build GRCh38
rs149155827 Chr.8: 120575141-120575141 on Build GRCh38
rs368125907 Chr.8: 120575129-120575129 on Build GRCh38
rs116157159 Chr.8: 120575109-120575109 on Build GRCh38
rs145253773 Chr.8: 120575119-120575119 on Build GRCh38
rs752725496 Chr.8: 120575204-120575204 on Build GRCh38
rs776449114 Chr.8: 120575155-120575155 on Build GRCh38
rs765333713 Chr.8: 120575198-120575198 on Build GRCh38
rs758659001 Chr.8: 120575207-120575207 on Build GRCh38
rs766198690 Chr.8: 120575056-120575056 on Build GRCh38
rs1007717190 Chr.8: 120575183-120575183 on Build GRCh38
rs776829908 Chr.8: 120575101-120575101 on Build GRCh38
rs200901214 Chr.8: 120575057-120575057 on Build GRCh38
rs747342268 Chr.8: 120575117-120575117 on Build GRCh38
rs377365462 Chr.8: 120575114-120575114 on Build GRCh38
rs373881265 Chr.8: 120575126-120575126 on Build GRCh38
rs765244166 Chr.8: 120575083-120575083 on Build GRCh38
rs888109063 Chr.8: 120575080-120575080 on Build GRCh38
rs759672190 Chr.8: 120575194-120575194 on Build GRCh38
rs868575731 Chr.8: 120575206-120575206 on Build GRCh38
rs771832700 Chr.8: 120575140-120575140 on Build GRCh38
rs760385622 Chr.8: 120575146-120575146 on Build GRCh38
rs765487117 Chr.8: 120575215-120575215 on Build GRCh38
rs143249644 Chr.8: 120575191-120575191 on Build GRCh38

View Full Product Details