Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151811266-151811495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RORC
Entrez Gene ID
6097
Gene Name
RAR related orphan receptor C
Gene Aliases
IMD42, NR1F3, RORG, RZR-GAMMA, RZRG, TOR
UniGene
Hs.256022
Gene Chromosome Location
Chr.1: 151806071-151832450 on Build GRCh38
Transcripts
NM_001001523.1 NM_005060.3 XM_006711484.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5377619 c.1339G>A p.E447K Chr.1: 151811381-151811381 on Build GRCh38
5992355 c.1289G>T p.R430L Chr.1: 151811431-151811431 on Build GRCh38
5901079 c.1513C>T p.H505Y Chr.1: 151811369-151811369 on Build GRCh38
5377620 c.1501G>A p.E501K Chr.1: 151811381-151811381 on Build GRCh38
4820993 c.1465G>C p.E489Q Chr.1: 151811417-151811417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763253904 Chr.1: 151811454-151811454 on Build GRCh38
rs376963484 Chr.1: 151811455-151811455 on Build GRCh38
rs569307472 Chr.1: 151811387-151811387 on Build GRCh38
rs761762343 Chr.1: 151811326-151811326 on Build GRCh38
rs750071909 Chr.1: 151811397-151811397 on Build GRCh38
rs199643455 Chr.1: 151811415-151811415 on Build GRCh38
rs911262602 Chr.1: 151811392-151811392 on Build GRCh38
rs200720891 Chr.1: 151811460-151811460 on Build GRCh38
rs201250362 Chr.1: 151811446-151811446 on Build GRCh38
rs760348811 Chr.1: 151811465-151811465 on Build GRCh38
rs769094214 Chr.1: 151811428-151811428 on Build GRCh38
rs200854121 Chr.1: 151811364-151811364 on Build GRCh38
rs199757303 Chr.1: 151811453-151811453 on Build GRCh38
rs148168558 Chr.1: 151811416-151811416 on Build GRCh38
rs763996284 Chr.1: 151811466-151811466 on Build GRCh38
rs987415040 Chr.1: 151811445-151811445 on Build GRCh38
rs199663055 Chr.1: 151811303-151811303 on Build GRCh38
rs756040895 Chr.1: 151811466-151811469 on Build GRCh38
rs975512932 Chr.1: 151811352-151811352 on Build GRCh38
rs867040756 Chr.1: 151811345-151811345 on Build GRCh38
rs777198149 Chr.1: 151811325-151811325 on Build GRCh38
rs202117520 Chr.1: 151811380-151811380 on Build GRCh38
rs772394939 Chr.1: 151811290-151811290 on Build GRCh38
rs200556721 Chr.1: 151811386-151811386 on Build GRCh38
rs879179527 Chr.1: 151811451-151811451 on Build GRCh38
rs201848485 Chr.1: 151811357-151811357 on Build GRCh38
rs863225092 Chr.1: 151811399-151811399 on Build GRCh38
rs113195254 Chr.1: 151811329-151811329 on Build GRCh38
rs781505074 Chr.1: 151811434-151811434 on Build GRCh38
rs752599135 Chr.1: 151811346-151811346 on Build GRCh38
rs201021982 Chr.1: 151811338-151811338 on Build GRCh38
rs747498049 Chr.1: 151811306-151811306 on Build GRCh38
rs200283675 Chr.1: 151811362-151811362 on Build GRCh38
rs144826157 Chr.1: 151811391-151811391 on Build GRCh38
rs769733906 Chr.1: 151811444-151811444 on Build GRCh38
rs557348384 Chr.1: 151811317-151811317 on Build GRCh38
rs572976073 Chr.1: 151811439-151811439 on Build GRCh38
rs767417110 Chr.1: 151811330-151811330 on Build GRCh38
rs200235475 Chr.1: 151811423-151811423 on Build GRCh38

View Full Product Details