Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48936925-48937194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNF8
Entrez Gene ID
11267
Gene Name
SNF8, ESCRT-II complex subunit
Gene Aliases
Dot3, EAP30, VPS22
UniGene
Hs.127249
Gene Chromosome Location
Chr.17: 48930096-48944842 on Build GRCh38
Transcripts
NM_001317192.1 NM_001317193.1 NM_001317194.1 NM_007241.3 NR_133679.1 XM_017024075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919119 c.268A>T p.M90L Chr.17: 48937101-48937101 on Build GRCh38
4841251 c.295G>A p.E99K Chr.17: 48937074-48937074 on Build GRCh38
6786111 c.294C>T p.Y98Y Chr.17: 48937075-48937075 on Build GRCh38
7152749 c.342G>A p.R114R Chr.17: 48937027-48937027 on Build GRCh38
4728683 c.283delG p.D95fs*6 Chr.17: 48937086-48937086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150340023 Chr.17: 48937070-48937070 on Build GRCh38
rs775693541 Chr.17: 48937002-48937002 on Build GRCh38
rs182202830 Chr.17: 48936970-48936970 on Build GRCh38
rs200399045 Chr.17: 48937065-48937065 on Build GRCh38
rs745975120 Chr.17: 48936994-48936994 on Build GRCh38
rs185256464 Chr.17: 48937045-48937045 on Build GRCh38
rs751738449 Chr.17: 48937028-48937028 on Build GRCh38
rs763236773 Chr.17: 48937092-48937092 on Build GRCh38
rs996182216 Chr.17: 48936947-48936947 on Build GRCh38
rs148219925 Chr.17: 48937044-48937044 on Build GRCh38
rs978772273 Chr.17: 48937157-48937157 on Build GRCh38
rs750722478 Chr.17: 48937084-48937084 on Build GRCh38
rs765521924 Chr.17: 48936980-48936980 on Build GRCh38
rs763111564 Chr.17: 48937158-48937159 on Build GRCh38
rs769008499 Chr.17: 48937071-48937071 on Build GRCh38
rs373056828 Chr.17: 48937053-48937053 on Build GRCh38
rs774459422 Chr.17: 48937075-48937075 on Build GRCh38
rs979549540 Chr.17: 48936948-48936948 on Build GRCh38
rs753569007 Chr.17: 48937048-48937048 on Build GRCh38
rs765038423 Chr.17: 48937149-48937149 on Build GRCh38
rs764220340 Chr.17: 48937007-48937007 on Build GRCh38
rs371124582 Chr.17: 48936980-48936980 on Build GRCh38
rs770183694 Chr.17: 48937001-48937001 on Build GRCh38
rs775991547 Chr.17: 48937068-48937068 on Build GRCh38
rs759995685 Chr.17: 48936991-48936991 on Build GRCh38
rs747613030 Chr.17: 48936977-48936977 on Build GRCh38
rs778440474 Chr.17: 48937050-48937050 on Build GRCh38
rs754748926 Chr.17: 48937140-48937140 on Build GRCh38
rs753748950 Chr.17: 48937131-48937131 on Build GRCh38
rs201483230 Chr.17: 48937039-48937039 on Build GRCh38
rs199555139 Chr.17: 48937120-48937120 on Build GRCh38
rs746422678 Chr.17: 48937057-48937057 on Build GRCh38
rs558743408 Chr.17: 48937049-48937049 on Build GRCh38
rs1005240136 Chr.17: 48937029-48937029 on Build GRCh38
rs773406269 Chr.17: 48937093-48937093 on Build GRCh38
rs763291180 Chr.17: 48937003-48937003 on Build GRCh38
rs767656400 Chr.17: 48937040-48937040 on Build GRCh38
rs534726666 Chr.17: 48937150-48937150 on Build GRCh38
rs777233502 Chr.17: 48937056-48937056 on Build GRCh38
rs749012298 Chr.17: 48936962-48936962 on Build GRCh38
rs144811357 Chr.17: 48936981-48936981 on Build GRCh38
rs750421797 Chr.17: 48937042-48937042 on Build GRCh38
rs916934460 Chr.17: 48937141-48937141 on Build GRCh38
rs760754929 Chr.17: 48937102-48937102 on Build GRCh38
rs766421717 Chr.17: 48937106-48937106 on Build GRCh38
rs954644741 Chr.17: 48937005-48937005 on Build GRCh38
rs140203050 Chr.17: 48937054-48937054 on Build GRCh38
rs755999959 Chr.17: 48936959-48936959 on Build GRCh38
rs778549461 Chr.17: 48936976-48936976 on Build GRCh38
rs762091985 Chr.17: 48937090-48937090 on Build GRCh38
rs568449612 Chr.17: 48936961-48936961 on Build GRCh38
rs958324223 Chr.17: 48937074-48937074 on Build GRCh38

View Full Product Details