Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64213460-64213733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF180
Entrez Gene ID
285671
Gene Name
ring finger protein 180
Gene Aliases
RINES
UniGene
Hs.657843
Gene Chromosome Location
Chr.5: 64165311-64372879 on Build GRCh38
Transcripts
NM_001113561.2 NM_001323291.1 NM_001323292.1 NM_178532.4 XM_017009383.1 XM_017009384.1 XM_017009385.1 XM_017009386.1 XM_017009387.1 XM_017009388.1 XM_017009389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6546284 c.337G>T p.V113L Chr.5: 64213663-64213663 on Build GRCh38
5935047 c.287G>A p.G96D Chr.5: 64213613-64213613 on Build GRCh38
5014491 c.361G>A p.D121N Chr.5: 64213687-64213687 on Build GRCh38
1438129 c.361G>T p.D121Y Chr.5: 64213687-64213687 on Build GRCh38
1438130 c.361G>T p.D121Y Chr.5: 64213687-64213687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764421850 Chr.5: 64213546-64213546 on Build GRCh38
rs750783386 Chr.5: 64213577-64213581 on Build GRCh38
rs142785620 Chr.5: 64213648-64213648 on Build GRCh38
rs148499067 Chr.5: 64213634-64213634 on Build GRCh38
rs909588639 Chr.5: 64213490-64213490 on Build GRCh38
rs375289875 Chr.5: 64213613-64213613 on Build GRCh38
rs763255059 Chr.5: 64213612-64213632 on Build GRCh38
rs760202147 Chr.5: 64213508-64213508 on Build GRCh38
rs906000604 Chr.5: 64213556-64213556 on Build GRCh38
rs770388333 Chr.5: 64213513-64213513 on Build GRCh38
rs762116789 Chr.5: 64213687-64213687 on Build GRCh38
rs745857742 Chr.5: 64213658-64213658 on Build GRCh38
rs767563672 Chr.5: 64213689-64213689 on Build GRCh38
rs375332267 Chr.5: 64213498-64213499 on Build GRCh38
rs567688078 Chr.5: 64213561-64213561 on Build GRCh38
rs367689363 Chr.5: 64213667-64213667 on Build GRCh38
rs749667360 Chr.5: 64213631-64213631 on Build GRCh38
rs1007209006 Chr.5: 64213701-64213701 on Build GRCh38
rs12519901 Chr.5: 64213522-64213522 on Build GRCh38
rs764999316 Chr.5: 64213527-64213527 on Build GRCh38
rs370850914 Chr.5: 64213637-64213637 on Build GRCh38
rs202006196 Chr.5: 64213604-64213604 on Build GRCh38
rs776191605 Chr.5: 64213653-64213653 on Build GRCh38
rs559542003 Chr.5: 64213499-64213499 on Build GRCh38
rs933234928 Chr.5: 64213520-64213520 on Build GRCh38
rs375073151 Chr.5: 64213644-64213644 on Build GRCh38
rs878882078 Chr.5: 64213603-64213603 on Build GRCh38
rs762860713 Chr.5: 64213676-64213676 on Build GRCh38
rs777434556 Chr.5: 64213638-64213638 on Build GRCh38
rs775230760 Chr.5: 64213669-64213669 on Build GRCh38
rs957793731 Chr.5: 64213483-64213483 on Build GRCh38
rs1057196899 Chr.5: 64213636-64213636 on Build GRCh38
rs868295762 Chr.5: 64213601-64213601 on Build GRCh38
rs752530810 Chr.5: 64213529-64213529 on Build GRCh38
rs750953555 Chr.5: 64213605-64213605 on Build GRCh38
rs776473882 Chr.5: 64213519-64213519 on Build GRCh38
rs140094514 Chr.5: 64213690-64213690 on Build GRCh38
rs552409529 Chr.5: 64213496-64213496 on Build GRCh38
rs1014263895 Chr.5: 64213668-64213668 on Build GRCh38
rs751780220 Chr.5: 64213587-64213587 on Build GRCh38
rs146084819 Chr.5: 64213682-64213682 on Build GRCh38
rs780361444 Chr.5: 64213627-64213627 on Build GRCh38
rs371172901 Chr.5: 64213681-64213681 on Build GRCh38
rs373845774 Chr.5: 64213663-64213663 on Build GRCh38
rs756092169 Chr.5: 64213695-64213695 on Build GRCh38
rs200955638 Chr.5: 64213528-64213528 on Build GRCh38
rs199920917 Chr.5: 64213537-64213537 on Build GRCh38
rs199996968 Chr.5: 64213673-64213673 on Build GRCh38
rs757440879 Chr.5: 64213602-64213602 on Build GRCh38
rs1048543210 Chr.5: 64213489-64213490 on Build GRCh38

View Full Product Details