Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130405520-130405789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS8
Entrez Gene ID
11095
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 8
Gene Aliases
ADAM-TS8, METH2
UniGene
Hs.271605
Gene Chromosome Location
Chr.11: 130404923-130428993 on Build GRCh38
Transcripts
NM_007037.5 XM_017017145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4019351 c.2653A>T p.T885S Chr.11: 130405662-130405662 on Build GRCh38
6131664 c.2558C>A p.T853N Chr.11: 130405757-130405757 on Build GRCh38
3445335 c.2568C>T p.I856I Chr.11: 130405747-130405747 on Build GRCh38
4659665 c.2613T>C p.S871S Chr.11: 130405702-130405702 on Build GRCh38
3445336 c.2481C>T p.I827I Chr.11: 130405747-130405747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750922134 Chr.11: 130405700-130405700 on Build GRCh38
rs373074592 Chr.11: 130405739-130405739 on Build GRCh38
rs199626173 Chr.11: 130405677-130405677 on Build GRCh38
rs747729320 Chr.11: 130405639-130405639 on Build GRCh38
rs565933413 Chr.11: 130405727-130405727 on Build GRCh38
rs768909867 Chr.11: 130405683-130405683 on Build GRCh38
rs763738205 Chr.11: 130405665-130405665 on Build GRCh38
rs766628377 Chr.11: 130405704-130405704 on Build GRCh38
rs778544983 Chr.11: 130405756-130405756 on Build GRCh38
rs1000565767 Chr.11: 130405687-130405687 on Build GRCh38
rs769891870 Chr.11: 130405563-130405563 on Build GRCh38
rs549603780 Chr.11: 130405599-130405599 on Build GRCh38
rs754672038 Chr.11: 130405728-130405728 on Build GRCh38
rs561556338 Chr.11: 130405561-130405561 on Build GRCh38
rs775580905 Chr.11: 130405581-130405581 on Build GRCh38
rs369686049 Chr.11: 130405729-130405729 on Build GRCh38
rs767722116 Chr.11: 130405569-130405569 on Build GRCh38
rs751192644 Chr.11: 130405624-130405624 on Build GRCh38
rs763124711 Chr.11: 130405548-130405548 on Build GRCh38
rs775007123 Chr.11: 130405566-130405566 on Build GRCh38
rs774647812 Chr.11: 130405585-130405585 on Build GRCh38
rs770666720 Chr.11: 130405664-130405664 on Build GRCh38
rs751994009 Chr.11: 130405725-130405725 on Build GRCh38
rs779715250 Chr.11: 130405610-130405610 on Build GRCh38
rs766302837 Chr.11: 130405562-130405563 on Build GRCh38
rs760788962 Chr.11: 130405678-130405678 on Build GRCh38
rs770953224 Chr.11: 130405745-130405745 on Build GRCh38
rs754724987 Chr.11: 130405636-130405636 on Build GRCh38
rs61753089 Chr.11: 130405638-130405638 on Build GRCh38
rs376504625 Chr.11: 130405765-130405765 on Build GRCh38
rs749046687 Chr.11: 130405659-130405659 on Build GRCh38
rs752196725 Chr.11: 130405731-130405731 on Build GRCh38
rs768081291 Chr.11: 130405553-130405553 on Build GRCh38
rs761892374 Chr.11: 130405695-130405695 on Build GRCh38
rs1021785890 Chr.11: 130405615-130405615 on Build GRCh38
rs758690876 Chr.11: 130405609-130405609 on Build GRCh38
rs748075301 Chr.11: 130405554-130405554 on Build GRCh38
rs762299796 Chr.11: 130405592-130405592 on Build GRCh38
rs1032314257 Chr.11: 130405647-130405647 on Build GRCh38
rs376567515 Chr.11: 130405597-130405597 on Build GRCh38
rs35281339 Chr.11: 130405582-130405582 on Build GRCh38
rs1041214738 Chr.11: 130405565-130405565 on Build GRCh38
rs776676907 Chr.11: 130405684-130405684 on Build GRCh38
rs769843029 Chr.11: 130405642-130405642 on Build GRCh38
rs199804043 Chr.11: 130405645-130405645 on Build GRCh38
rs750737688 Chr.11: 130405600-130405600 on Build GRCh38
rs749209098 Chr.11: 130405744-130405744 on Build GRCh38
rs765661608 Chr.11: 130405696-130405696 on Build GRCh38
rs999093985 Chr.11: 130405641-130405641 on Build GRCh38
rs373240029 Chr.11: 130405654-130405654 on Build GRCh38
rs762716573 Chr.11: 130405567-130405567 on Build GRCh38
rs753970758 Chr.11: 130405587-130405587 on Build GRCh38
rs746458796 Chr.11: 130405544-130405544 on Build GRCh38
rs755691092 Chr.11: 130405738-130405738 on Build GRCh38
rs187157580 Chr.11: 130405703-130405703 on Build GRCh38

View Full Product Details