Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56465999-56466272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
UniGene
Hs.604915
Gene Chromosome Location
Chr.6: 56457987-56954671 on Build GRCh38
Transcripts
NM_001144769.2 NM_001144770.1 NM_015548.4 NM_183380.3 XM_005249310.3 XM_005249315.3 XM_005249316.2 XM_005249318.2 XM_005249319.2 XM_005249320.4 XM_005249322.4 XM_005249323.3 XM_005249324.2 XM_011514824.2 XM_011514825.2 XM_011514826.2 XM_017011205.1 XM_017011206.1 XM_017011207.1 XM_017011208.1 XM_017011209.1 XM_017011210.1 XM_017011211.1 XM_017011212.1 XM_017011213.1 XM_017011214.1 XM_017011215.1 XM_017011216.1 XM_017011217.1 XM_017011218.1 XM_017011219.1 XM_017011220.1 XM_017011221.1 XM_017011222.1 XM_017011223.1 XM_017011224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3158505 c.21883C>T p.R7295* Chr.6: 56466174-56466174 on Build GRCh38
5414574 c.21970C>T p.P7324S Chr.6: 56466087-56466087 on Build GRCh38
6649817 c.21862-3C>G p.? Chr.6: 56466198-56466198 on Build GRCh38
5731728 c.21941C>T p.A7314V Chr.6: 56466116-56466116 on Build GRCh38
5731727 c.14780C>T p.A4927V Chr.6: 56466116-56466116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762215995 Chr.6: 56466124-56466124 on Build GRCh38
rs769154986 Chr.6: 56466070-56466070 on Build GRCh38
rs371363863 Chr.6: 56466215-56466215 on Build GRCh38
rs780204641 Chr.6: 56466057-56466057 on Build GRCh38
rs778796501 Chr.6: 56466188-56466188 on Build GRCh38
rs759254829 Chr.6: 56466165-56466165 on Build GRCh38
rs757537466 Chr.6: 56466035-56466035 on Build GRCh38
rs1035898014 Chr.6: 56466028-56466028 on Build GRCh38
rs1010931452 Chr.6: 56466050-56466050 on Build GRCh38
rs779049758 Chr.6: 56466046-56466046 on Build GRCh38
rs73747833 Chr.6: 56466049-56466049 on Build GRCh38
rs1003001591 Chr.6: 56466023-56466023 on Build GRCh38
rs1006050474 Chr.6: 56466053-56466053 on Build GRCh38
rs968091482 Chr.6: 56466097-56466097 on Build GRCh38
rs979075859 Chr.6: 56466152-56466152 on Build GRCh38
rs764187003 Chr.6: 56466180-56466180 on Build GRCh38
rs527354100 Chr.6: 56466138-56466138 on Build GRCh38
rs770587646 Chr.6: 56466130-56466130 on Build GRCh38
rs776974015 Chr.6: 56466113-56466113 on Build GRCh38
rs199834581 Chr.6: 56466199-56466199 on Build GRCh38
rs1014589062 Chr.6: 56466060-56466060 on Build GRCh38
rs996091356 Chr.6: 56466043-56466043 on Build GRCh38
rs769136844 Chr.6: 56466236-56466236 on Build GRCh38
rs779930356 Chr.6: 56466226-56466226 on Build GRCh38
rs1041067644 Chr.6: 56466020-56466020 on Build GRCh38
rs1050334346 Chr.6: 56466047-56466047 on Build GRCh38
rs139952901 Chr.6: 56466212-56466212 on Build GRCh38
rs747156298 Chr.6: 56466228-56466228 on Build GRCh38
rs748897258 Chr.6: 56466059-56466061 on Build GRCh38
rs766884642 Chr.6: 56466170-56466170 on Build GRCh38
rs147332345 Chr.6: 56466063-56466063 on Build GRCh38
rs753660669 Chr.6: 56466181-56466181 on Build GRCh38
rs757155481 Chr.6: 56466184-56466184 on Build GRCh38
rs752263952 Chr.6: 56466172-56466172 on Build GRCh38
rs920975458 Chr.6: 56466155-56466155 on Build GRCh38

View Full Product Details