Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12016305-12016556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928464
Entrez Gene ID
101928464
Gene Name
uncharacterized LOC101928464
Gene Aliases
-
UniGene
Hs.631643
Gene Chromosome Location
Chr.19: 11987617-12046275 on Build GRCh38
Transcripts
NR_134927.1 NR_134928.1 NR_134929.1 NM_001080411.2 NM_001308346.1 NM_001308348.1 NM_001308351.1 NM_001308355.1 NM_001308357.1 XM_006722661.3 XM_011527725.2 XM_011527728.2 XM_011527729.2 XM_017026413.1

Gene Symbol
ZNF433
Entrez Gene ID
163059
Gene Name
zinc finger protein 433
Gene Aliases
-
UniGene
Hs.667776
Gene Chromosome Location
Chr.19: 12014717-12035741 on Build GRCh38
Transcripts
NR_134927.1 NR_134928.1 NR_134929.1 NM_001080411.2 NM_001308346.1 NM_001308348.1 NM_001308351.1 NM_001308355.1 NM_001308357.1 XM_006722661.3 XM_011527725.2 XM_011527728.2 XM_011527729.2 XM_017026413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
991120 c.548C>T p.S183L Chr.19: 12016319-12016319 on Build GRCh38
6353984 c.554T>C p.L185P Chr.19: 12016313-12016313 on Build GRCh38
991121 c.526G>A p.G176R Chr.19: 12016341-12016341 on Build GRCh38
5767636 c.349T>G p.S117A Chr.19: 12016518-12016518 on Build GRCh38
4750057 c.455A>C p.N152T Chr.19: 12016412-12016412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747071444 Chr.19: 12016403-12016404 on Build GRCh38
rs202218817 Chr.19: 12016439-12016439 on Build GRCh38
rs898049979 Chr.19: 12016407-12016407 on Build GRCh38
rs547164452 Chr.19: 12016371-12016371 on Build GRCh38
rs778069284 Chr.19: 12016351-12016354 on Build GRCh38
rs988075227 Chr.19: 12016488-12016488 on Build GRCh38
rs771437839 Chr.19: 12016373-12016373 on Build GRCh38
rs758360967 Chr.19: 12016386-12016387 on Build GRCh38
rs778658700 Chr.19: 12016357-12016358 on Build GRCh38
rs968864049 Chr.19: 12016526-12016526 on Build GRCh38
rs374806336 Chr.19: 12016342-12016342 on Build GRCh38
rs201469236 Chr.19: 12016468-12016468 on Build GRCh38
rs780861439 Chr.19: 12016475-12016475 on Build GRCh38
rs771029161 Chr.19: 12016419-12016420 on Build GRCh38
rs367931065 Chr.19: 12016408-12016408 on Build GRCh38
rs767730957 Chr.19: 12016383-12016383 on Build GRCh38
rs528475989 Chr.19: 12016378-12016378 on Build GRCh38
rs758362373 Chr.19: 12016445-12016445 on Build GRCh38
rs149059930 Chr.19: 12016409-12016409 on Build GRCh38
rs377092636 Chr.19: 12016341-12016341 on Build GRCh38
rs745310674 Chr.19: 12016454-12016454 on Build GRCh38
rs776199468 Chr.19: 12016494-12016494 on Build GRCh38
rs1026726932 Chr.19: 12016397-12016397 on Build GRCh38
rs772696381 Chr.19: 12016521-12016521 on Build GRCh38
rs746292304 Chr.19: 12016507-12016507 on Build GRCh38
rs1002689972 Chr.19: 12016419-12016419 on Build GRCh38
rs772806888 Chr.19: 12016502-12016502 on Build GRCh38
rs760106630 Chr.19: 12016380-12016380 on Build GRCh38
rs779658291 Chr.19: 12016458-12016458 on Build GRCh38
rs993543228 Chr.19: 12016394-12016394 on Build GRCh38
rs746862390 Chr.19: 12016461-12016461 on Build GRCh38
rs772657302 Chr.19: 12016527-12016527 on Build GRCh38
rs781331414 Chr.19: 12016477-12016478 on Build GRCh38
rs775998676 Chr.19: 12016386-12016386 on Build GRCh38
rs757335027 Chr.19: 12016410-12016410 on Build GRCh38
rs769133572 Chr.19: 12016498-12016498 on Build GRCh38
rs752441635 Chr.19: 12016359-12016360 on Build GRCh38
rs777698060 Chr.19: 12016400-12016402 on Build GRCh38
rs765381751 Chr.19: 12016415-12016415 on Build GRCh38
rs776105145 Chr.19: 12016348-12016348 on Build GRCh38
rs747634888 Chr.19: 12016480-12016480 on Build GRCh38
rs770533887 Chr.19: 12016428-12016428 on Build GRCh38
rs1035535063 Chr.19: 12016436-12016436 on Build GRCh38

View Full Product Details