Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33280147-33280374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF362
Entrez Gene ID
149076
Gene Name
zinc finger protein 362
Gene Aliases
RN, lin-29
UniGene
Hs.743358
Gene Chromosome Location
Chr.1: 33256307-33300719 on Build GRCh38
Transcripts
NM_152493.2 XM_005270507.2 XM_005270508.4 XM_017000415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
385382 c.505A>G p.S169G Chr.1: 33280279-33280279 on Build GRCh38
6829653 c.582_583insA p.I197fs*34 Chr.1: 33280356-33280357 on Build GRCh38
1341899 c.507delC p.P171fs*39 Chr.1: 33280281-33280281 on Build GRCh38
1341900 c.536T>A p.I179N Chr.1: 33280310-33280310 on Build GRCh38
6829657 c.542G>A p.G181D Chr.1: 33280316-33280316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751574009 Chr.1: 33280287-33280287 on Build GRCh38
rs753772595 Chr.1: 33280282-33280282 on Build GRCh38
rs745425657 Chr.1: 33280335-33280335 on Build GRCh38
rs770585965 Chr.1: 33280240-33280240 on Build GRCh38
rs763991180 Chr.1: 33280182-33280182 on Build GRCh38
rs773956283 Chr.1: 33280245-33280245 on Build GRCh38
rs143005937 Chr.1: 33280180-33280180 on Build GRCh38
rs749779241 Chr.1: 33280226-33280226 on Build GRCh38
rs781327251 Chr.1: 33280302-33280302 on Build GRCh38
rs770180996 Chr.1: 33280220-33280220 on Build GRCh38
rs900235192 Chr.1: 33280331-33280331 on Build GRCh38
rs372351139 Chr.1: 33280275-33280275 on Build GRCh38
rs752960107 Chr.1: 33280320-33280320 on Build GRCh38
rs762490200 Chr.1: 33280181-33280186 on Build GRCh38
rs767449978 Chr.1: 33280174-33280174 on Build GRCh38
rs746737705 Chr.1: 33280281-33280281 on Build GRCh38
rs776450205 Chr.1: 33280351-33280351 on Build GRCh38
rs759348133 Chr.1: 33280228-33280233 on Build GRCh38
rs766172709 Chr.1: 33280192-33280197 on Build GRCh38
rs775049086 Chr.1: 33280337-33280337 on Build GRCh38
rs200063572 Chr.1: 33280286-33280286 on Build GRCh38
rs755154398 Chr.1: 33280190-33280190 on Build GRCh38
rs756484246 Chr.1: 33280321-33280321 on Build GRCh38
rs369076583 Chr.1: 33280333-33280333 on Build GRCh38
rs761245001 Chr.1: 33280172-33280177 on Build GRCh38
rs758516719 Chr.1: 33280187-33280187 on Build GRCh38
rs759264251 Chr.1: 33280248-33280248 on Build GRCh38
rs200855447 Chr.1: 33280251-33280251 on Build GRCh38
rs140453245 Chr.1: 33280323-33280323 on Build GRCh38
rs769178681 Chr.1: 33280171-33280172 on Build GRCh38
rs760670740 Chr.1: 33280179-33280179 on Build GRCh38
rs779836581 Chr.1: 33280181-33280181 on Build GRCh38
rs767427280 Chr.1: 33280331-33280332 on Build GRCh38
rs768361109 Chr.1: 33280349-33280349 on Build GRCh38
rs888619293 Chr.1: 33280338-33280338 on Build GRCh38
rs774176287 Chr.1: 33280191-33280192 on Build GRCh38
rs775799780 Chr.1: 33280169-33280174 on Build GRCh38
rs374737076 Chr.1: 33280285-33280285 on Build GRCh38
rs911845368 Chr.1: 33280253-33280253 on Build GRCh38
rs1053031763 Chr.1: 33280250-33280250 on Build GRCh38
rs780087741 Chr.1: 33280188-33280188 on Build GRCh38
rs775191592 Chr.1: 33280257-33280257 on Build GRCh38
rs760357804 Chr.1: 33280340-33280340 on Build GRCh38
rs138130115 Chr.1: 33280336-33280336 on Build GRCh38
rs778183086 Chr.1: 33280221-33280221 on Build GRCh38
rs201989290 Chr.1: 33280290-33280290 on Build GRCh38
rs748404073 Chr.1: 33280216-33280216 on Build GRCh38
rs772835675 Chr.1: 33280180-33280181 on Build GRCh38
rs781543299 Chr.1: 33280208-33280208 on Build GRCh38

View Full Product Details