Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179813562-179813819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM163A
Entrez Gene ID
148753
Gene Name
family with sequence similarity 163 member A
Gene Aliases
C1orf76, NDSP
UniGene
Hs.400696
Gene Chromosome Location
Chr.1: 179727763-179816198 on Build GRCh38
Transcripts
NM_173509.2 XM_005244913.2 XM_006711174.2 XM_006711175.2 XM_006711176.2 XM_006711177.1 XM_011509217.1 XM_011509221.1 XM_011509226.1 XM_011509228.1 XM_017000386.1 XM_017000387.1 XM_017000388.1 XM_017000389.1 XM_017000390.1 XM_017000391.1 XM_017000392.1 XM_017000393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844661 c.125T>C p.V42A Chr.1: 179813810-179813810 on Build GRCh38
677825 c.124G>T p.V42F Chr.1: 179813809-179813809 on Build GRCh38
6059884 c.120C>A p.T40T Chr.1: 179813805-179813805 on Build GRCh38
7164307 c.133G>A p.E45K Chr.1: 179813818-179813818 on Build GRCh38
7238379 c.120C>T p.T40T Chr.1: 179813805-179813805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754954531 Chr.1: 179813773-179813773 on Build GRCh38
rs533657501 Chr.1: 179813622-179813622 on Build GRCh38
rs750281157 Chr.1: 179813750-179813750 on Build GRCh38
rs771004626 Chr.1: 179813679-179813679 on Build GRCh38
rs988432492 Chr.1: 179813677-179813677 on Build GRCh38
rs909956293 Chr.1: 179813765-179813765 on Build GRCh38
rs1029813594 Chr.1: 179813599-179813599 on Build GRCh38
rs778086353 Chr.1: 179813799-179813799 on Build GRCh38
rs569383151 Chr.1: 179813753-179813753 on Build GRCh38
rs754094660 Chr.1: 179813759-179813759 on Build GRCh38
rs768223408 Chr.1: 179813778-179813778 on Build GRCh38
rs1006655719 Chr.1: 179813636-179813636 on Build GRCh38
rs996815853 Chr.1: 179813720-179813720 on Build GRCh38
rs867596478 Chr.1: 179813707-179813707 on Build GRCh38
rs372609404 Chr.1: 179813731-179813731 on Build GRCh38
rs751534575 Chr.1: 179813745-179813745 on Build GRCh38
rs747283916 Chr.1: 179813800-179813800 on Build GRCh38
rs772605030 Chr.1: 179813798-179813798 on Build GRCh38
rs767396668 Chr.1: 179813748-179813748 on Build GRCh38
rs746355433 Chr.1: 179813701-179813701 on Build GRCh38
rs375661436 Chr.1: 179813774-179813774 on Build GRCh38
rs1032293031 Chr.1: 179813732-179813732 on Build GRCh38
rs958753470 Chr.1: 179813739-179813739 on Build GRCh38
rs77831891 Chr.1: 179813650-179813650 on Build GRCh38
rs899873960 Chr.1: 179813700-179813700 on Build GRCh38
rs762324889 Chr.1: 179813733-179813733 on Build GRCh38
rs116039137 Chr.1: 179813725-179813725 on Build GRCh38
rs113586611 Chr.1: 179813583-179813583 on Build GRCh38
rs564766396 Chr.1: 179813597-179813597 on Build GRCh38
rs748694211 Chr.1: 179813746-179813746 on Build GRCh38
rs777637338 Chr.1: 179813748-179813749 on Build GRCh38
rs761897764 Chr.1: 179813747-179813747 on Build GRCh38
rs941475425 Chr.1: 179813662-179813662 on Build GRCh38
rs763660117 Chr.1: 179813734-179813734 on Build GRCh38
rs781598381 Chr.1: 179813590-179813590 on Build GRCh38
rs555224813 Chr.1: 179813705-179813705 on Build GRCh38
rs567036463 Chr.1: 179813683-179813683 on Build GRCh38
rs147916028 Chr.1: 179813726-179813726 on Build GRCh38
rs780090453 Chr.1: 179813758-179813758 on Build GRCh38
rs1033141978 Chr.1: 179813644-179813644 on Build GRCh38
rs146731765 Chr.1: 179813792-179813792 on Build GRCh38

View Full Product Details