Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120446308-120446581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATC
Entrez Gene ID
283459
Gene Name
glutamyl-tRNA amidotransferase subunit C
Gene Aliases
15E1.2
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120446438-120463753 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_016399.2

Gene Symbol
TRIAP1
Entrez Gene ID
51499
Gene Name
TP53 regulated inhibitor of apoptosis 1
Gene Aliases
HSPC132, MDM35, P53CSV, WF-1
UniGene
Hs.69499
Gene Chromosome Location
Chr.12: 120443961-120446412 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_016399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6807757 c.20C>T p.A7V Chr.12: 120446353-120446353 on Build GRCh38
6135589 c.53G>C p.C18S Chr.12: 120446320-120446320 on Build GRCh38
6000493 c.17A>G p.E6G Chr.12: 120446356-120446356 on Build GRCh38
6571306 c.31C>T p.R11W Chr.12: 120446511-120446511 on Build GRCh38
5517690 c.59A>G p.N20S Chr.12: 120446314-120446314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752262697 Chr.12: 120446373-120446373 on Build GRCh38
rs761316481 Chr.12: 120446512-120446512 on Build GRCh38
rs2235217 Chr.12: 120446525-120446525 on Build GRCh38
rs769918086 Chr.12: 120446418-120446418 on Build GRCh38
rs377312589 Chr.12: 120446495-120446495 on Build GRCh38
rs201599988 Chr.12: 120446388-120446388 on Build GRCh38
rs779378921 Chr.12: 120446445-120446445 on Build GRCh38
rs780456003 Chr.12: 120446342-120446342 on Build GRCh38
rs142254855 Chr.12: 120446480-120446480 on Build GRCh38
rs61734361 Chr.12: 120446502-120446502 on Build GRCh38
rs945448635 Chr.12: 120446490-120446490 on Build GRCh38
rs1000838329 Chr.12: 120446384-120446384 on Build GRCh38
rs142599020 Chr.12: 120446344-120446344 on Build GRCh38
rs766847472 Chr.12: 120446463-120446463 on Build GRCh38
rs756353947 Chr.12: 120446541-120446541 on Build GRCh38
rs759317993 Chr.12: 120446440-120446440 on Build GRCh38
rs781523005 Chr.12: 120446470-120446470 on Build GRCh38
rs768372222 Chr.12: 120446507-120446507 on Build GRCh38
rs753346194 Chr.12: 120446456-120446456 on Build GRCh38
rs371546739 Chr.12: 120446481-120446481 on Build GRCh38
rs755797453 Chr.12: 120446385-120446387 on Build GRCh38
rs757694666 Chr.12: 120446420-120446423 on Build GRCh38
rs117692929 Chr.12: 120446450-120446450 on Build GRCh38
rs773200634 Chr.12: 120446358-120446358 on Build GRCh38
rs773252043 Chr.12: 120446422-120446422 on Build GRCh38
rs774037459 Chr.12: 120446439-120446439 on Build GRCh38
rs752272387 Chr.12: 120446373-120446373 on Build GRCh38
rs17431446 Chr.12: 120446488-120446488 on Build GRCh38
rs750986851 Chr.12: 120446483-120446485 on Build GRCh38
rs764394826 Chr.12: 120446384-120446385 on Build GRCh38
rs758903722 Chr.12: 120446398-120446398 on Build GRCh38
rs905195344 Chr.12: 120446383-120446383 on Build GRCh38
rs749225531 Chr.12: 120446430-120446430 on Build GRCh38
rs200895047 Chr.12: 120446397-120446397 on Build GRCh38
rs776709841 Chr.12: 120446353-120446353 on Build GRCh38
rs564753747 Chr.12: 120446412-120446412 on Build GRCh38
rs770787459 Chr.12: 120446483-120446483 on Build GRCh38
rs747344933 Chr.12: 120446408-120446408 on Build GRCh38
rs756298995 Chr.12: 120446476-120446476 on Build GRCh38
rs1045565977 Chr.12: 120446503-120446503 on Build GRCh38
rs200932941 Chr.12: 120446395-120446395 on Build GRCh38
rs1026923599 Chr.12: 120446370-120446370 on Build GRCh38
rs1055404770 Chr.12: 120446355-120446355 on Build GRCh38
rs755623349 Chr.12: 120446381-120446381 on Build GRCh38
rs780312146 Chr.12: 120446400-120446400 on Build GRCh38
rs755165176 Chr.12: 120446411-120446411 on Build GRCh38
rs532063291 Chr.12: 120446562-120446562 on Build GRCh38
rs763748905 Chr.12: 120446453-120446453 on Build GRCh38
rs376552989 Chr.12: 120446552-120446552 on Build GRCh38
rs540772404 Chr.12: 120446465-120446465 on Build GRCh38
rs377336416 Chr.12: 120446445-120446445 on Build GRCh38
rs759142396 Chr.12: 120446367-120446367 on Build GRCh38
rs752964473 Chr.12: 120446474-120446474 on Build GRCh38
rs368556877 Chr.12: 120446522-120446522 on Build GRCh38
rs147621480 Chr.12: 120446361-120446361 on Build GRCh38
rs1003941678 Chr.12: 120446538-120446538 on Build GRCh38
rs145225296 Chr.12: 120446544-120446544 on Build GRCh38
rs147874381 Chr.12: 120446494-120446494 on Build GRCh38
rs201519820 Chr.12: 120446491-120446491 on Build GRCh38
rs368215298 Chr.12: 120446333-120446333 on Build GRCh38
rs767286445 Chr.12: 120446443-120446443 on Build GRCh38
rs557491914 Chr.12: 120446497-120446497 on Build GRCh38
rs745792428 Chr.12: 120446492-120446492 on Build GRCh38
rs754216471 Chr.12: 120446396-120446396 on Build GRCh38
rs767231229 Chr.12: 120446368-120446368 on Build GRCh38
rs549680688 Chr.12: 120446508-120446508 on Build GRCh38
rs752165199 Chr.12: 120446523-120446523 on Build GRCh38
rs748164331 Chr.12: 120446356-120446356 on Build GRCh38
rs1002419164 Chr.12: 120446347-120446347 on Build GRCh38
rs769726145 Chr.12: 120446357-120446357 on Build GRCh38
rs949491199 Chr.12: 120446337-120446337 on Build GRCh38
rs1010153028 Chr.12: 120446528-120446528 on Build GRCh38
rs1041593573 Chr.12: 120446533-120446533 on Build GRCh38
rs199833922 Chr.12: 120446524-120446524 on Build GRCh38
rs753021870 Chr.12: 120446535-120446535 on Build GRCh38
rs781047730 Chr.12: 120446431-120446431 on Build GRCh38

View Full Product Details