Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119172285-119172510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRX1
Entrez Gene ID
79671
Gene Name
NLR family member X1
Gene Aliases
CLR11.3, DLNB26, NOD26, NOD5, NOD9
UniGene
Hs.524082
Gene Chromosome Location
Chr.11: 119168334-119184016 on Build GRCh38
Transcripts
NM_001282143.1 NM_001282144.1 NM_001282358.1 NM_024618.3 XM_005271669.1 XM_005271670.1 XM_005271671.1 XM_005271672.1 XM_006718904.1 XM_011542980.1 XM_017018294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425405 c.114A>T p.Q38H Chr.11: 119172399-119172399 on Build GRCh38
3444124 c.83C>T p.P28L Chr.11: 119172368-119172368 on Build GRCh38
110483 c.124C>T p.P42S Chr.11: 119172409-119172409 on Build GRCh38
7425406 c.114A>T p.Q38H Chr.11: 119172399-119172399 on Build GRCh38
4462759 c.125C>T p.P42L Chr.11: 119172410-119172410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775689434 Chr.11: 119172361-119172361 on Build GRCh38
rs778429968 Chr.11: 119172472-119172472 on Build GRCh38
rs537131759 Chr.11: 119172317-119172317 on Build GRCh38
rs759515461 Chr.11: 119172406-119172406 on Build GRCh38
rs777542512 Chr.11: 119172331-119172331 on Build GRCh38
rs373080354 Chr.11: 119172436-119172436 on Build GRCh38
rs772389031 Chr.11: 119172358-119172358 on Build GRCh38
rs776914089 Chr.11: 119172366-119172366 on Build GRCh38
rs760586955 Chr.11: 119172424-119172424 on Build GRCh38
rs770012581 Chr.11: 119172401-119172401 on Build GRCh38
rs552852958 Chr.11: 119172439-119172439 on Build GRCh38
rs60612369 Chr.11: 119172362-119172362 on Build GRCh38
rs953195851 Chr.11: 119172367-119172367 on Build GRCh38
rs768753625 Chr.11: 119172364-119172364 on Build GRCh38
rs188616690 Chr.11: 119172410-119172410 on Build GRCh38
rs577488289 Chr.11: 119172445-119172445 on Build GRCh38
rs774391248 Chr.11: 119172405-119172405 on Build GRCh38
rs771369503 Chr.11: 119172315-119172315 on Build GRCh38
rs761922653 Chr.11: 119172383-119172383 on Build GRCh38
rs755698473 Chr.11: 119172319-119172319 on Build GRCh38
rs985939928 Chr.11: 119172389-119172389 on Build GRCh38
rs748745248 Chr.11: 119172337-119172337 on Build GRCh38
rs201371895 Chr.11: 119172346-119172346 on Build GRCh38
rs752575821 Chr.11: 119172318-119172318 on Build GRCh38
rs752419860 Chr.11: 119172416-119172416 on Build GRCh38
rs924311356 Chr.11: 119172476-119172476 on Build GRCh38
rs754770235 Chr.11: 119172312-119172312 on Build GRCh38
rs142957206 Chr.11: 119172409-119172409 on Build GRCh38
rs555322970 Chr.11: 119172348-119172348 on Build GRCh38
rs769088403 Chr.11: 119172437-119172438 on Build GRCh38
rs375464688 Chr.11: 119172311-119172311 on Build GRCh38

View Full Product Details