Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 56801775-56802048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTX2
Entrez Gene ID
5015
Gene Name
orthodenticle homeobox 2
Gene Aliases
CPHD6, MCOPS5
UniGene
Hs.288655
Gene Chromosome Location
Chr.14: 56800707-56810476 on Build GRCh38
Transcripts
NM_001270523.1 NM_001270524.1 NM_001270525.1 NM_021728.3 NM_172337.2 NR_073034.1 NR_073036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6140845 c.745C>A p.Q249K Chr.14: 56801884-56801884 on Build GRCh38
4773992 c.614G>A p.G205E Chr.14: 56802015-56802015 on Build GRCh38
2245566 c.684T>C p.S228S Chr.14: 56801945-56801945 on Build GRCh38
2245564 c.706A>C p.T236P Chr.14: 56801923-56801923 on Build GRCh38
956425 c.644C>A p.P215H Chr.14: 56801985-56801985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750626422 Chr.14: 56801956-56801956 on Build GRCh38
rs867979918 Chr.14: 56801842-56801842 on Build GRCh38
rs758228717 Chr.14: 56801871-56801871 on Build GRCh38
rs529946021 Chr.14: 56801947-56801947 on Build GRCh38
rs769304167 Chr.14: 56801905-56801905 on Build GRCh38
rs1006268708 Chr.14: 56801901-56801901 on Build GRCh38
rs781096873 Chr.14: 56801865-56801865 on Build GRCh38
rs139800030 Chr.14: 56801872-56801872 on Build GRCh38
rs977939396 Chr.14: 56801864-56801864 on Build GRCh38
rs780929725 Chr.14: 56801986-56801986 on Build GRCh38
rs144449264 Chr.14: 56801916-56801916 on Build GRCh38
rs772782540 Chr.14: 56801914-56801914 on Build GRCh38
rs746941491 Chr.14: 56801880-56801880 on Build GRCh38
rs548455339 Chr.14: 56801968-56801968 on Build GRCh38
rs1026307269 Chr.14: 56801885-56801885 on Build GRCh38
rs928898444 Chr.14: 56801955-56801955 on Build GRCh38
rs764571884 Chr.14: 56801936-56801936 on Build GRCh38
rs907201088 Chr.14: 56802013-56802013 on Build GRCh38
rs754929157 Chr.14: 56801974-56801974 on Build GRCh38
rs745480462 Chr.14: 56801936-56801944 on Build GRCh38
rs142743327 Chr.14: 56801966-56801966 on Build GRCh38
rs150982073 Chr.14: 56801988-56801988 on Build GRCh38
rs993488539 Chr.14: 56801860-56801860 on Build GRCh38
rs779220317 Chr.14: 56801832-56801832 on Build GRCh38
rs751376563 Chr.14: 56801973-56801973 on Build GRCh38
rs916022550 Chr.14: 56801961-56801961 on Build GRCh38
rs768514532 Chr.14: 56801881-56801881 on Build GRCh38
rs868121860 Chr.14: 56801950-56801950 on Build GRCh38
rs747665958 Chr.14: 56801898-56801898 on Build GRCh38
rs370761964 Chr.14: 56801931-56801931 on Build GRCh38
rs199650198 Chr.14: 56801886-56801886 on Build GRCh38
rs761733794 Chr.14: 56801949-56801949 on Build GRCh38
rs868295626 Chr.14: 56801895-56801895 on Build GRCh38
rs143131956 Chr.14: 56801890-56801890 on Build GRCh38
rs765441049 Chr.14: 56801952-56801952 on Build GRCh38
rs775876318 Chr.14: 56801924-56801924 on Build GRCh38
rs374968145 Chr.14: 56801965-56801965 on Build GRCh38
rs750679604 Chr.14: 56801847-56801847 on Build GRCh38

View Full Product Details