Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121781732-121781969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCB1
Entrez Gene ID
9657
Gene Name
IQ motif containing B1
Gene Aliases
NPHP5, PIQ, SLSN5
UniGene
Hs.604110
Gene Chromosome Location
Chr.3: 121769761-121835079 on Build GRCh38
Transcripts
NM_001023570.3 NM_001023571.3 NM_001319107.1 NR_134968.1 XM_005247911.3 XM_005247912.2 XM_011513335.2 XM_017007537.1 XM_017007538.1 XM_017007539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3120999 c.1400G>C p.R467T Chr.3: 121781753-121781753 on Build GRCh38
5397904 c.1399A>G p.R467G Chr.3: 121781754-121781754 on Build GRCh38
3121002 c.1290C>T p.F430F Chr.3: 121781863-121781863 on Build GRCh38
1579592 c.1303C>T p.R435C Chr.3: 121781850-121781850 on Build GRCh38
4583836 c.1368T>G p.V456V Chr.3: 121781785-121781785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760905369 Chr.3: 121781787-121781787 on Build GRCh38
rs926668992 Chr.3: 121781844-121781844 on Build GRCh38
rs866982675 Chr.3: 121781790-121781790 on Build GRCh38
rs146832128 Chr.3: 121781849-121781849 on Build GRCh38
rs752682393 Chr.3: 121781808-121781808 on Build GRCh38
rs779234709 Chr.3: 121781910-121781910 on Build GRCh38
rs1049368885 Chr.3: 121781881-121781882 on Build GRCh38
rs748257203 Chr.3: 121781914-121781914 on Build GRCh38
rs142536967 Chr.3: 121781842-121781842 on Build GRCh38
rs915510870 Chr.3: 121781826-121781826 on Build GRCh38
rs138646990 Chr.3: 121781873-121781873 on Build GRCh38
rs753426941 Chr.3: 121781792-121781792 on Build GRCh38
rs767201620 Chr.3: 121781889-121781889 on Build GRCh38
rs780735130 Chr.3: 121781903-121781904 on Build GRCh38
rs764717043 Chr.3: 121781797-121781797 on Build GRCh38
rs147708058 Chr.3: 121781760-121781760 on Build GRCh38
rs886057824 Chr.3: 121781809-121781809 on Build GRCh38
rs772003773 Chr.3: 121781818-121781818 on Build GRCh38
rs766072332 Chr.3: 121781789-121781789 on Build GRCh38
rs757143428 Chr.3: 121781880-121781880 on Build GRCh38
rs768788882 Chr.3: 121781758-121781758 on Build GRCh38
rs201319850 Chr.3: 121781872-121781872 on Build GRCh38
rs754160972 Chr.3: 121781890-121781890 on Build GRCh38
rs758347399 Chr.3: 121781922-121781922 on Build GRCh38
rs11920543 Chr.3: 121781850-121781850 on Build GRCh38
rs776326876 Chr.3: 121781866-121781866 on Build GRCh38
rs377629008 Chr.3: 121781833-121781833 on Build GRCh38
rs768913508 Chr.3: 121781839-121781839 on Build GRCh38
rs17849995 Chr.3: 121781852-121781852 on Build GRCh38
rs768911174 Chr.3: 121781925-121781928 on Build GRCh38
rs761109901 Chr.3: 121781857-121781857 on Build GRCh38
rs772369157 Chr.3: 121781784-121781784 on Build GRCh38
rs374241434 Chr.3: 121781793-121781793 on Build GRCh38
rs758519096 Chr.3: 121781882-121781882 on Build GRCh38
rs747497450 Chr.3: 121781914-121781917 on Build GRCh38
rs150129899 Chr.3: 121781858-121781858 on Build GRCh38
rs867772426 Chr.3: 121781820-121781820 on Build GRCh38
rs749515337 Chr.3: 121781834-121781834 on Build GRCh38
rs535158012 Chr.3: 121781918-121781918 on Build GRCh38
rs377265463 Chr.3: 121781819-121781819 on Build GRCh38
rs780327292 Chr.3: 121781829-121781829 on Build GRCh38
rs751386502 Chr.3: 121781876-121781876 on Build GRCh38
rs773593065 Chr.3: 121781785-121781785 on Build GRCh38
rs121918244 Chr.3: 121781772-121781772 on Build GRCh38
rs758303358 Chr.3: 121781813-121781813 on Build GRCh38
rs756943739 Chr.3: 121781821-121781821 on Build GRCh38
rs755216787 Chr.3: 121781908-121781908 on Build GRCh38
rs991890780 Chr.3: 121781770-121781770 on Build GRCh38
rs776989189 Chr.3: 121781874-121781874 on Build GRCh38
rs201474218 Chr.3: 121781807-121781807 on Build GRCh38
rs780089248 Chr.3: 121781882-121781884 on Build GRCh38

View Full Product Details