Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44104559-44104800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEBP1
Entrez Gene ID
165
Gene Name
AE binding protein 1
Gene Aliases
ACLP
UniGene
Hs.439463
Gene Chromosome Location
Chr.7: 44104320-44114565 on Build GRCh38
Transcripts
NM_001129.4 XM_011515162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354655 c.74C>T p.P25L Chr.7: 44104739-44104739 on Build GRCh38
6321303 c.126G>T p.L42L Chr.7: 44104791-44104791 on Build GRCh38
4886086 c.71G>T p.R24L Chr.7: 44104736-44104736 on Build GRCh38
6408983 c.60C>A p.C20* Chr.7: 44104725-44104725 on Build GRCh38
4956217 c.93C>T p.D31D Chr.7: 44104758-44104758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746317181 Chr.7: 44104714-44104714 on Build GRCh38
rs772653486 Chr.7: 44104724-44104724 on Build GRCh38
rs762832519 Chr.7: 44104662-44104662 on Build GRCh38
rs759336775 Chr.7: 44104779-44104779 on Build GRCh38
rs757535956 Chr.7: 44104705-44104705 on Build GRCh38
rs28362520 Chr.7: 44104602-44104602 on Build GRCh38
rs28362521 Chr.7: 44104685-44104685 on Build GRCh38
rs761505637 Chr.7: 44104632-44104632 on Build GRCh38
rs773068687 Chr.7: 44104655-44104655 on Build GRCh38
rs777965585 Chr.7: 44104695-44104695 on Build GRCh38
rs572403133 Chr.7: 44104686-44104686 on Build GRCh38
rs564360927 Chr.7: 44104744-44104744 on Build GRCh38
rs774219756 Chr.7: 44104767-44104767 on Build GRCh38
rs752310632 Chr.7: 44104729-44104729 on Build GRCh38
rs13241299 Chr.7: 44104727-44104727 on Build GRCh38
rs747660366 Chr.7: 44104746-44104746 on Build GRCh38
rs902326529 Chr.7: 44104669-44104669 on Build GRCh38
rs943255542 Chr.7: 44104625-44104625 on Build GRCh38
rs937655398 Chr.7: 44104578-44104578 on Build GRCh38
rs779371963 Chr.7: 44104708-44104708 on Build GRCh38
rs766049309 Chr.7: 44104764-44104764 on Build GRCh38
rs764989792 Chr.7: 44104638-44104638 on Build GRCh38
rs375195748 Chr.7: 44104747-44104747 on Build GRCh38
rs901700807 Chr.7: 44104641-44104641 on Build GRCh38
rs772980753 Chr.7: 44104761-44104761 on Build GRCh38
rs754975816 Chr.7: 44104684-44104684 on Build GRCh38
rs1031972849 Chr.7: 44104745-44104745 on Build GRCh38
rs766250601 Chr.7: 44104667-44104667 on Build GRCh38
rs575362804 Chr.7: 44104692-44104692 on Build GRCh38
rs917657328 Chr.7: 44104605-44104605 on Build GRCh38
rs933150154 Chr.7: 44104649-44104649 on Build GRCh38
rs751421512 Chr.7: 44104681-44104681 on Build GRCh38
rs767491556 Chr.7: 44104780-44104780 on Build GRCh38
rs1038877120 Chr.7: 44104637-44104637 on Build GRCh38
rs545702407 Chr.7: 44104702-44104702 on Build GRCh38
rs574534169 Chr.7: 44104613-44104613 on Build GRCh38
rs1042211294 Chr.7: 44104653-44104653 on Build GRCh38

View Full Product Details