Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149198360-149198617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF282
Entrez Gene ID
8427
Gene Name
zinc finger protein 282
Gene Aliases
HUB1
UniGene
Hs.729056
Gene Chromosome Location
Chr.7: 149195462-149226248 on Build GRCh38
Transcripts
NM_001303481.1 NM_003575.3 XM_006716151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1087124 c.401C>A p.A134D Chr.7: 149198568-149198568 on Build GRCh38
7114622 c.322G>A p.V108M Chr.7: 149198489-149198489 on Build GRCh38
3879167 c.273G>A p.L91L Chr.7: 149198440-149198440 on Build GRCh38
1449073 c.287T>C p.I96T Chr.7: 149198454-149198454 on Build GRCh38
5872192 c.388G>A p.E130K Chr.7: 149198555-149198555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752657322 Chr.7: 149198512-149198512 on Build GRCh38
rs760179364 Chr.7: 149198427-149198427 on Build GRCh38
rs771595296 Chr.7: 149198420-149198420 on Build GRCh38
rs142017242 Chr.7: 149198386-149198386 on Build GRCh38
rs150660033 Chr.7: 149198473-149198473 on Build GRCh38
rs764978893 Chr.7: 149198509-149198509 on Build GRCh38
rs780379063 Chr.7: 149198554-149198554 on Build GRCh38
rs922814828 Chr.7: 149198574-149198574 on Build GRCh38
rs778860364 Chr.7: 149198400-149198400 on Build GRCh38
rs781419599 Chr.7: 149198545-149198545 on Build GRCh38
rs780428606 Chr.7: 149198412-149198412 on Build GRCh38
rs536766991 Chr.7: 149198597-149198597 on Build GRCh38
rs768927441 Chr.7: 149198417-149198417 on Build GRCh38
rs199638052 Chr.7: 149198418-149198418 on Build GRCh38
rs746077313 Chr.7: 149198405-149198405 on Build GRCh38
rs891614728 Chr.7: 149198394-149198394 on Build GRCh38
rs759336110 Chr.7: 149198500-149198500 on Build GRCh38
rs190208939 Chr.7: 149198413-149198413 on Build GRCh38
rs758327150 Chr.7: 149198539-149198539 on Build GRCh38
rs149374266 Chr.7: 149198584-149198584 on Build GRCh38
rs752780037 Chr.7: 149198423-149198423 on Build GRCh38
rs758564810 Chr.7: 149198548-149198548 on Build GRCh38
rs752607755 Chr.7: 149198401-149198401 on Build GRCh38
rs139886507 Chr.7: 149198583-149198583 on Build GRCh38
rs571953952 Chr.7: 149198480-149198480 on Build GRCh38
rs1025936565 Chr.7: 149198558-149198558 on Build GRCh38
rs377732853 Chr.7: 149198479-149198479 on Build GRCh38
rs537694957 Chr.7: 149198551-149198551 on Build GRCh38
rs202090175 Chr.7: 149198404-149198404 on Build GRCh38
rs749508582 Chr.7: 149198563-149198563 on Build GRCh38
rs750103234 Chr.7: 149198577-149198578 on Build GRCh38
rs964169105 Chr.7: 149198424-149198424 on Build GRCh38
rs770209921 Chr.7: 149198410-149198410 on Build GRCh38
rs146346133 Chr.7: 149198596-149198596 on Build GRCh38
rs753560268 Chr.7: 149198382-149198382 on Build GRCh38
rs765954214 Chr.7: 149198458-149198458 on Build GRCh38
rs370207856 Chr.7: 149198575-149198575 on Build GRCh38
rs376294814 Chr.7: 149198402-149198402 on Build GRCh38
rs755341684 Chr.7: 149198569-149198569 on Build GRCh38

View Full Product Details