Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3018327-3018586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
UniGene
Hs.274873
Gene Chromosome Location
Chr.11: 3000922-3057451 on Build GRCh38
Transcripts
NM_001014437.2 NM_001194997.1 NM_001751.5 NM_139273.3 NR_036542.1 XM_006718341.3 XM_017018389.1 XM_017018390.1 XM_017018391.1 XM_017018392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620252 c.1318C>T p.R440W Chr.11: 3018509-3018509 on Build GRCh38
4875157 c.1362C>A p.T454T Chr.11: 3018465-3018465 on Build GRCh38
6620250 c.1528C>T p.R510W Chr.11: 3018509-3018509 on Build GRCh38
926241 c.1572C>A p.T524T Chr.11: 3018465-3018465 on Build GRCh38
3666261 c.1558T>A p.S520T Chr.11: 3018479-3018479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772663320 Chr.11: 3018538-3018538 on Build GRCh38
rs937727677 Chr.11: 3018348-3018349 on Build GRCh38
rs150252960 Chr.11: 3018417-3018417 on Build GRCh38
rs763061632 Chr.11: 3018561-3018561 on Build GRCh38
rs761110823 Chr.11: 3018541-3018541 on Build GRCh38
rs755064829 Chr.11: 3018395-3018395 on Build GRCh38
rs752839619 Chr.11: 3018508-3018508 on Build GRCh38
rs929256507 Chr.11: 3018399-3018399 on Build GRCh38
rs780883071 Chr.11: 3018516-3018516 on Build GRCh38
rs771491699 Chr.11: 3018544-3018544 on Build GRCh38
rs768147476 Chr.11: 3018447-3018447 on Build GRCh38
rs943649307 Chr.11: 3018368-3018368 on Build GRCh38
rs373346810 Chr.11: 3018433-3018433 on Build GRCh38
rs377202281 Chr.11: 3018380-3018380 on Build GRCh38
rs746776555 Chr.11: 3018510-3018510 on Build GRCh38
rs766676110 Chr.11: 3018481-3018481 on Build GRCh38
rs756342519 Chr.11: 3018374-3018374 on Build GRCh38
rs368361569 Chr.11: 3018432-3018432 on Build GRCh38
rs754068451 Chr.11: 3018486-3018486 on Build GRCh38
rs755084329 Chr.11: 3018489-3018489 on Build GRCh38
rs547458078 Chr.11: 3018359-3018359 on Build GRCh38
rs903414295 Chr.11: 3018405-3018405 on Build GRCh38
rs544038154 Chr.11: 3018498-3018498 on Build GRCh38
rs190331761 Chr.11: 3018440-3018440 on Build GRCh38
rs768059814 Chr.11: 3018367-3018367 on Build GRCh38
rs372843593 Chr.11: 3018550-3018550 on Build GRCh38
rs769328079 Chr.11: 3018478-3018478 on Build GRCh38
rs750822353 Chr.11: 3018373-3018373 on Build GRCh38
rs1054700138 Chr.11: 3018493-3018493 on Build GRCh38
rs571697572 Chr.11: 3018426-3018426 on Build GRCh38
rs116421839 Chr.11: 3018551-3018551 on Build GRCh38
rs753911734 Chr.11: 3018394-3018394 on Build GRCh38
rs778899177 Chr.11: 3018403-3018403 on Build GRCh38
rs758364577 Chr.11: 3018424-3018424 on Build GRCh38
rs765518888 Chr.11: 3018552-3018552 on Build GRCh38
rs201215281 Chr.11: 3018509-3018509 on Build GRCh38
rs372923129 Chr.11: 3018365-3018365 on Build GRCh38
rs934355969 Chr.11: 3018463-3018463 on Build GRCh38
rs774915903 Chr.11: 3018443-3018443 on Build GRCh38
rs368933650 Chr.11: 3018555-3018555 on Build GRCh38
rs745573425 Chr.11: 3018521-3018521 on Build GRCh38
rs774767177 Chr.11: 3018363-3018363 on Build GRCh38
rs773892828 Chr.11: 3018539-3018539 on Build GRCh38
rs976715705 Chr.11: 3018375-3018375 on Build GRCh38
rs757095435 Chr.11: 3018511-3018511 on Build GRCh38
rs759001057 Chr.11: 3018358-3018358 on Build GRCh38
rs769490997 Chr.11: 3018528-3018528 on Build GRCh38
rs369480020 Chr.11: 3018537-3018537 on Build GRCh38
rs745359785 Chr.11: 3018435-3018435 on Build GRCh38
rs776070715 Chr.11: 3018357-3018357 on Build GRCh38
rs1038234371 Chr.11: 3018499-3018499 on Build GRCh38
rs746829564 Chr.11: 3018427-3018427 on Build GRCh38
rs762159255 Chr.11: 3018446-3018446 on Build GRCh38

View Full Product Details