Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33394933-33395184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP7
Entrez Gene ID
364
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap, GLYCQTL
UniGene
Hs.455323
Gene Chromosome Location
Chr.9: 33383135-33402682 on Build GRCh38
Transcripts
NM_001170.2 NM_001318156.1 NM_001318157.1 NM_001318158.1 NR_134513.1 NR_134514.1 NR_134515.1 XM_005251453.3 XM_006716765.3 XM_011517866.2 XM_011517867.2 XM_017014700.1 XM_017014701.1 XM_017014702.1 XM_017014703.1 XM_017014704.1 XM_017014705.1 XM_017014706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5932353 c.113C>T p.A38V Chr.9: 33395106-33395106 on Build GRCh38
1489943 c.85_86insGG p.L29fs*15 Chr.9: 33395136-33395137 on Build GRCh38
6183312 c.86A>T p.Q29L Chr.9: 33395133-33395133 on Build GRCh38
4373768 c.100C>A p.R34R Chr.9: 33395119-33395119 on Build GRCh38
1580243 c.142A>T p.M48L Chr.9: 33395080-33395080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756009474 Chr.9: 33395136-33395147 on Build GRCh38
rs764563140 Chr.9: 33395094-33395094 on Build GRCh38
rs373384368 Chr.9: 33395137-33395138 on Build GRCh38
rs145775825 Chr.9: 33395143-33395143 on Build GRCh38
rs776199160 Chr.9: 33395092-33395092 on Build GRCh38
rs766484596 Chr.9: 33395161-33395161 on Build GRCh38
rs2381001 Chr.9: 33395068-33395068 on Build GRCh38
rs745617679 Chr.9: 33395133-33395134 on Build GRCh38
rs192851993 Chr.9: 33395136-33395136 on Build GRCh38
rs570250104 Chr.9: 33395036-33395036 on Build GRCh38
rs771584246 Chr.9: 33395069-33395069 on Build GRCh38
rs747841241 Chr.9: 33395119-33395119 on Build GRCh38
rs72707431 Chr.9: 33395041-33395041 on Build GRCh38
rs976701092 Chr.9: 33395040-33395040 on Build GRCh38
rs377239361 Chr.9: 33395059-33395059 on Build GRCh38
rs2381002 Chr.9: 33395104-33395104 on Build GRCh38
rs543495409 Chr.9: 33395122-33395122 on Build GRCh38
rs72707429 Chr.9: 33395033-33395033 on Build GRCh38
rs760196810 Chr.9: 33395087-33395087 on Build GRCh38
rs558128835 Chr.9: 33395096-33395096 on Build GRCh38
rs761356013 Chr.9: 33395034-33395034 on Build GRCh38
rs779982613 Chr.9: 33395136-33395137 on Build GRCh38
rs767011829 Chr.9: 33394974-33394974 on Build GRCh38
rs774787164 Chr.9: 33395138-33395138 on Build GRCh38
rs753118331 Chr.9: 33395043-33395043 on Build GRCh38
rs2381000 Chr.9: 33395042-33395042 on Build GRCh38
rs536174500 Chr.9: 33395051-33395051 on Build GRCh38
rs79803453 Chr.9: 33395052-33395052 on Build GRCh38
rs1021417721 Chr.9: 33395007-33395007 on Build GRCh38
rs760862357 Chr.9: 33395158-33395158 on Build GRCh38
rs2381004 Chr.9: 33395121-33395121 on Build GRCh38
rs781227239 Chr.9: 33395024-33395024 on Build GRCh38
rs529625335 Chr.9: 33395082-33395082 on Build GRCh38
rs750700265 Chr.9: 33395148-33395148 on Build GRCh38
rs781156267 Chr.9: 33395134-33395134 on Build GRCh38
rs200054774 Chr.9: 33395058-33395058 on Build GRCh38
rs758900960 Chr.9: 33395046-33395046 on Build GRCh38
rs751859079 Chr.9: 33395049-33395049 on Build GRCh38
rs968439382 Chr.9: 33395014-33395014 on Build GRCh38
rs534661939 Chr.9: 33395090-33395090 on Build GRCh38
rs550440525 Chr.9: 33395028-33395028 on Build GRCh38
rs2380999 Chr.9: 33395032-33395032 on Build GRCh38
rs148012859 Chr.9: 33395105-33395105 on Build GRCh38
rs894105429 Chr.9: 33394992-33394992 on Build GRCh38
rs776252656 Chr.9: 33395129-33395129 on Build GRCh38
rs763689078 Chr.9: 33395118-33395118 on Build GRCh38
rs2381003 Chr.9: 33395110-33395110 on Build GRCh38
rs12343286 Chr.9: 33394976-33394976 on Build GRCh38
rs1056815600 Chr.9: 33394983-33394983 on Build GRCh38
rs1003174466 Chr.9: 33394960-33394960 on Build GRCh38
rs757394464 Chr.9: 33395050-33395050 on Build GRCh38
rs745850826 Chr.9: 33395027-33395027 on Build GRCh38
rs762121366 Chr.9: 33395099-33395099 on Build GRCh38
rs539538326 Chr.9: 33394967-33394967 on Build GRCh38
rs780091659 Chr.9: 33395109-33395109 on Build GRCh38
rs773993278 Chr.9: 33395061-33395061 on Build GRCh38
rs373620197 Chr.9: 33395162-33395162 on Build GRCh38
rs201476223 Chr.9: 33395037-33395037 on Build GRCh38
rs756053895 Chr.9: 33395054-33395054 on Build GRCh38
rs775283438 Chr.9: 33395030-33395030 on Build GRCh38
rs767860854 Chr.9: 33395144-33395144 on Build GRCh38
rs772863468 Chr.9: 33395077-33395077 on Build GRCh38
rs2381005 Chr.9: 33395137-33395137 on Build GRCh38
rs3860978 Chr.9: 33395012-33395012 on Build GRCh38
rs1012677516 Chr.9: 33395006-33395006 on Build GRCh38

View Full Product Details