Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8330105-8330344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC45A1
Entrez Gene ID
50651
Gene Name
solute carrier family 45 member 1
Gene Aliases
DNB5
UniGene
Hs.463036
Gene Chromosome Location
Chr.1: 8318085-8344167 on Build GRCh38
Transcripts
NM_001080397.2 XM_011541530.1 XM_011541531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6064142 c.841G>A p.V281M Chr.1: 8330232-8330232 on Build GRCh38
6064141 c.739G>A p.V247M Chr.1: 8330232-8330232 on Build GRCh38
4849153 c.915C>T p.L305L Chr.1: 8330306-8330306 on Build GRCh38
5431235 c.720C>T p.L240L Chr.1: 8330213-8330213 on Build GRCh38
6127433 c.849C>T p.G283G Chr.1: 8330240-8330240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748362131 Chr.1: 8330314-8330314 on Build GRCh38
rs147116462 Chr.1: 8330279-8330279 on Build GRCh38
rs1010050365 Chr.1: 8330173-8330173 on Build GRCh38
rs12565834 Chr.1: 8330186-8330186 on Build GRCh38
rs552752178 Chr.1: 8330154-8330154 on Build GRCh38
rs775819094 Chr.1: 8330234-8330234 on Build GRCh38
rs577905830 Chr.1: 8330195-8330195 on Build GRCh38
rs746271502 Chr.1: 8330166-8330168 on Build GRCh38
rs771912671 Chr.1: 8330204-8330204 on Build GRCh38
rs1054247883 Chr.1: 8330143-8330143 on Build GRCh38
rs201967766 Chr.1: 8330237-8330237 on Build GRCh38
rs138517419 Chr.1: 8330303-8330303 on Build GRCh38
rs746636610 Chr.1: 8330318-8330318 on Build GRCh38
rs201971465 Chr.1: 8330317-8330317 on Build GRCh38
rs750596930 Chr.1: 8330299-8330299 on Build GRCh38
rs779136241 Chr.1: 8330243-8330243 on Build GRCh38
rs1018026210 Chr.1: 8330217-8330217 on Build GRCh38
rs770516473 Chr.1: 8330268-8330268 on Build GRCh38
rs150216502 Chr.1: 8330267-8330267 on Build GRCh38
rs763910011 Chr.1: 8330293-8330293 on Build GRCh38
rs56125713 Chr.1: 8330324-8330324 on Build GRCh38
rs761337589 Chr.1: 8330197-8330197 on Build GRCh38
rs780476814 Chr.1: 8330180-8330180 on Build GRCh38
rs143965615 Chr.1: 8330327-8330327 on Build GRCh38
rs1025564135 Chr.1: 8330184-8330184 on Build GRCh38
rs191155736 Chr.1: 8330234-8330234 on Build GRCh38
rs775305042 Chr.1: 8330280-8330280 on Build GRCh38
rs368791442 Chr.1: 8330174-8330174 on Build GRCh38
rs200094048 Chr.1: 8330307-8330307 on Build GRCh38
rs200341486 Chr.1: 8330213-8330213 on Build GRCh38
rs755422661 Chr.1: 8330311-8330311 on Build GRCh38
rs186437701 Chr.1: 8330231-8330231 on Build GRCh38
rs144351866 Chr.1: 8330315-8330315 on Build GRCh38
rs370872627 Chr.1: 8330205-8330205 on Build GRCh38
rs151069350 Chr.1: 8330261-8330261 on Build GRCh38
rs2289730 Chr.1: 8330153-8330153 on Build GRCh38
rs760204807 Chr.1: 8330207-8330207 on Build GRCh38
rs768857561 Chr.1: 8330192-8330192 on Build GRCh38
rs772931544 Chr.1: 8330263-8330263 on Build GRCh38
rs376145020 Chr.1: 8330169-8330169 on Build GRCh38
rs150790768 Chr.1: 8330260-8330260 on Build GRCh38
rs950286511 Chr.1: 8330274-8330274 on Build GRCh38
rs757409091 Chr.1: 8330232-8330232 on Build GRCh38
rs769241812 Chr.1: 8330240-8330240 on Build GRCh38
rs34646197 Chr.1: 8330275-8330275 on Build GRCh38
rs764910891 Chr.1: 8330229-8330229 on Build GRCh38
rs144740978 Chr.1: 8330219-8330219 on Build GRCh38
rs781246207 Chr.1: 8330164-8330164 on Build GRCh38
rs1007002791 Chr.1: 8330128-8330128 on Build GRCh38
rs554626444 Chr.1: 8330238-8330238 on Build GRCh38
rs766341691 Chr.1: 8330306-8330306 on Build GRCh38
rs762670620 Chr.1: 8330292-8330292 on Build GRCh38
rs768998199 Chr.1: 8330273-8330273 on Build GRCh38
rs776253005 Chr.1: 8330272-8330272 on Build GRCh38

View Full Product Details