Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44777579-44777852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS8
Entrez Gene ID
6202
Gene Name
ribosomal protein S8
Gene Aliases
S8
UniGene
Hs.512675
Gene Chromosome Location
Chr.1: 44775574-44778740 on Build GRCh38
Transcripts
NM_001012.1 NR_001456.1

Gene Symbol
SNORD38A
Entrez Gene ID
94162
Gene Name
small nucleolar RNA, C/D box 38A
Gene Aliases
RNU38A, U38A
UniGene
Gene Chromosome Location
Chr.1: 44777842-44777912 on Build GRCh38
Transcripts
NM_001012.1 NR_001456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118653 c.363G>T p.L121L Chr.1: 44777765-44777765 on Build GRCh38
3805379 c.213T>G p.C71W Chr.1: 44777615-44777615 on Build GRCh38
5754460 c.326A>G p.Y109C Chr.1: 44777728-44777728 on Build GRCh38
681209 c.366C>T p.G122G Chr.1: 44777768-44777768 on Build GRCh38
4165204 c.291G>A p.V97V Chr.1: 44777693-44777693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770526961 Chr.1: 44777813-44777814 on Build GRCh38
rs201608642 Chr.1: 44777830-44777830 on Build GRCh38
rs773875567 Chr.1: 44777711-44777711 on Build GRCh38
rs748034169 Chr.1: 44777657-44777657 on Build GRCh38
rs757691384 Chr.1: 44777752-44777752 on Build GRCh38
rs748169878 Chr.1: 44777793-44777793 on Build GRCh38
rs900337622 Chr.1: 44777679-44777679 on Build GRCh38
rs755155687 Chr.1: 44777776-44777776 on Build GRCh38
rs772973833 Chr.1: 44777663-44777663 on Build GRCh38
rs750781753 Chr.1: 44777756-44777756 on Build GRCh38
rs765976878 Chr.1: 44777705-44777705 on Build GRCh38
rs879107881 Chr.1: 44777790-44777790 on Build GRCh38
rs11537864 Chr.1: 44777667-44777667 on Build GRCh38
rs11537868 Chr.1: 44777706-44777706 on Build GRCh38
rs145889888 Chr.1: 44777694-44777694 on Build GRCh38
rs78279677 Chr.1: 44777726-44777726 on Build GRCh38
rs769592026 Chr.1: 44777801-44777801 on Build GRCh38
rs995974053 Chr.1: 44777723-44777723 on Build GRCh38
rs374555700 Chr.1: 44777623-44777623 on Build GRCh38
rs781121879 Chr.1: 44777786-44777786 on Build GRCh38
rs773022878 Chr.1: 44777808-44777808 on Build GRCh38
rs769691487 Chr.1: 44777662-44777662 on Build GRCh38
rs780316149 Chr.1: 44777611-44777611 on Build GRCh38
rs878910802 Chr.1: 44777791-44777791 on Build GRCh38
rs11537863 Chr.1: 44777669-44777669 on Build GRCh38
rs768497991 Chr.1: 44777634-44777634 on Build GRCh38
rs956352747 Chr.1: 44777818-44777818 on Build GRCh38
rs25687 Chr.1: 44777639-44777639 on Build GRCh38
rs747096386 Chr.1: 44777771-44777771 on Build GRCh38
rs780371213 Chr.1: 44777759-44777759 on Build GRCh38
rs374348518 Chr.1: 44777828-44777828 on Build GRCh38
rs553120843 Chr.1: 44777681-44777681 on Build GRCh38
rs754433878 Chr.1: 44777748-44777748 on Build GRCh38
rs749034987 Chr.1: 44777811-44777811 on Build GRCh38
rs764510923 Chr.1: 44777739-44777739 on Build GRCh38
rs375461739 Chr.1: 44777714-44777714 on Build GRCh38
rs1031658374 Chr.1: 44777762-44777762 on Build GRCh38
rs141076332 Chr.1: 44777666-44777666 on Build GRCh38
rs1022232205 Chr.1: 44777725-44777725 on Build GRCh38
rs765682906 Chr.1: 44777755-44777755 on Build GRCh38
rs11537870 Chr.1: 44777730-44777730 on Build GRCh38
rs758720829 Chr.1: 44777757-44777757 on Build GRCh38
rs769984020 Chr.1: 44777807-44777807 on Build GRCh38
rs879235759 Chr.1: 44777792-44777792 on Build GRCh38

View Full Product Details