Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 115113099-115113338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH2
Entrez Gene ID
57631
Gene Name
leucine rich repeats and calponin homology domain containing 2
Gene Aliases
dA204F4.4
UniGene
Hs.65366
Gene Chromosome Location
Chr.X: 115110611-115234082 on Build GRCh38
Transcripts
NM_001243963.1 NM_020871.3 XM_006724724.3

SNP Detail

SNP ID Chromosome Location
rs782341959 Chr.X: 115113131-115113131 on Build GRCh38
rs199995730 Chr.X: 115113209-115113209 on Build GRCh38
rs782569379 Chr.X: 115113226-115113226 on Build GRCh38
rs782339786 Chr.X: 115113238-115113238 on Build GRCh38
rs369392135 Chr.X: 115113291-115113291 on Build GRCh38
rs201883620 Chr.X: 115113284-115113284 on Build GRCh38
rs782271580 Chr.X: 115113185-115113185 on Build GRCh38
rs192687026 Chr.X: 115113169-115113169 on Build GRCh38
rs41304492 Chr.X: 115113143-115113143 on Build GRCh38
rs782420699 Chr.X: 115113307-115113307 on Build GRCh38
rs143396803 Chr.X: 115113132-115113132 on Build GRCh38
rs375999930 Chr.X: 115113231-115113231 on Build GRCh38

View Full Product Details