Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51416549-51416818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129083
Entrez Gene ID
100129083
Gene Name
uncharacterized LOC100129083
Gene Aliases
-
UniGene
Hs.284813
Gene Chromosome Location
Chr.19: 51415906-51417425 on Build GRCh38
Transcripts
NM_001256795.1 NM_001171156.1 NM_001171157.1 NM_001171158.1 NM_001171159.1 NM_001171161.1 NM_001322105.1 NM_033130.4 XM_005259366.4 XM_005259367.3 XM_011527439.1

Gene Symbol
SIGLEC10
Entrez Gene ID
89790
Gene Name
sialic acid binding Ig like lectin 10
Gene Aliases
PRO940, SIGLEC-10, SLG2
UniGene
Hs.284813
Gene Chromosome Location
Chr.19: 51410021-51417803 on Build GRCh38
Transcripts
NM_001256795.1 NM_001171156.1 NM_001171157.1 NM_001171158.1 NM_001171159.1 NM_001171161.1 NM_001322105.1 NM_033130.4 XM_005259366.4 XM_005259367.3 XM_011527439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960350 c.422-5C>G p.? Chr.19: 51416781-51416781 on Build GRCh38
7236257 c.422-5C>A p.? Chr.19: 51416781-51416781 on Build GRCh38
440110 c.606C>A p.F202L Chr.19: 51416766-51416766 on Build GRCh38
6382302 c.677C>T p.A226V Chr.19: 51416695-51416695 on Build GRCh38
228720 c.615A>C p.R205S Chr.19: 51416757-51416757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149388582 Chr.19: 51416731-51416731 on Build GRCh38
rs201392477 Chr.19: 51416764-51416764 on Build GRCh38
rs768791040 Chr.19: 51416774-51416774 on Build GRCh38
rs752673152 Chr.19: 51416568-51416568 on Build GRCh38
rs747987229 Chr.19: 51416778-51416778 on Build GRCh38
rs373611666 Chr.19: 51416625-51416625 on Build GRCh38
rs754227847 Chr.19: 51416740-51416740 on Build GRCh38
rs111865145 Chr.19: 51416640-51416640 on Build GRCh38
rs867779814 Chr.19: 51416789-51416789 on Build GRCh38
rs760706201 Chr.19: 51416569-51416569 on Build GRCh38
rs546225166 Chr.19: 51416756-51416756 on Build GRCh38
rs61741677 Chr.19: 51416744-51416744 on Build GRCh38
rs555268636 Chr.19: 51416675-51416675 on Build GRCh38
rs778070006 Chr.19: 51416603-51416603 on Build GRCh38
rs764884110 Chr.19: 51416717-51416717 on Build GRCh38
rs760652924 Chr.19: 51416626-51416626 on Build GRCh38
rs780414009 Chr.19: 51416763-51416763 on Build GRCh38
rs565321156 Chr.19: 51416591-51416591 on Build GRCh38
rs539060597 Chr.19: 51416597-51416597 on Build GRCh38
rs758084278 Chr.19: 51416642-51416642 on Build GRCh38
rs749376631 Chr.19: 51416659-51416662 on Build GRCh38
rs71358836 Chr.19: 51416598-51416598 on Build GRCh38
rs763937261 Chr.19: 51416571-51416571 on Build GRCh38
rs749862287 Chr.19: 51416677-51416677 on Build GRCh38
rs765826865 Chr.19: 51416662-51416662 on Build GRCh38
rs569661331 Chr.19: 51416618-51416618 on Build GRCh38
rs757792226 Chr.19: 51416745-51416745 on Build GRCh38
rs768915755 Chr.19: 51416696-51416696 on Build GRCh38
rs751260939 Chr.19: 51416732-51416732 on Build GRCh38
rs770768237 Chr.19: 51416797-51416797 on Build GRCh38
rs753720011 Chr.19: 51416581-51416581 on Build GRCh38
rs776706879 Chr.19: 51416697-51416697 on Build GRCh38
rs781346556 Chr.19: 51416777-51416777 on Build GRCh38
rs776761821 Chr.19: 51416630-51416630 on Build GRCh38
rs71358835 Chr.19: 51416586-51416586 on Build GRCh38
rs754521676 Chr.19: 51416674-51416674 on Build GRCh38
rs757554352 Chr.19: 51416680-51416680 on Build GRCh38
rs557047713 Chr.19: 51416684-51416684 on Build GRCh38
rs746164618 Chr.19: 51416620-51416620 on Build GRCh38
rs764094700 Chr.19: 51416628-51416628 on Build GRCh38
rs751099328 Chr.19: 51416672-51416672 on Build GRCh38
rs754468151 Chr.19: 51416738-51416738 on Build GRCh38
rs779111009 Chr.19: 51416750-51416750 on Build GRCh38
rs370940644 Chr.19: 51416730-51416730 on Build GRCh38
rs761815082 Chr.19: 51416702-51416702 on Build GRCh38
rs148042318 Chr.19: 51416757-51416757 on Build GRCh38
rs199986765 Chr.19: 51416739-51416739 on Build GRCh38
rs769800706 Chr.19: 51416793-51416793 on Build GRCh38
rs761501008 Chr.19: 51416639-51416639 on Build GRCh38
rs770306736 Chr.19: 51416601-51416601 on Build GRCh38
rs749977708 Chr.19: 51416641-51416641 on Build GRCh38
rs573808895 Chr.19: 51416685-51416685 on Build GRCh38
rs9304711 Chr.19: 51416695-51416695 on Build GRCh38
rs755494017 Chr.19: 51416745-51416747 on Build GRCh38
rs375483042 Chr.19: 51416794-51416794 on Build GRCh38
rs771372573 Chr.19: 51416617-51416617 on Build GRCh38
rs779337135 Chr.19: 51416681-51416681 on Build GRCh38
rs757119053 Chr.19: 51416583-51416583 on Build GRCh38
rs773081551 Chr.19: 51416727-51416727 on Build GRCh38
rs900819415 Chr.19: 51416580-51416580 on Build GRCh38
rs749620128 Chr.19: 51416606-51416606 on Build GRCh38
rs750150502 Chr.19: 51416593-51416593 on Build GRCh38

View Full Product Details