Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93794795-93795054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED17
Entrez Gene ID
9440
Gene Name
mediator complex subunit 17
Gene Aliases
CRSP6, CRSP77, DRIP80, TRAP80
UniGene
Hs.444931
Gene Chromosome Location
Chr.11: 93784239-93813330 on Build GRCh38
Transcripts
NM_004268.4 XM_011543068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700167 c.994A>G p.I332V Chr.11: 93795042-93795042 on Build GRCh38
4456301 c.1004C>T p.P335L Chr.11: 93795052-93795052 on Build GRCh38
6070783 c.963A>T p.Q321H Chr.11: 93795011-93795011 on Build GRCh38
7424354 c.867A>T p.P289P Chr.11: 93794915-93794915 on Build GRCh38
5360407 c.982delA p.N329fs*43 Chr.11: 93795030-93795030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773324510 Chr.11: 93794891-93794891 on Build GRCh38
rs756613156 Chr.11: 93794955-93794955 on Build GRCh38
rs368306104 Chr.11: 93794908-93794908 on Build GRCh38
rs766241332 Chr.11: 93795027-93795027 on Build GRCh38
rs1021508489 Chr.11: 93794841-93794841 on Build GRCh38
rs2276311 Chr.11: 93795026-93795026 on Build GRCh38
rs780547843 Chr.11: 93794958-93794958 on Build GRCh38
rs374004657 Chr.11: 93794871-93794871 on Build GRCh38
rs956100028 Chr.11: 93795021-93795021 on Build GRCh38
rs751313909 Chr.11: 93794986-93794986 on Build GRCh38
rs111678421 Chr.11: 93794828-93794828 on Build GRCh38
rs766055940 Chr.11: 93794888-93794888 on Build GRCh38
rs369672198 Chr.11: 93795015-93795015 on Build GRCh38
rs200790049 Chr.11: 93794975-93794975 on Build GRCh38
rs997048492 Chr.11: 93794833-93794833 on Build GRCh38
rs370657428 Chr.11: 93794869-93794869 on Build GRCh38
rs143018025 Chr.11: 93794938-93794938 on Build GRCh38
rs746341041 Chr.11: 93794861-93794861 on Build GRCh38
rs762043140 Chr.11: 93794887-93794887 on Build GRCh38
rs749631821 Chr.11: 93794964-93794964 on Build GRCh38
rs781710873 Chr.11: 93794860-93794860 on Build GRCh38
rs375348736 Chr.11: 93794946-93794946 on Build GRCh38
rs200139509 Chr.11: 93794913-93794913 on Build GRCh38
rs774578517 Chr.11: 93794885-93794885 on Build GRCh38
rs768769792 Chr.11: 93794884-93794884 on Build GRCh38
rs749627360 Chr.11: 93794880-93794880 on Build GRCh38
rs773455328 Chr.11: 93795020-93795020 on Build GRCh38
rs866980922 Chr.11: 93794979-93794980 on Build GRCh38
rs772329319 Chr.11: 93795009-93795009 on Build GRCh38
rs754743877 Chr.11: 93794932-93794932 on Build GRCh38
rs777239716 Chr.11: 93794939-93794939 on Build GRCh38
rs577266297 Chr.11: 93794999-93794999 on Build GRCh38
rs774781719 Chr.11: 93794992-93794992 on Build GRCh38
rs372057965 Chr.11: 93794937-93794937 on Build GRCh38
rs757886740 Chr.11: 93794858-93794858 on Build GRCh38
rs748351387 Chr.11: 93795006-93795006 on Build GRCh38
rs536602728 Chr.11: 93794843-93794843 on Build GRCh38
rs147992585 Chr.11: 93794889-93794889 on Build GRCh38
rs533311520 Chr.11: 93794857-93794857 on Build GRCh38

View Full Product Details