Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159535212-159535459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10J5
Entrez Gene ID
127385
Gene Name
olfactory receptor family 10 subfamily J member 5
Gene Aliases
OR1-28
UniGene
Hs.553587
Gene Chromosome Location
Chr.1: 159535078-159536007 on Build GRCh38
Transcripts
NM_001004469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6459484 c.627C>T p.P209P Chr.1: 159535381-159535381 on Build GRCh38
5132510 c.735T>C p.T245T Chr.1: 159535273-159535273 on Build GRCh38
1335373 c.669C>G p.S223S Chr.1: 159535339-159535339 on Build GRCh38
5032712 c.604A>C p.S202R Chr.1: 159535404-159535404 on Build GRCh38
385807 c.782C>T p.P261L Chr.1: 159535226-159535226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765687991 Chr.1: 159535294-159535294 on Build GRCh38
rs763491362 Chr.1: 159535306-159535306 on Build GRCh38
rs771058077 Chr.1: 159535353-159535353 on Build GRCh38
rs558725254 Chr.1: 159535317-159535317 on Build GRCh38
rs141462119 Chr.1: 159535310-159535310 on Build GRCh38
rs139662085 Chr.1: 159535422-159535422 on Build GRCh38
rs746689307 Chr.1: 159535379-159535379 on Build GRCh38
rs770118223 Chr.1: 159535381-159535381 on Build GRCh38
rs775232295 Chr.1: 159535376-159535376 on Build GRCh38
rs973892154 Chr.1: 159535289-159535289 on Build GRCh38
rs766887460 Chr.1: 159535409-159535409 on Build GRCh38
rs745390587 Chr.1: 159535374-159535374 on Build GRCh38
rs776796325 Chr.1: 159535275-159535275 on Build GRCh38
rs1049910396 Chr.1: 159535300-159535300 on Build GRCh38
rs113532286 Chr.1: 159535238-159535238 on Build GRCh38
rs1043861225 Chr.1: 159535407-159535407 on Build GRCh38
rs35393723 Chr.1: 159535311-159535311 on Build GRCh38
rs750911188 Chr.1: 159535298-159535298 on Build GRCh38
rs973715167 Chr.1: 159535256-159535256 on Build GRCh38
rs773636771 Chr.1: 159535394-159535394 on Build GRCh38
rs772585571 Chr.1: 159535362-159535362 on Build GRCh38
rs752937659 Chr.1: 159535415-159535415 on Build GRCh38
rs762272541 Chr.1: 159535280-159535280 on Build GRCh38
rs867721846 Chr.1: 159535303-159535303 on Build GRCh38
rs755991763 Chr.1: 159535336-159535336 on Build GRCh38
rs866207118 Chr.1: 159535320-159535320 on Build GRCh38
rs771586405 Chr.1: 159535375-159535375 on Build GRCh38
rs774041317 Chr.1: 159535234-159535234 on Build GRCh38
rs374885592 Chr.1: 159535359-159535359 on Build GRCh38
rs895397289 Chr.1: 159535365-159535365 on Build GRCh38
rs781479522 Chr.1: 159535324-159535324 on Build GRCh38
rs577028454 Chr.1: 159535338-159535338 on Build GRCh38
rs774226295 Chr.1: 159535410-159535410 on Build GRCh38
rs372971358 Chr.1: 159535358-159535358 on Build GRCh38
rs555085601 Chr.1: 159535262-159535262 on Build GRCh38
rs775955636 Chr.1: 159535368-159535369 on Build GRCh38
rs747807028 Chr.1: 159535405-159535406 on Build GRCh38
rs760718758 Chr.1: 159535406-159535406 on Build GRCh38
rs746356653 Chr.1: 159535302-159535304 on Build GRCh38
rs771812122 Chr.1: 159535265-159535265 on Build GRCh38
rs749206305 Chr.1: 159535350-159535350 on Build GRCh38
rs56786307 Chr.1: 159535412-159535412 on Build GRCh38
rs544179663 Chr.1: 159535411-159535411 on Build GRCh38
rs759103313 Chr.1: 159535254-159535254 on Build GRCh38
rs932431583 Chr.1: 159535278-159535278 on Build GRCh38
rs752391373 Chr.1: 159535328-159535328 on Build GRCh38
rs763976151 Chr.1: 159535425-159535425 on Build GRCh38

View Full Product Details