Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40415322-40415589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IVD
Entrez Gene ID
3712
Gene Name
isovaleryl-CoA dehydrogenase
Gene Aliases
ACAD2
UniGene
Hs.513646
Gene Chromosome Location
Chr.15: 40405485-40435948 on Build GRCh38
Transcripts
NM_001159508.1 NM_002225.3 XM_005254350.2 XM_006720492.2 XM_006720495.2 XM_017022149.1 XM_017022150.1 XM_017022151.1 XM_017022152.1 XM_017022153.1 XM_017022154.1 XM_017022155.1 XM_017022156.1 XM_017022157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1708012 c.919C>T p.P307S Chr.15: 40415432-40415432 on Build GRCh38
5596890 c.942C>T p.A314A Chr.15: 40415455-40415455 on Build GRCh38
3500891 c.967C>T p.Q323* Chr.15: 40415480-40415480 on Build GRCh38
5054265 c.900G>A p.A300A Chr.15: 40415413-40415413 on Build GRCh38
5383221 c.920C>T p.P307L Chr.15: 40415433-40415433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761076572 Chr.15: 40415400-40415400 on Build GRCh38
rs1014196997 Chr.15: 40415548-40415548 on Build GRCh38
rs367814475 Chr.15: 40415482-40415482 on Build GRCh38
rs941878883 Chr.15: 40415358-40415358 on Build GRCh38
rs531429598 Chr.15: 40415490-40415490 on Build GRCh38
rs751049911 Chr.15: 40415419-40415419 on Build GRCh38
rs770558545 Chr.15: 40415509-40415509 on Build GRCh38
rs772633374 Chr.15: 40415389-40415389 on Build GRCh38
rs778109931 Chr.15: 40415377-40415377 on Build GRCh38
rs750569823 Chr.15: 40415396-40415396 on Build GRCh38
rs777379216 Chr.15: 40415455-40415455 on Build GRCh38
rs377570597 Chr.15: 40415360-40415360 on Build GRCh38
rs771061679 Chr.15: 40415380-40415380 on Build GRCh38
rs776468115 Chr.15: 40415373-40415373 on Build GRCh38
rs757967808 Chr.15: 40415353-40415353 on Build GRCh38
rs375048123 Chr.15: 40415352-40415352 on Build GRCh38
rs148072949 Chr.15: 40415444-40415444 on Build GRCh38
rs113361784 Chr.15: 40415489-40415489 on Build GRCh38
rs754100750 Chr.15: 40415470-40415470 on Build GRCh38
rs767108729 Chr.15: 40415522-40415522 on Build GRCh38
rs759116078 Chr.15: 40415425-40415425 on Build GRCh38
rs117793361 Chr.15: 40415558-40415558 on Build GRCh38
rs1044885399 Chr.15: 40415347-40415347 on Build GRCh38
rs375250023 Chr.15: 40415382-40415382 on Build GRCh38
rs755470515 Chr.15: 40415543-40415543 on Build GRCh38
rs138427412 Chr.15: 40415413-40415413 on Build GRCh38
rs754906922 Chr.15: 40415362-40415362 on Build GRCh38
rs73385117 Chr.15: 40415564-40415564 on Build GRCh38
rs752699525 Chr.15: 40415529-40415529 on Build GRCh38
rs762673197 Chr.15: 40415506-40415506 on Build GRCh38
rs564690986 Chr.15: 40415410-40415410 on Build GRCh38
rs757351801 Chr.15: 40415480-40415480 on Build GRCh38
rs28940889 Chr.15: 40415454-40415454 on Build GRCh38
rs781141864 Chr.15: 40415368-40415368 on Build GRCh38
rs796051983 Chr.15: 40415412-40415412 on Build GRCh38
rs149247895 Chr.15: 40415417-40415417 on Build GRCh38
rs760468999 Chr.15: 40415530-40415530 on Build GRCh38
rs572691367 Chr.15: 40415375-40415375 on Build GRCh38
rs144418134 Chr.15: 40415438-40415438 on Build GRCh38
rs547914696 Chr.15: 40415510-40415510 on Build GRCh38
rs772115680 Chr.15: 40415485-40415485 on Build GRCh38
rs776804343 Chr.15: 40415491-40415491 on Build GRCh38
rs370481272 Chr.15: 40415378-40415378 on Build GRCh38
rs759169323 Chr.15: 40415520-40415520 on Build GRCh38
rs941828243 Chr.15: 40415561-40415561 on Build GRCh38

View Full Product Details