Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127480999-127481272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IWS1
Entrez Gene ID
55677
Gene Name
IWS1, SUPT6H interacting protein
Gene Aliases
-
UniGene
Hs.469879
Gene Chromosome Location
Chr.2: 127480807-127526886 on Build GRCh38
Transcripts
NM_017969.2 XM_005263707.3 XM_006712626.3 XM_017004467.1 XM_017004468.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5621064 c.2452C>T p.P818S Chr.2: 127481052-127481052 on Build GRCh38
3042333 c.2404_2405insA p.S802fs*12 Chr.2: 127481099-127481100 on Build GRCh38
4819264 c.2389G>A p.D797N Chr.2: 127481115-127481115 on Build GRCh38
4642482 c.2398A>G p.K800E Chr.2: 127481106-127481106 on Build GRCh38
4185680 c.2353A>G p.I785V Chr.2: 127481151-127481151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746341246 Chr.2: 127481207-127481207 on Build GRCh38
rs1047793197 Chr.2: 127481089-127481089 on Build GRCh38
rs944265258 Chr.2: 127481144-127481144 on Build GRCh38
rs764739153 Chr.2: 127481083-127481083 on Build GRCh38
rs752965778 Chr.2: 127481038-127481038 on Build GRCh38
rs185201768 Chr.2: 127481162-127481162 on Build GRCh38
rs371511015 Chr.2: 127481026-127481026 on Build GRCh38
rs749577368 Chr.2: 127481168-127481168 on Build GRCh38
rs559974254 Chr.2: 127481231-127481231 on Build GRCh38
rs762745846 Chr.2: 127481224-127481224 on Build GRCh38
rs200512921 Chr.2: 127481213-127481213 on Build GRCh38
rs368270703 Chr.2: 127481217-127481217 on Build GRCh38
rs375178274 Chr.2: 127481199-127481199 on Build GRCh38
rs140149850 Chr.2: 127481111-127481111 on Build GRCh38
rs751070474 Chr.2: 127481226-127481226 on Build GRCh38
rs755008316 Chr.2: 127481104-127481104 on Build GRCh38
rs996602495 Chr.2: 127481180-127481180 on Build GRCh38
rs138004420 Chr.2: 127481167-127481167 on Build GRCh38
rs776503180 Chr.2: 127481123-127481123 on Build GRCh38
rs774461386 Chr.2: 127481204-127481204 on Build GRCh38
rs753544801 Chr.2: 127481155-127481156 on Build GRCh38
rs1024938468 Chr.2: 127481247-127481247 on Build GRCh38
rs144079075 Chr.2: 127481218-127481218 on Build GRCh38
rs532214439 Chr.2: 127481244-127481244 on Build GRCh38
rs761126248 Chr.2: 127481067-127481067 on Build GRCh38
rs151146769 Chr.2: 127481095-127481095 on Build GRCh38
rs750095579 Chr.2: 127481085-127481085 on Build GRCh38
rs781217062 Chr.2: 127481108-127481108 on Build GRCh38
rs756790547 Chr.2: 127481176-127481177 on Build GRCh38
rs755945966 Chr.2: 127481153-127481153 on Build GRCh38
rs765959833 Chr.2: 127481225-127481225 on Build GRCh38
rs558416528 Chr.2: 127481101-127481101 on Build GRCh38
rs766048884 Chr.2: 127481099-127481099 on Build GRCh38
rs767785597 Chr.2: 127481246-127481246 on Build GRCh38

View Full Product Details