Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50625444-50625669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSA
Entrez Gene ID
410
Gene Name
arylsulfatase A
Gene Aliases
MLD
UniGene
Hs.731715
Gene Chromosome Location
Chr.22: 50622754-50628173 on Build GRCh38
Transcripts
NM_000487.5 NM_001085425.2 NM_001085426.2 NM_001085427.2 NM_001085428.2 XM_011530690.2 XM_011530691.2 XM_017028800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5133568 c.1175G>A p.R392Q Chr.22: 50625614-50625614 on Build GRCh38
6605260 c.1130_1132delTCT p.F377delF Chr.22: 50625657-50625659 on Build GRCh38
5638708 c.1189A>G p.K397E Chr.22: 50625600-50625600 on Build GRCh38
6354175 c.1178C>G p.T393S Chr.22: 50625611-50625611 on Build GRCh38
5393922 c.1121C>T p.S374F Chr.22: 50625668-50625668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6151426 Chr.22: 50625566-50625566 on Build GRCh38
rs187599274 Chr.22: 50625531-50625531 on Build GRCh38
rs762674 Chr.22: 50625559-50625559 on Build GRCh38
rs1007814225 Chr.22: 50625473-50625473 on Build GRCh38
rs74315479 Chr.22: 50625639-50625639 on Build GRCh38
rs759592253 Chr.22: 50625492-50625492 on Build GRCh38
rs398123413 Chr.22: 50625474-50625474 on Build GRCh38
rs747155690 Chr.22: 50625632-50625632 on Build GRCh38
rs373219209 Chr.22: 50625609-50625609 on Build GRCh38
rs770243080 Chr.22: 50625468-50625468 on Build GRCh38
rs368163068 Chr.22: 50625530-50625530 on Build GRCh38
rs1002298028 Chr.22: 50625623-50625623 on Build GRCh38
rs752151238 Chr.22: 50625535-50625535 on Build GRCh38
rs199476370 Chr.22: 50625633-50625633 on Build GRCh38
rs750260214 Chr.22: 50625508-50625508 on Build GRCh38
rs746072398 Chr.22: 50625554-50625554 on Build GRCh38
rs372705596 Chr.22: 50625561-50625561 on Build GRCh38
rs369490134 Chr.22: 50625583-50625583 on Build GRCh38
rs6151425 Chr.22: 50625640-50625640 on Build GRCh38
rs752206330 Chr.22: 50625592-50625592 on Build GRCh38
rs199476376 Chr.22: 50625594-50625594 on Build GRCh38
rs74315480 Chr.22: 50625615-50625615 on Build GRCh38
rs756625827 Chr.22: 50625542-50625542 on Build GRCh38
rs904986667 Chr.22: 50625622-50625622 on Build GRCh38
rs754346314 Chr.22: 50625549-50625549 on Build GRCh38
rs743616 Chr.22: 50625611-50625611 on Build GRCh38
rs772146580 Chr.22: 50625555-50625555 on Build GRCh38
rs568502611 Chr.22: 50625551-50625551 on Build GRCh38
rs963103843 Chr.22: 50625509-50625509 on Build GRCh38
rs761875290 Chr.22: 50625505-50625505 on Build GRCh38
rs748620030 Chr.22: 50625467-50625467 on Build GRCh38
rs199476376 Chr.22: 50625594-50625594 on Build GRCh38
rs373829750 Chr.22: 50625550-50625550 on Build GRCh38
rs747030880 Chr.22: 50625558-50625558 on Build GRCh38
rs111508121 Chr.22: 50625577-50625577 on Build GRCh38
rs755522172 Chr.22: 50625549-50625550 on Build GRCh38
rs373961176 Chr.22: 50625502-50625502 on Build GRCh38
rs74925710 Chr.22: 50625466-50625466 on Build GRCh38
rs570701266 Chr.22: 50625516-50625516 on Build GRCh38
rs199476391 Chr.22: 50625614-50625614 on Build GRCh38
rs910692005 Chr.22: 50625520-50625520 on Build GRCh38
rs968267549 Chr.22: 50625495-50625495 on Build GRCh38
rs374150980 Chr.22: 50625479-50625479 on Build GRCh38
rs1021214650 Chr.22: 50625471-50625471 on Build GRCh38
rs778131215 Chr.22: 50625469-50625469 on Build GRCh38
rs979995446 Chr.22: 50625502-50625503 on Build GRCh38
rs762744940 Chr.22: 50625515-50625515 on Build GRCh38
rs557367346 Chr.22: 50625565-50625565 on Build GRCh38
rs772169618 Chr.22: 50625501-50625501 on Build GRCh38
rs757603437 Chr.22: 50625612-50625612 on Build GRCh38
rs80338820 Chr.22: 50625578-50625578 on Build GRCh38
rs149088203 Chr.22: 50625634-50625634 on Build GRCh38

View Full Product Details