Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44104747-44104986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEBP1
Entrez Gene ID
165
Gene Name
AE binding protein 1
Gene Aliases
ACLP
UniGene
Hs.439463
Gene Chromosome Location
Chr.7: 44104320-44114565 on Build GRCh38
Transcripts
NM_001129.4 XM_011515162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321303 c.126G>T p.L42L Chr.7: 44104791-44104791 on Build GRCh38
5476765 c.156C>T p.D52D Chr.7: 44104821-44104821 on Build GRCh38
4164346 c.247G>A p.A83T Chr.7: 44104912-44104912 on Build GRCh38
3849704 c.199C>T p.R67* Chr.7: 44104864-44104864 on Build GRCh38
6177805 c.149G>A p.R50Q Chr.7: 44104814-44104814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773952615 Chr.7: 44104901-44104901 on Build GRCh38
rs112298043 Chr.7: 44104878-44104878 on Build GRCh38
rs770632747 Chr.7: 44104892-44104892 on Build GRCh38
rs759336775 Chr.7: 44104779-44104779 on Build GRCh38
rs750811336 Chr.7: 44104819-44104819 on Build GRCh38
rs909174059 Chr.7: 44104957-44104957 on Build GRCh38
rs758785378 Chr.7: 44104811-44104812 on Build GRCh38
rs777461260 Chr.7: 44104878-44104878 on Build GRCh38
rs780694276 Chr.7: 44104837-44104837 on Build GRCh38
rs530069122 Chr.7: 44104952-44104952 on Build GRCh38
rs760645622 Chr.7: 44104918-44104918 on Build GRCh38
rs1025828193 Chr.7: 44104824-44104824 on Build GRCh38
rs141126404 Chr.7: 44104808-44104808 on Build GRCh38
rs546918357 Chr.7: 44104811-44104811 on Build GRCh38
rs977787733 Chr.7: 44104965-44104965 on Build GRCh38
rs769397846 Chr.7: 44104866-44104866 on Build GRCh38
rs759368887 Chr.7: 44104904-44104904 on Build GRCh38
rs774219756 Chr.7: 44104767-44104767 on Build GRCh38
rs994227969 Chr.7: 44104822-44104822 on Build GRCh38
rs764204380 Chr.7: 44104797-44104797 on Build GRCh38
rs766049309 Chr.7: 44104764-44104764 on Build GRCh38
rs572753883 Chr.7: 44104902-44104902 on Build GRCh38
rs764118719 Chr.7: 44104924-44104924 on Build GRCh38
rs758758357 Chr.7: 44104835-44104835 on Build GRCh38
rs893407626 Chr.7: 44104790-44104790 on Build GRCh38
rs984395825 Chr.7: 44104870-44104870 on Build GRCh38
rs753935573 Chr.7: 44104947-44104947 on Build GRCh38
rs754028506 Chr.7: 44104807-44104807 on Build GRCh38
rs748847396 Chr.7: 44104879-44104879 on Build GRCh38
rs1013558383 Chr.7: 44104799-44104799 on Build GRCh38
rs917558654 Chr.7: 44104922-44104922 on Build GRCh38
rs559299616 Chr.7: 44104862-44104862 on Build GRCh38
rs771909045 Chr.7: 44104911-44104911 on Build GRCh38
rs767491556 Chr.7: 44104780-44104780 on Build GRCh38
rs752694956 Chr.7: 44104786-44104786 on Build GRCh38
rs201978706 Chr.7: 44104788-44104788 on Build GRCh38
rs971997979 Chr.7: 44104859-44104859 on Build GRCh38
rs775379801 Chr.7: 44104912-44104912 on Build GRCh38

View Full Product Details