Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73383500-73383743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA7
Entrez Gene ID
310
Gene Name
annexin A7
Gene Aliases
ANX7, SNX, SYNEXIN
UniGene
Hs.631827
Gene Chromosome Location
Chr.10: 73375101-73414085 on Build GRCh38
Transcripts
NM_001156.4 NM_001320879.1 NM_001320880.1 NM_004034.3 XM_017016162.1 XM_017016163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1685663 c.728delT p.L243fs*1 Chr.10: 73383662-73383662 on Build GRCh38
1627608 c.742G>A p.E248K Chr.10: 73383648-73383648 on Build GRCh38
5021108 c.776C>T p.T259M Chr.10: 73383614-73383614 on Build GRCh38
1603711 c.700-4A>G p.? Chr.10: 73383694-73383694 on Build GRCh38
1963712 c.775A>G p.T259A Chr.10: 73383615-73383615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751437176 Chr.10: 73383650-73383650 on Build GRCh38
rs769213073 Chr.10: 73383606-73383606 on Build GRCh38
rs367910001 Chr.10: 73383665-73383665 on Build GRCh38
rs780076617 Chr.10: 73383692-73383692 on Build GRCh38
rs552035180 Chr.10: 73383569-73383570 on Build GRCh38
rs200262714 Chr.10: 73383597-73383597 on Build GRCh38
rs747416168 Chr.10: 73383695-73383695 on Build GRCh38
rs779126828 Chr.10: 73383538-73383538 on Build GRCh38
rs777227063 Chr.10: 73383702-73383702 on Build GRCh38
rs779000183 Chr.10: 73383673-73383673 on Build GRCh38
rs201902462 Chr.10: 73383699-73383699 on Build GRCh38
rs757267292 Chr.10: 73383533-73383533 on Build GRCh38
rs187605730 Chr.10: 73383607-73383607 on Build GRCh38
rs745938968 Chr.10: 73383678-73383678 on Build GRCh38
rs562367196 Chr.10: 73383630-73383630 on Build GRCh38
rs947658264 Chr.10: 73383611-73383611 on Build GRCh38
rs762743606 Chr.10: 73383621-73383621 on Build GRCh38
rs772264554 Chr.10: 73383590-73383590 on Build GRCh38
rs199621428 Chr.10: 73383558-73383558 on Build GRCh38
rs746046245 Chr.10: 73383546-73383546 on Build GRCh38
rs745872465 Chr.10: 73383659-73383659 on Build GRCh38
rs558866290 Chr.10: 73383591-73383591 on Build GRCh38
rs754661103 Chr.10: 73383652-73383652 on Build GRCh38
rs142515398 Chr.10: 73383614-73383614 on Build GRCh38
rs758689567 Chr.10: 73383685-73383685 on Build GRCh38
rs113051613 Chr.10: 73383613-73383613 on Build GRCh38
rs372263988 Chr.10: 73383711-73383711 on Build GRCh38
rs767501517 Chr.10: 73383653-73383653 on Build GRCh38
rs772818416 Chr.10: 73383618-73383618 on Build GRCh38
rs1057111891 Chr.10: 73383676-73383676 on Build GRCh38
rs375873686 Chr.10: 73383664-73383664 on Build GRCh38
rs927346247 Chr.10: 73383627-73383627 on Build GRCh38
rs368588192 Chr.10: 73383572-73383572 on Build GRCh38
rs142756017 Chr.10: 73383564-73383564 on Build GRCh38

View Full Product Details